Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 31025023-31025521 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP4S1
Entrez Gene ID
11154
Gene Name
adaptor related protein complex 4 sigma 1 subunit
Gene Aliases
AP47B, CLA20, CLAPS4, CPSQ6, SPG52
UniGene
Hs.293411
Gene Chromosome Location
Chr.14: 31025106-31096450 on Build GRCh38
Transcripts
NM_001128126.2 NM_001254726.1 NM_001254727.1 NM_001254729.1 NM_007077.4 XM_011536371.2 NM_001083893.1 NM_014574.3 XM_005267569.3 XM_005267570.3

Gene Symbol
STRN3
Entrez Gene ID
29966
Gene Name
striatin 3
Gene Aliases
PPP2R6B, SG2NA
UniGene
Hs.401843
Gene Chromosome Location
Chr.14: 30893799-31026528 on Build GRCh38
Transcripts
NM_001128126.2 NM_001254726.1 NM_001254727.1 NM_001254729.1 NM_007077.4 XM_011536371.2 NM_001083893.1 NM_014574.3 XM_005267569.3 XM_005267570.3

SNP Detail

SNP ID Chromosome Location
rs963738481 Chr.14: 31025153-31025154 on Build GRCh38
rs990488971 Chr.14: 31025496-31025496 on Build GRCh38
rs566690585 Chr.14: 31025307-31025307 on Build GRCh38
rs894903517 Chr.14: 31025329-31025329 on Build GRCh38
rs912444411 Chr.14: 31025212-31025212 on Build GRCh38
rs962513220 Chr.14: 31025339-31025339 on Build GRCh38
rs947357641 Chr.14: 31025203-31025203 on Build GRCh38
rs112406943 Chr.14: 31025455-31025455 on Build GRCh38
rs936286205 Chr.14: 31025474-31025474 on Build GRCh38
rs555256918 Chr.14: 31025160-31025160 on Build GRCh38
rs766102911 Chr.14: 31025249-31025252 on Build GRCh38
rs113885370 Chr.14: 31025282-31025282 on Build GRCh38
rs11625235 Chr.14: 31025371-31025371 on Build GRCh38
rs928886891 Chr.14: 31025432-31025432 on Build GRCh38
rs179738 Chr.14: 31025284-31025284 on Build GRCh38
rs886664055 Chr.14: 31025381-31025381 on Build GRCh38
rs1016281336 Chr.14: 31025149-31025154 on Build GRCh38
rs568908450 Chr.14: 31025234-31025234 on Build GRCh38
rs978030538 Chr.14: 31025466-31025466 on Build GRCh38
rs555728241 Chr.14: 31025400-31025400 on Build GRCh38
rs551562684 Chr.14: 31025290-31025290 on Build GRCh38
rs1015736335 Chr.14: 31025303-31025303 on Build GRCh38
rs116515302 Chr.14: 31025054-31025054 on Build GRCh38
rs762250453 Chr.14: 31025167-31025170 on Build GRCh38
rs1029536448 Chr.14: 31025268-31025268 on Build GRCh38
rs963346227 Chr.14: 31025401-31025401 on Build GRCh38
rs1025616108 Chr.14: 31025098-31025098 on Build GRCh38
rs981597263 Chr.14: 31025056-31025056 on Build GRCh38
rs1005271351 Chr.14: 31025138-31025138 on Build GRCh38
rs1029000863 Chr.14: 31025430-31025430 on Build GRCh38
rs528919079 Chr.14: 31025192-31025192 on Build GRCh38
rs112266697 Chr.14: 31025101-31025101 on Build GRCh38
rs1005124748 Chr.14: 31025396-31025396 on Build GRCh38
rs934803743 Chr.14: 31025438-31025438 on Build GRCh38
rs928751086 Chr.14: 31025061-31025061 on Build GRCh38
rs775554692 Chr.14: 31025161-31025164 on Build GRCh38
rs925223582 Chr.14: 31025472-31025472 on Build GRCh38
rs916144537 Chr.14: 31025049-31025049 on Build GRCh38
rs981657759 Chr.14: 31025426-31025426 on Build GRCh38
rs776204016 Chr.14: 31025368-31025368 on Build GRCh38
rs75089409 Chr.14: 31025257-31025257 on Build GRCh38
rs1013793342 Chr.14: 31025084-31025084 on Build GRCh38
rs1013724402 Chr.14: 31025359-31025359 on Build GRCh38
rs924951899 Chr.14: 31025274-31025274 on Build GRCh38
rs966966595 Chr.14: 31025457-31025457 on Build GRCh38
rs150559645 Chr.14: 31025162-31025162 on Build GRCh38
rs1054871823 Chr.14: 31025324-31025324 on Build GRCh38
rs970294307 Chr.14: 31025404-31025404 on Build GRCh38
rs550643989 Chr.14: 31025207-31025207 on Build GRCh38
rs574765760 Chr.14: 31025445-31025445 on Build GRCh38
rs1046580155 Chr.14: 31025375-31025375 on Build GRCh38
rs996759889 Chr.14: 31025259-31025259 on Build GRCh38
rs534053025 Chr.14: 31025385-31025385 on Build GRCh38
rs1053586278 Chr.14: 31025147-31025147 on Build GRCh38
rs945325878 Chr.14: 31025256-31025256 on Build GRCh38
rs936359577 Chr.14: 31025278-31025278 on Build GRCh38

View Full Product Details