Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 49894291-49894800 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARF6
Entrez Gene ID
382
Gene Name
ADP ribosylation factor 6
Gene Aliases
-
UniGene
Hs.525330
Gene Chromosome Location
Chr.14: 49893018-49897054 on Build GRCh38
Transcripts
NM_001663.3

SNP Detail

SNP ID Chromosome Location
rs768722630 Chr.14: 49894572-49894573 on Build GRCh38
rs560814967 Chr.14: 49894648-49894648 on Build GRCh38
rs548311221 Chr.14: 49894343-49894343 on Build GRCh38
rs747046547 Chr.14: 49894364-49894364 on Build GRCh38
rs373961802 Chr.14: 49894452-49894452 on Build GRCh38
rs1046530644 Chr.14: 49894662-49894662 on Build GRCh38
rs181594154 Chr.14: 49894382-49894382 on Build GRCh38
rs15382 Chr.14: 49894620-49894620 on Build GRCh38
rs1052463598 Chr.14: 49894597-49894597 on Build GRCh38
rs760916651 Chr.14: 49894545-49894545 on Build GRCh38
rs888733764 Chr.14: 49894756-49894756 on Build GRCh38
rs1039313380 Chr.14: 49894436-49894436 on Build GRCh38
rs554347117 Chr.14: 49894530-49894530 on Build GRCh38
rs965534052 Chr.14: 49894406-49894406 on Build GRCh38
rs537919229 Chr.14: 49894513-49894513 on Build GRCh38
rs764412657 Chr.14: 49894646-49894646 on Build GRCh38
rs949516974 Chr.14: 49894625-49894625 on Build GRCh38
rs540226077 Chr.14: 49894551-49894551 on Build GRCh38
rs767570165 Chr.14: 49894346-49894346 on Build GRCh38
rs533916032 Chr.14: 49894350-49894350 on Build GRCh38
rs35848359 Chr.14: 49894474-49894474 on Build GRCh38
rs531794083 Chr.14: 49894332-49894332 on Build GRCh38
rs1003874953 Chr.14: 49894382-49894387 on Build GRCh38
rs976595705 Chr.14: 49894410-49894410 on Build GRCh38
rs368376617 Chr.14: 49894453-49894453 on Build GRCh38
rs566425091 Chr.14: 49894362-49894362 on Build GRCh38
rs910764521 Chr.14: 49894425-49894425 on Build GRCh38
rs562847169 Chr.14: 49894330-49894330 on Build GRCh38
rs985015448 Chr.14: 49894421-49894421 on Build GRCh38
rs575259203 Chr.14: 49894486-49894486 on Build GRCh38
rs913969287 Chr.14: 49894613-49894613 on Build GRCh38
rs112211624 Chr.14: 49894356-49894356 on Build GRCh38
rs942387627 Chr.14: 49894432-49894432 on Build GRCh38
rs978676335 Chr.14: 49894492-49894492 on Build GRCh38
rs138800769 Chr.14: 49894474-49894475 on Build GRCh38
rs371978931 Chr.14: 49894325-49894325 on Build GRCh38
rs15381 Chr.14: 49894699-49894699 on Build GRCh38
rs577569307 Chr.14: 49894660-49894660 on Build GRCh38
rs558678193 Chr.14: 49894440-49894440 on Build GRCh38
rs190349237 Chr.14: 49894345-49894345 on Build GRCh38
rs879247552 Chr.14: 49894454-49894456 on Build GRCh38
rs1026575178 Chr.14: 49894351-49894351 on Build GRCh38
rs936645810 Chr.14: 49894596-49894596 on Build GRCh38
rs61983787 Chr.14: 49894474-49894474 on Build GRCh38
rs756865603 Chr.14: 49894574-49894574 on Build GRCh38
rs1002420982 Chr.14: 49894716-49894716 on Build GRCh38
rs921117263 Chr.14: 49894431-49894431 on Build GRCh38
rs925896832 Chr.14: 49894494-49894494 on Build GRCh38
rs1048625048 Chr.14: 49894722-49894722 on Build GRCh38
rs933758010 Chr.14: 49894446-49894446 on Build GRCh38
rs1047023740 Chr.14: 49894336-49894336 on Build GRCh38
rs778068699 Chr.14: 49894435-49894435 on Build GRCh38
rs574521612 Chr.14: 49894532-49894532 on Build GRCh38
rs1015290520 Chr.14: 49894387-49894387 on Build GRCh38
rs546431753 Chr.14: 49894713-49894713 on Build GRCh38

View Full Product Details