Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 50005897-50006407 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC01588
Entrez Gene ID
283551
Gene Name
long intergenic non-protein coding RNA 1588
Gene Aliases
C14orf182
UniGene
Hs.292580
Gene Chromosome Location
Chr.14: 49981669-50007520 on Build GRCh38
Transcripts
NR_126496.1 NR_126497.1 NR_126498.1 NR_126500.1

SNP Detail

SNP ID Chromosome Location
rs902744355 Chr.14: 50006085-50006085 on Build GRCh38
rs8004022 Chr.14: 50005964-50005964 on Build GRCh38
rs11621806 Chr.14: 50006248-50006248 on Build GRCh38
rs11626611 Chr.14: 50006380-50006380 on Build GRCh38
rs973015428 Chr.14: 50006227-50006227 on Build GRCh38
rs80221861 Chr.14: 50006056-50006056 on Build GRCh38
rs961220426 Chr.14: 50005997-50005997 on Build GRCh38
rs185540715 Chr.14: 50006348-50006348 on Build GRCh38
rs527990861 Chr.14: 50005986-50005986 on Build GRCh38
rs1043973153 Chr.14: 50006069-50006069 on Build GRCh38
rs907238064 Chr.14: 50005993-50005993 on Build GRCh38
rs928608097 Chr.14: 50006354-50006354 on Build GRCh38
rs547326355 Chr.14: 50006065-50006065 on Build GRCh38
rs762923149 Chr.14: 50006037-50006037 on Build GRCh38
rs1029438079 Chr.14: 50006153-50006153 on Build GRCh38
rs148640745 Chr.14: 50006205-50006205 on Build GRCh38
rs555345894 Chr.14: 50006271-50006271 on Build GRCh38
rs528493065 Chr.14: 50006024-50006024 on Build GRCh38
rs979033161 Chr.14: 50006286-50006286 on Build GRCh38
rs571061587 Chr.14: 50006095-50006095 on Build GRCh38
rs757648199 Chr.14: 50005985-50005985 on Build GRCh38
rs112596506 Chr.14: 50006135-50006135 on Build GRCh38
rs996268189 Chr.14: 50006143-50006143 on Build GRCh38
rs561255928 Chr.14: 50006184-50006184 on Build GRCh38
rs559476261 Chr.14: 50005968-50005968 on Build GRCh38
rs887760994 Chr.14: 50006173-50006173 on Build GRCh38
rs553766889 Chr.14: 50006334-50006334 on Build GRCh38
rs898032223 Chr.14: 50006086-50006086 on Build GRCh38
rs949335765 Chr.14: 50005978-50005978 on Build GRCh38
rs11621844 Chr.14: 50006297-50006297 on Build GRCh38
rs927703638 Chr.14: 50006007-50006007 on Build GRCh38
rs997785522 Chr.14: 50006114-50006114 on Build GRCh38
rs1052289902 Chr.14: 50005922-50005922 on Build GRCh38
rs901964230 Chr.14: 50006115-50006115 on Build GRCh38
rs1034851281 Chr.14: 50006241-50006241 on Build GRCh38
rs960765873 Chr.14: 50006244-50006244 on Build GRCh38
rs972524815 Chr.14: 50006004-50006004 on Build GRCh38
rs887932239 Chr.14: 50006011-50006011 on Build GRCh38
rs779458428 Chr.14: 50006033-50006033 on Build GRCh38
rs569542931 Chr.14: 50006337-50006337 on Build GRCh38
rs569807683 Chr.14: 50006225-50006225 on Build GRCh38
rs935510224 Chr.14: 50006092-50006092 on Build GRCh38
rs1011101655 Chr.14: 50006255-50006255 on Build GRCh38
rs539626105 Chr.14: 50006128-50006128 on Build GRCh38
rs909742931 Chr.14: 50005988-50005988 on Build GRCh38
rs1016876593 Chr.14: 50006211-50006211 on Build GRCh38
rs142141087 Chr.14: 50006274-50006274 on Build GRCh38
rs1055230769 Chr.14: 50006031-50006031 on Build GRCh38
rs1015118314 Chr.14: 50006042-50006042 on Build GRCh38
rs7492428 Chr.14: 50005944-50005944 on Build GRCh38
rs1040978688 Chr.14: 50006349-50006349 on Build GRCh38
rs961241438 Chr.14: 50006224-50006224 on Build GRCh38

View Full Product Details