Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 50006444-50006945 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC01588
Entrez Gene ID
283551
Gene Name
long intergenic non-protein coding RNA 1588
Gene Aliases
C14orf182
UniGene
Hs.292580
Gene Chromosome Location
Chr.14: 49981669-50007520 on Build GRCh38
Transcripts
NR_126496.1 NR_126497.1 NR_126498.1 NR_126500.1

SNP Detail

SNP ID Chromosome Location
rs79477892 Chr.14: 50006635-50006635 on Build GRCh38
rs1030649796 Chr.14: 50006851-50006851 on Build GRCh38
rs979106743 Chr.14: 50006605-50006605 on Build GRCh38
rs7359055 Chr.14: 50006514-50006514 on Build GRCh38
rs76561243 Chr.14: 50006636-50006636 on Build GRCh38
rs991032233 Chr.14: 50006557-50006557 on Build GRCh38
rs200105071 Chr.14: 50006641-50006641 on Build GRCh38
rs796952095 Chr.14: 50006636-50006636 on Build GRCh38
rs1003354954 Chr.14: 50006773-50006773 on Build GRCh38
rs909219388 Chr.14: 50006669-50006669 on Build GRCh38
rs909333783 Chr.14: 50006704-50006704 on Build GRCh38
rs888764140 Chr.14: 50006495-50006495 on Build GRCh38
rs773026483 Chr.14: 50006889-50006889 on Build GRCh38
rs896359514 Chr.14: 50006718-50006718 on Build GRCh38
rs112291776 Chr.14: 50006513-50006514 on Build GRCh38
rs897809733 Chr.14: 50006710-50006710 on Build GRCh38
rs760797961 Chr.14: 50006905-50006905 on Build GRCh38
rs77644224 Chr.14: 50006513-50006513 on Build GRCh38
rs1030603635 Chr.14: 50006487-50006487 on Build GRCh38
rs959240920 Chr.14: 50006796-50006796 on Build GRCh38
rs1056395944 Chr.14: 50006716-50006716 on Build GRCh38
rs897107583 Chr.14: 50006500-50006500 on Build GRCh38
rs1053378805 Chr.14: 50006720-50006720 on Build GRCh38
rs6572636 Chr.14: 50006686-50006686 on Build GRCh38
rs1003262187 Chr.14: 50006715-50006715 on Build GRCh38
rs7359046 Chr.14: 50006654-50006654 on Build GRCh38
rs6572638 Chr.14: 50006779-50006779 on Build GRCh38
rs553787669 Chr.14: 50006700-50006700 on Build GRCh38
rs189617899 Chr.14: 50006884-50006884 on Build GRCh38
rs970611274 Chr.14: 50006612-50006612 on Build GRCh38
rs967806257 Chr.14: 50006755-50006755 on Build GRCh38
rs7359045 Chr.14: 50006521-50006521 on Build GRCh38
rs6572637 Chr.14: 50006726-50006726 on Build GRCh38
rs1020731260 Chr.14: 50006756-50006756 on Build GRCh38
rs992550753 Chr.14: 50006644-50006644 on Build GRCh38
rs970897599 Chr.14: 50006769-50006769 on Build GRCh38
rs549972497 Chr.14: 50006717-50006717 on Build GRCh38
rs923602872 Chr.14: 50006615-50006615 on Build GRCh38
rs776580686 Chr.14: 50006557-50006557 on Build GRCh38
rs1023422957 Chr.14: 50006749-50006749 on Build GRCh38
rs140365749 Chr.14: 50006708-50006708 on Build GRCh38
rs926532974 Chr.14: 50006621-50006622 on Build GRCh38
rs976345205 Chr.14: 50006621-50006621 on Build GRCh38
rs994084208 Chr.14: 50006512-50006512 on Build GRCh38
rs937931069 Chr.14: 50006642-50006642 on Build GRCh38
rs1007149549 Chr.14: 50006497-50006497 on Build GRCh38
rs577563306 Chr.14: 50006873-50006873 on Build GRCh38
rs11332873 Chr.14: 50006622-50006622 on Build GRCh38
rs935611377 Chr.14: 50006617-50006617 on Build GRCh38
rs1050999563 Chr.14: 50006696-50006696 on Build GRCh38
rs756392480 Chr.14: 50006483-50006483 on Build GRCh38
rs7148405 Chr.14: 50006880-50006880 on Build GRCh38
rs533617601 Chr.14: 50006709-50006709 on Build GRCh38
rs555407355 Chr.14: 50006815-50006815 on Build GRCh38
rs561685200 Chr.14: 50006579-50006579 on Build GRCh38
rs967732540 Chr.14: 50006585-50006585 on Build GRCh38

View Full Product Details