Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 57268500-57268988 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP5M1
Entrez Gene ID
55745
Gene Name
adaptor related protein complex 5 mu 1 subunit
Gene Aliases
C14orf108, MUDENG, Mu5, MuD
UniGene
Hs.597349
Gene Chromosome Location
Chr.14: 57268888-57298742 on Build GRCh38
Transcripts
NM_018229.3 NR_026895.1 XM_006720196.2 XM_011536940.2 NM_006544.3 XM_005267272.3

Gene Symbol
EXOC5
Entrez Gene ID
10640
Gene Name
exocyst complex component 5
Gene Aliases
HSEC10, PRO1912, SEC10, SEC10L1, SEC10P
UniGene
Hs.743987
Gene Chromosome Location
Chr.14: 57202476-57268899 on Build GRCh38
Transcripts
NM_018229.3 NR_026895.1 XM_006720196.2 XM_011536940.2 NM_006544.3 XM_005267272.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6417491 c.14C>T p.A5V Chr.14: 57268635-57268635 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1011229290 Chr.14: 57268529-57268529 on Build GRCh38
rs774306711 Chr.14: 57268616-57268616 on Build GRCh38
rs548872646 Chr.14: 57268927-57268927 on Build GRCh38
rs904057726 Chr.14: 57268838-57268838 on Build GRCh38
rs552343604 Chr.14: 57268725-57268725 on Build GRCh38
rs992572746 Chr.14: 57268722-57268722 on Build GRCh38
rs1055262831 Chr.14: 57268844-57268844 on Build GRCh38
rs750748852 Chr.14: 57268656-57268656 on Build GRCh38
rs150717393 Chr.14: 57268588-57268589 on Build GRCh38
rs915915687 Chr.14: 57268852-57268852 on Build GRCh38
rs1013808811 Chr.14: 57268849-57268849 on Build GRCh38
rs1028269129 Chr.14: 57268535-57268535 on Build GRCh38
rs770735817 Chr.14: 57268692-57268692 on Build GRCh38
rs749047122 Chr.14: 57268688-57268688 on Build GRCh38
rs752365304 Chr.14: 57268639-57268639 on Build GRCh38
rs945802801 Chr.14: 57268827-57268827 on Build GRCh38
rs570342595 Chr.14: 57268948-57268948 on Build GRCh38
rs948981449 Chr.14: 57268702-57268702 on Build GRCh38
rs756754782 Chr.14: 57268953-57268953 on Build GRCh38
rs1001139858 Chr.14: 57268691-57268691 on Build GRCh38
rs914338393 Chr.14: 57268887-57268887 on Build GRCh38
rs952509326 Chr.14: 57268956-57268956 on Build GRCh38
rs961516341 Chr.14: 57268797-57268797 on Build GRCh38
rs928892597 Chr.14: 57268869-57268869 on Build GRCh38
rs752157595 Chr.14: 57268669-57268669 on Build GRCh38
rs976670814 Chr.14: 57268914-57268914 on Build GRCh38
rs34658299 Chr.14: 57268812-57268813 on Build GRCh38
rs762752478 Chr.14: 57268589-57268589 on Build GRCh38
rs961195116 Chr.14: 57268651-57268651 on Build GRCh38
rs963217910 Chr.14: 57268565-57268565 on Build GRCh38
rs207474873 Chr.14: 57268771-57268771 on Build GRCh38
rs889477937 Chr.14: 57268586-57268586 on Build GRCh38
rs748902493 Chr.14: 57268932-57268932 on Build GRCh38
rs781668380 Chr.14: 57268544-57268544 on Build GRCh38
rs777510477 Chr.14: 57268882-57268882 on Build GRCh38
rs763818511 Chr.14: 57268655-57268655 on Build GRCh38
rs931134681 Chr.14: 57268758-57268758 on Build GRCh38
rs558579506 Chr.14: 57268963-57268963 on Build GRCh38
rs994001306 Chr.14: 57268801-57268801 on Build GRCh38
rs934849698 Chr.14: 57268875-57268875 on Build GRCh38
rs1042868416 Chr.14: 57268924-57268924 on Build GRCh38
rs931248989 Chr.14: 57268521-57268521 on Build GRCh38
rs1041388585 Chr.14: 57268935-57268935 on Build GRCh38
rs760313636 Chr.14: 57268652-57268652 on Build GRCh38
rs895398205 Chr.14: 57268845-57268845 on Build GRCh38
rs1008149668 Chr.14: 57268784-57268784 on Build GRCh38
rs891541233 Chr.14: 57268664-57268664 on Build GRCh38
rs770792787 Chr.14: 57268613-57268613 on Build GRCh38
rs892869608 Chr.14: 57268727-57268727 on Build GRCh38
rs889471254 Chr.14: 57268918-57268918 on Build GRCh38
rs780822884 Chr.14: 57268596-57268608 on Build GRCh38
rs752309234 Chr.14: 57268688-57268688 on Build GRCh38
rs781450642 Chr.14: 57268577-57268577 on Build GRCh38
rs1041114758 Chr.14: 57268811-57268811 on Build GRCh38
rs773051689 Chr.14: 57268588-57268588 on Build GRCh38
rs547676439 Chr.14: 57268816-57268816 on Build GRCh38
rs777979767 Chr.14: 57268686-57268686 on Build GRCh38
rs928816291 Chr.14: 57268704-57268704 on Build GRCh38
rs972769981 Chr.14: 57268728-57268728 on Build GRCh38
rs1001033987 Chr.14: 57268843-57268843 on Build GRCh38
rs536560060 Chr.14: 57268915-57268915 on Build GRCh38
rs774251179 Chr.14: 57268696-57268696 on Build GRCh38
rs981675131 Chr.14: 57268861-57268861 on Build GRCh38
rs952156379 Chr.14: 57268834-57268834 on Build GRCh38
rs1022979304 Chr.14: 57268969-57268969 on Build GRCh38
rs768637069 Chr.14: 57268576-57268576 on Build GRCh38
rs766985070 Chr.14: 57268634-57268634 on Build GRCh38
rs748356797 Chr.14: 57268581-57268581 on Build GRCh38
rs911024082 Chr.14: 57268886-57268886 on Build GRCh38
rs769852391 Chr.14: 57268675-57268675 on Build GRCh38
rs947126342 Chr.14: 57268894-57268894 on Build GRCh38
rs943929702 Chr.14: 57268788-57268788 on Build GRCh38
rs934819467 Chr.14: 57268724-57268724 on Build GRCh38
rs780655303 Chr.14: 57268665-57268665 on Build GRCh38
rs970008498 Chr.14: 57268855-57268855 on Build GRCh38
rs1045923020 Chr.14: 57268703-57268703 on Build GRCh38
rs755815627 Chr.14: 57268846-57268846 on Build GRCh38
rs565273999 Chr.14: 57268674-57268674 on Build GRCh38
rs959442049 Chr.14: 57268717-57268717 on Build GRCh38
rs965685452 Chr.14: 57268900-57268900 on Build GRCh38
rs1049414321 Chr.14: 57268757-57268757 on Build GRCh38
rs537079690 Chr.14: 57268952-57268952 on Build GRCh38
rs566223693 Chr.14: 57268826-57268826 on Build GRCh38
rs769623800 Chr.14: 57268585-57268585 on Build GRCh38
rs961773148 Chr.14: 57268726-57268727 on Build GRCh38
rs911126609 Chr.14: 57268778-57268778 on Build GRCh38
rs1019518819 Chr.14: 57268792-57268792 on Build GRCh38
rs755051718 Chr.14: 57268673-57268673 on Build GRCh38
rs1027017839 Chr.14: 57268864-57268864 on Build GRCh38
rs758998153 Chr.14: 57268618-57268618 on Build GRCh38
rs1011284191 Chr.14: 57268738-57268738 on Build GRCh38

View Full Product Details