Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 57268884-57269386 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP5M1
Entrez Gene ID
55745
Gene Name
adaptor related protein complex 5 mu 1 subunit
Gene Aliases
C14orf108, MUDENG, Mu5, MuD
UniGene
Hs.597349
Gene Chromosome Location
Chr.14: 57268888-57298742 on Build GRCh38
Transcripts
NM_018229.3 NR_026895.1 XM_006720196.2 XM_011536940.2 NM_006544.3

Gene Symbol
EXOC5
Entrez Gene ID
10640
Gene Name
exocyst complex component 5
Gene Aliases
HSEC10, PRO1912, SEC10, SEC10L1, SEC10P
UniGene
Hs.743987
Gene Chromosome Location
Chr.14: 57202476-57268899 on Build GRCh38
Transcripts
NM_018229.3 NR_026895.1 XM_006720196.2 XM_011536940.2 NM_006544.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7179336 c.34G>A p.E12K Chr.14: 57269348-57269348 on Build GRCh38
6271596 c.34G>T p.E12* Chr.14: 57269348-57269348 on Build GRCh38
3386592 c.17T>G p.V6G Chr.14: 57269331-57269331 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1018345480 Chr.14: 57269259-57269259 on Build GRCh38
rs548872646 Chr.14: 57268927-57268927 on Build GRCh38
rs929914185 Chr.14: 57269106-57269106 on Build GRCh38
rs768327293 Chr.14: 57269359-57269359 on Build GRCh38
rs916764177 Chr.14: 57269154-57269154 on Build GRCh38
rs947510856 Chr.14: 57269033-57269033 on Build GRCh38
rs375804884 Chr.14: 57269296-57269296 on Build GRCh38
rs763383170 Chr.14: 57269309-57269309 on Build GRCh38
rs114716822 Chr.14: 57269018-57269018 on Build GRCh38
rs34722917 Chr.14: 57269330-57269330 on Build GRCh38
rs570342595 Chr.14: 57268948-57268948 on Build GRCh38
rs756754782 Chr.14: 57268953-57268953 on Build GRCh38
rs1006898947 Chr.14: 57268988-57268988 on Build GRCh38
rs753546606 Chr.14: 57269324-57269324 on Build GRCh38
rs952509326 Chr.14: 57268956-57268956 on Build GRCh38
rs760517302 Chr.14: 57269023-57269023 on Build GRCh38
rs565465442 Chr.14: 57269051-57269051 on Build GRCh38
rs976670814 Chr.14: 57268914-57268914 on Build GRCh38
rs746454257 Chr.14: 57269354-57269354 on Build GRCh38
rs775258434 Chr.14: 57269104-57269104 on Build GRCh38
rs766885880 Chr.14: 57269314-57269314 on Build GRCh38
rs748902493 Chr.14: 57268932-57268932 on Build GRCh38
rs756253018 Chr.14: 57269329-57269329 on Build GRCh38
rs965220928 Chr.14: 57269007-57269007 on Build GRCh38
rs943707440 Chr.14: 57269059-57269059 on Build GRCh38
rs558579506 Chr.14: 57268963-57268963 on Build GRCh38
rs1048765992 Chr.14: 57269182-57269182 on Build GRCh38
rs888422022 Chr.14: 57269217-57269218 on Build GRCh38
rs752104842 Chr.14: 57269315-57269315 on Build GRCh38
rs1012023693 Chr.14: 57269201-57269201 on Build GRCh38
rs914746488 Chr.14: 57269031-57269031 on Build GRCh38
rs1042868416 Chr.14: 57268924-57268924 on Build GRCh38
rs763799904 Chr.14: 57269302-57269302 on Build GRCh38
rs140243229 Chr.14: 57269196-57269196 on Build GRCh38
rs138148216 Chr.14: 57269344-57269344 on Build GRCh38
rs778345161 Chr.14: 57269302-57269303 on Build GRCh38
rs1041388585 Chr.14: 57268935-57268935 on Build GRCh38
rs181867118 Chr.14: 57269060-57269060 on Build GRCh38
rs778748769 Chr.14: 57269338-57269338 on Build GRCh38
rs889471254 Chr.14: 57268918-57268918 on Build GRCh38
rs760235201 Chr.14: 57269286-57269286 on Build GRCh38
rs1003322657 Chr.14: 57269250-57269250 on Build GRCh38
rs768449392 Chr.14: 57269215-57269215 on Build GRCh38
rs981390418 Chr.14: 57268986-57268986 on Build GRCh38
rs1032702857 Chr.14: 57269361-57269361 on Build GRCh38
rs200149814 Chr.14: 57269331-57269331 on Build GRCh38
rs943759975 Chr.14: 57269350-57269350 on Build GRCh38
rs7152744 Chr.14: 57269236-57269236 on Build GRCh38
rs536560060 Chr.14: 57268915-57268915 on Build GRCh38
rs756572168 Chr.14: 57269300-57269301 on Build GRCh38
rs757015812 Chr.14: 57269335-57269335 on Build GRCh38
rs930194690 Chr.14: 57269207-57269207 on Build GRCh38
rs1022979304 Chr.14: 57268969-57268969 on Build GRCh38
rs745371920 Chr.14: 57269049-57269049 on Build GRCh38
rs970063075 Chr.14: 57268985-57268985 on Build GRCh38
rs745806872 Chr.14: 57269343-57269343 on Build GRCh38
rs937091920 Chr.14: 57269130-57269130 on Build GRCh38
rs925582717 Chr.14: 57269113-57269113 on Build GRCh38
rs991236012 Chr.14: 57269144-57269144 on Build GRCh38
rs897180138 Chr.14: 57268972-57268972 on Build GRCh38
rs753565782 Chr.14: 57269304-57269304 on Build GRCh38
rs563275698 Chr.14: 57269256-57269256 on Build GRCh38
rs910961477 Chr.14: 57269056-57269056 on Build GRCh38
rs775527320 Chr.14: 57269353-57269353 on Build GRCh38
rs537079690 Chr.14: 57268952-57268952 on Build GRCh38
rs148345093 Chr.14: 57268984-57268984 on Build GRCh38
rs994125009 Chr.14: 57268980-57268980 on Build GRCh38
rs749569877 Chr.14: 57269361-57269369 on Build GRCh38
rs778311795 Chr.14: 57268987-57268987 on Build GRCh38
rs749562779 Chr.14: 57269333-57269333 on Build GRCh38
rs985219986 Chr.14: 57269054-57269054 on Build GRCh38
rs774803819 Chr.14: 57269284-57269284 on Build GRCh38
rs745798809 Chr.14: 57268990-57268990 on Build GRCh38
rs538144687 Chr.14: 57269320-57269320 on Build GRCh38
rs978434283 Chr.14: 57269080-57269080 on Build GRCh38

View Full Product Details