Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 57269263-57269754 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP5M1
Entrez Gene ID
55745
Gene Name
adaptor related protein complex 5 mu 1 subunit
Gene Aliases
C14orf108, MUDENG, Mu5, MuD
UniGene
Hs.597349
Gene Chromosome Location
Chr.14: 57268888-57298742 on Build GRCh38
Transcripts
NM_018229.3 NR_026895.1 XM_006720196.2 XM_011536940.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7179336 c.34G>A p.E12K Chr.14: 57269348-57269348 on Build GRCh38
6271596 c.34G>T p.E12* Chr.14: 57269348-57269348 on Build GRCh38
3386592 c.17T>G p.V6G Chr.14: 57269331-57269331 on Build GRCh38
5053995 c.74G>A p.R25K Chr.14: 57269388-57269388 on Build GRCh38
5448671 c.74+3A>C p.? Chr.14: 57269391-57269391 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs888475962 Chr.14: 57269594-57269594 on Build GRCh38
rs1044870088 Chr.14: 57269540-57269540 on Build GRCh38
rs759952904 Chr.14: 57269378-57269378 on Build GRCh38
rs998649864 Chr.14: 57269630-57269630 on Build GRCh38
rs768327293 Chr.14: 57269359-57269359 on Build GRCh38
rs766834609 Chr.14: 57269374-57269374 on Build GRCh38
rs754057145 Chr.14: 57269428-57269428 on Build GRCh38
rs375804884 Chr.14: 57269296-57269296 on Build GRCh38
rs763383170 Chr.14: 57269309-57269309 on Build GRCh38
rs34722917 Chr.14: 57269330-57269330 on Build GRCh38
rs753546606 Chr.14: 57269324-57269324 on Build GRCh38
rs767874845 Chr.14: 57269398-57269398 on Build GRCh38
rs746454257 Chr.14: 57269354-57269354 on Build GRCh38
rs891806092 Chr.14: 57269609-57269609 on Build GRCh38
rs753269919 Chr.14: 57269489-57269489 on Build GRCh38
rs756485272 Chr.14: 57269518-57269518 on Build GRCh38
rs766885880 Chr.14: 57269314-57269314 on Build GRCh38
rs745608208 Chr.14: 57269435-57269435 on Build GRCh38
rs756253018 Chr.14: 57269329-57269329 on Build GRCh38
rs949798357 Chr.14: 57269473-57269473 on Build GRCh38
rs534235047 Chr.14: 57269703-57269703 on Build GRCh38
rs1032923649 Chr.14: 57269701-57269701 on Build GRCh38
rs758235461 Chr.14: 57269439-57269439 on Build GRCh38
rs1002959853 Chr.14: 57269591-57269591 on Build GRCh38
rs150326144 Chr.14: 57269536-57269536 on Build GRCh38
rs752104842 Chr.14: 57269315-57269315 on Build GRCh38
rs763799904 Chr.14: 57269302-57269302 on Build GRCh38
rs918277117 Chr.14: 57269422-57269422 on Build GRCh38
rs138148216 Chr.14: 57269344-57269344 on Build GRCh38
rs778345161 Chr.14: 57269302-57269303 on Build GRCh38
rs775998464 Chr.14: 57269424-57269424 on Build GRCh38
rs1010640150 Chr.14: 57269620-57269620 on Build GRCh38
rs554535090 Chr.14: 57269455-57269455 on Build GRCh38
rs778748769 Chr.14: 57269338-57269338 on Build GRCh38
rs958347987 Chr.14: 57269386-57269386 on Build GRCh38
rs965906679 Chr.14: 57269704-57269704 on Build GRCh38
rs760235201 Chr.14: 57269286-57269286 on Build GRCh38
rs976542968 Chr.14: 57269716-57269716 on Build GRCh38
rs1036158121 Chr.14: 57269593-57269593 on Build GRCh38
rs1032702857 Chr.14: 57269361-57269361 on Build GRCh38
rs200149814 Chr.14: 57269331-57269331 on Build GRCh38
rs199518257 Chr.14: 57269373-57269373 on Build GRCh38
rs943759975 Chr.14: 57269350-57269350 on Build GRCh38
rs774341000 Chr.14: 57269420-57269420 on Build GRCh38
rs374122487 Chr.14: 57269400-57269400 on Build GRCh38
rs764587546 Chr.14: 57269685-57269685 on Build GRCh38
rs942707513 Chr.14: 57269598-57269598 on Build GRCh38
rs1021908554 Chr.14: 57269626-57269626 on Build GRCh38
rs757015812 Chr.14: 57269335-57269335 on Build GRCh38
rs756572168 Chr.14: 57269300-57269301 on Build GRCh38
rs547835264 Chr.14: 57269556-57269556 on Build GRCh38
rs745806872 Chr.14: 57269343-57269343 on Build GRCh38
rs756165870 Chr.14: 57269404-57269404 on Build GRCh38
rs764198621 Chr.14: 57269418-57269418 on Build GRCh38
rs918318870 Chr.14: 57269717-57269717 on Build GRCh38
rs530346103 Chr.14: 57269643-57269643 on Build GRCh38
rs753565782 Chr.14: 57269304-57269304 on Build GRCh38
rs1020148285 Chr.14: 57269635-57269635 on Build GRCh38
rs775527320 Chr.14: 57269353-57269353 on Build GRCh38
rs760085817 Chr.14: 57269433-57269433 on Build GRCh38
rs111628238 Chr.14: 57269469-57269469 on Build GRCh38
rs929603415 Chr.14: 57269436-57269436 on Build GRCh38
rs559719524 Chr.14: 57269639-57269639 on Build GRCh38
rs749569877 Chr.14: 57269361-57269369 on Build GRCh38
rs878887169 Chr.14: 57269520-57269520 on Build GRCh38
rs749562779 Chr.14: 57269333-57269333 on Build GRCh38
rs538144687 Chr.14: 57269320-57269320 on Build GRCh38
rs574333380 Chr.14: 57269463-57269463 on Build GRCh38

View Full Product Details