Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 68874424-68874920 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTN1
Entrez Gene ID
87
Gene Name
actinin alpha 1
Gene Aliases
BDPLT15
UniGene
Hs.235750
Gene Chromosome Location
Chr.14: 68874123-68979366 on Build GRCh38
Transcripts
NM_001102.3 NM_001130004.1 NM_001130005.1 XM_011537265.2 XM_011537266.2 XM_011537267.2 XM_011537268.2 XM_017021720.1 XM_017021721.1 XM_017021722.1 XM_017021723.1 XM_017021725.1 XM_017021726.1 XM_017021727.1 XM_017021728.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6592992 c.2737G>A p.D913N Chr.14: 68874867-68874867 on Build GRCh38
7354088 c.2662G>A p.G888S Chr.14: 68874876-68874876 on Build GRCh38
4659128 c.2665G>A p.E889K Chr.14: 68874873-68874873 on Build GRCh38
4659130 c.2651C>T p.T884M Chr.14: 68874887-68874887 on Build GRCh38
6592966 c.2720C>T p.A907V Chr.14: 68874884-68874884 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1052417072 Chr.14: 68874694-68874694 on Build GRCh38
rs578254736 Chr.14: 68874736-68874736 on Build GRCh38
rs1013902422 Chr.14: 68874499-68874499 on Build GRCh38
rs541725585 Chr.14: 68874682-68874682 on Build GRCh38
rs111795768 Chr.14: 68874862-68874862 on Build GRCh38
rs1013518228 Chr.14: 68874817-68874817 on Build GRCh38
rs541262717 Chr.14: 68874892-68874892 on Build GRCh38
rs774807508 Chr.14: 68874851-68874851 on Build GRCh38
rs758983643 Chr.14: 68874846-68874846 on Build GRCh38
rs11557771 Chr.14: 68874526-68874526 on Build GRCh38
rs113418960 Chr.14: 68874531-68874531 on Build GRCh38
rs937362483 Chr.14: 68874667-68874669 on Build GRCh38
rs1025278134 Chr.14: 68874819-68874819 on Build GRCh38
rs545396188 Chr.14: 68874836-68874836 on Build GRCh38
rs12237 Chr.14: 68874483-68874483 on Build GRCh38
rs974918917 Chr.14: 68874670-68874670 on Build GRCh38
rs753420287 Chr.14: 68874810-68874810 on Build GRCh38
rs765483044 Chr.14: 68874802-68874802 on Build GRCh38
rs759103863 Chr.14: 68874877-68874877 on Build GRCh38
rs370350860 Chr.14: 68874840-68874840 on Build GRCh38
rs764889893 Chr.14: 68874884-68874884 on Build GRCh38
rs760633640 Chr.14: 68874786-68874786 on Build GRCh38
rs996505111 Chr.14: 68874820-68874820 on Build GRCh38
rs745956137 Chr.14: 68874839-68874839 on Build GRCh38
rs762175698 Chr.14: 68874854-68874854 on Build GRCh38
rs955084884 Chr.14: 68874853-68874853 on Build GRCh38
rs754618872 Chr.14: 68874824-68874824 on Build GRCh38
rs1024661306 Chr.14: 68874566-68874566 on Build GRCh38
rs553739767 Chr.14: 68874707-68874707 on Build GRCh38
rs752368324 Chr.14: 68874899-68874899 on Build GRCh38
rs777665467 Chr.14: 68874828-68874828 on Build GRCh38
rs14582 Chr.14: 68874761-68874761 on Build GRCh38
rs71770347 Chr.14: 68874487-68874487 on Build GRCh38
rs933603867 Chr.14: 68874678-68874678 on Build GRCh38
rs1015876409 Chr.14: 68874847-68874847 on Build GRCh38
rs1055886127 Chr.14: 68874667-68874670 on Build GRCh38
rs758246893 Chr.14: 68874826-68874826 on Build GRCh38
rs1033705565 Chr.14: 68874801-68874801 on Build GRCh38
rs941263059 Chr.14: 68874614-68874614 on Build GRCh38
rs192640536 Chr.14: 68874876-68874876 on Build GRCh38
rs182250143 Chr.14: 68874574-68874574 on Build GRCh38
rs922397645 Chr.14: 68874489-68874489 on Build GRCh38
rs547011909 Chr.14: 68874837-68874837 on Build GRCh38
rs773076904 Chr.14: 68874756-68874756 on Build GRCh38
rs933820257 Chr.14: 68874507-68874507 on Build GRCh38
rs757804466 Chr.14: 68874590-68874591 on Build GRCh38
rs746883304 Chr.14: 68874834-68874834 on Build GRCh38
rs1041886151 Chr.14: 68874744-68874744 on Build GRCh38
rs149327982 Chr.14: 68874827-68874827 on Build GRCh38
rs1043074779 Chr.14: 68874708-68874708 on Build GRCh38
rs945997783 Chr.14: 68874703-68874703 on Build GRCh38
rs913238190 Chr.14: 68874696-68874696 on Build GRCh38
rs922092956 Chr.14: 68874677-68874677 on Build GRCh38
rs1038704241 Chr.14: 68874630-68874630 on Build GRCh38
rs35738337 Chr.14: 68874737-68874738 on Build GRCh38
rs76521838 Chr.14: 68874486-68874486 on Build GRCh38
rs372486596 Chr.14: 68874825-68874825 on Build GRCh38
rs1000439473 Chr.14: 68874799-68874799 on Build GRCh38
rs983229235 Chr.14: 68874590-68874590 on Build GRCh38
rs1014352230 Chr.14: 68874711-68874711 on Build GRCh38
rs77044396 Chr.14: 68874487-68874487 on Build GRCh38
rs781078800 Chr.14: 68874838-68874838 on Build GRCh38
rs1033503556 Chr.14: 68874461-68874461 on Build GRCh38
rs768868163 Chr.14: 68874843-68874843 on Build GRCh38
rs920593877 Chr.14: 68874652-68874652 on Build GRCh38
rs749558617 Chr.14: 68874842-68874842 on Build GRCh38
rs962888664 Chr.14: 68874880-68874880 on Build GRCh38
rs375245539 Chr.14: 68874856-68874856 on Build GRCh38
rs903363952 Chr.14: 68874751-68874751 on Build GRCh38
rs188009772 Chr.14: 68874644-68874644 on Build GRCh38
rs3209273 Chr.14: 68874568-68874568 on Build GRCh38
rs959304247 Chr.14: 68874481-68874481 on Build GRCh38
rs1001606062 Chr.14: 68874737-68874737 on Build GRCh38
rs529075691 Chr.14: 68874604-68874604 on Build GRCh38

View Full Product Details