Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 68874822-68875320 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTN1
Entrez Gene ID
87
Gene Name
actinin alpha 1
Gene Aliases
BDPLT15
UniGene
Hs.235750
Gene Chromosome Location
Chr.14: 68874123-68979366 on Build GRCh38
Transcripts
NM_001102.3 NM_001130004.1 NM_001130005.1 XM_011537265.2 XM_011537266.2 XM_011537267.2 XM_011537268.2 XM_017021720.1 XM_017021721.1 XM_017021722.1 XM_017021723.1 XM_017021725.1 XM_017021726.1 XM_017021727.1 XM_017021728.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7354088 c.2662G>A p.G888S Chr.14: 68874876-68874876 on Build GRCh38
331776 c.2549G>C p.R850P Chr.14: 68874989-68874989 on Build GRCh38
1370749 c.2546G>A p.R849H Chr.14: 68874992-68874992 on Build GRCh38
4899190 c.2682C>T p.S894S Chr.14: 68874922-68874922 on Build GRCh38
4659128 c.2665G>A p.E889K Chr.14: 68874873-68874873 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756160188 Chr.14: 68874929-68874929 on Build GRCh38
rs535198159 Chr.14: 68875159-68875159 on Build GRCh38
rs2070278 Chr.14: 68875195-68875195 on Build GRCh38
rs201818845 Chr.14: 68875029-68875029 on Build GRCh38
rs113447035 Chr.14: 68875020-68875020 on Build GRCh38
rs567845825 Chr.14: 68875127-68875127 on Build GRCh38
rs752730807 Chr.14: 68875062-68875062 on Build GRCh38
rs148344567 Chr.14: 68874990-68874990 on Build GRCh38
rs772838152 Chr.14: 68875102-68875102 on Build GRCh38
rs771575455 Chr.14: 68874989-68874989 on Build GRCh38
rs767263355 Chr.14: 68874994-68874994 on Build GRCh38
rs766310248 Chr.14: 68875005-68875005 on Build GRCh38
rs371440985 Chr.14: 68874992-68874992 on Build GRCh38
rs772926735 Chr.14: 68875038-68875038 on Build GRCh38
rs751804758 Chr.14: 68875139-68875139 on Build GRCh38
rs374861899 Chr.14: 68875027-68875027 on Build GRCh38
rs750932524 Chr.14: 68875069-68875069 on Build GRCh38
rs759103863 Chr.14: 68874877-68874877 on Build GRCh38
rs780057510 Chr.14: 68874968-68874968 on Build GRCh38
rs370350860 Chr.14: 68874840-68874840 on Build GRCh38
rs199901015 Chr.14: 68874953-68874953 on Build GRCh38
rs1003873380 Chr.14: 68875292-68875294 on Build GRCh38
rs141668847 Chr.14: 68874904-68874904 on Build GRCh38
rs745956137 Chr.14: 68874839-68874839 on Build GRCh38
rs769389947 Chr.14: 68874944-68874944 on Build GRCh38
rs780144853 Chr.14: 68874932-68874932 on Build GRCh38
rs866338070 Chr.14: 68875046-68875046 on Build GRCh38
rs762945522 Chr.14: 68875055-68875056 on Build GRCh38
rs977533635 Chr.14: 68875162-68875162 on Build GRCh38
rs1015876409 Chr.14: 68874847-68874847 on Build GRCh38
rs761305316 Chr.14: 68874943-68874943 on Build GRCh38
rs775561996 Chr.14: 68875117-68875117 on Build GRCh38
rs771727287 Chr.14: 68875035-68875035 on Build GRCh38
rs756601539 Chr.14: 68875100-68875100 on Build GRCh38
rs756604906 Chr.14: 68875097-68875097 on Build GRCh38
rs569671786 Chr.14: 68875061-68875061 on Build GRCh38
rs747818083 Chr.14: 68875216-68875216 on Build GRCh38
rs924726466 Chr.14: 68875182-68875182 on Build GRCh38
rs770672420 Chr.14: 68875105-68875105 on Build GRCh38
rs917362007 Chr.14: 68875260-68875260 on Build GRCh38
rs763107679 Chr.14: 68875123-68875123 on Build GRCh38
rs764313700 Chr.14: 68875124-68875124 on Build GRCh38
rs900027830 Chr.14: 68875205-68875205 on Build GRCh38
rs751345444 Chr.14: 68875101-68875111 on Build GRCh38
rs748801956 Chr.14: 68875126-68875126 on Build GRCh38
rs781454018 Chr.14: 68875086-68875086 on Build GRCh38
rs917241748 Chr.14: 68875156-68875156 on Build GRCh38
rs367686061 Chr.14: 68874971-68874971 on Build GRCh38
rs143416481 Chr.14: 68874967-68874967 on Build GRCh38
rs755087579 Chr.14: 68874976-68874976 on Build GRCh38
rs147023729 Chr.14: 68874933-68874933 on Build GRCh38
rs955053748 Chr.14: 68875067-68875067 on Build GRCh38
rs757585091 Chr.14: 68874945-68874945 on Build GRCh38
rs199707794 Chr.14: 68875002-68875002 on Build GRCh38
rs77499007 Chr.14: 68874941-68874941 on Build GRCh38
rs768868163 Chr.14: 68874843-68874843 on Build GRCh38
rs767331088 Chr.14: 68875143-68875144 on Build GRCh38
rs763814462 Chr.14: 68874950-68874950 on Build GRCh38
rs749558617 Chr.14: 68874842-68874842 on Build GRCh38
rs962888664 Chr.14: 68874880-68874880 on Build GRCh38
rs375245539 Chr.14: 68874856-68874856 on Build GRCh38
rs41285480 Chr.14: 68875189-68875189 on Build GRCh38
rs547204209 Chr.14: 68874993-68874993 on Build GRCh38
rs41285478 Chr.14: 68875024-68875024 on Build GRCh38
rs765310464 Chr.14: 68875056-68875056 on Build GRCh38
rs755072170 Chr.14: 68875012-68875012 on Build GRCh38
rs11557769 Chr.14: 68874936-68874936 on Build GRCh38
rs758102806 Chr.14: 68874900-68874900 on Build GRCh38
rs745859336 Chr.14: 68874923-68874923 on Build GRCh38
rs926484025 Chr.14: 68875112-68875112 on Build GRCh38
rs773768447 Chr.14: 68875033-68875033 on Build GRCh38
rs111795768 Chr.14: 68874862-68874862 on Build GRCh38
rs143640239 Chr.14: 68875099-68875099 on Build GRCh38
rs541262717 Chr.14: 68874892-68874892 on Build GRCh38
rs774807508 Chr.14: 68874851-68874851 on Build GRCh38
rs1055860090 Chr.14: 68874981-68874981 on Build GRCh38
rs758983643 Chr.14: 68874846-68874846 on Build GRCh38
rs750327189 Chr.14: 68874962-68874962 on Build GRCh38
rs761651604 Chr.14: 68874957-68874957 on Build GRCh38
rs376070530 Chr.14: 68875013-68875013 on Build GRCh38
rs764403516 Chr.14: 68875073-68875073 on Build GRCh38
rs780483163 Chr.14: 68875111-68875111 on Build GRCh38
rs553627673 Chr.14: 68875171-68875171 on Build GRCh38
rs764889893 Chr.14: 68874884-68874884 on Build GRCh38
rs752923320 Chr.14: 68875014-68875014 on Build GRCh38
rs138192933 Chr.14: 68875237-68875237 on Build GRCh38
rs762175698 Chr.14: 68874854-68874854 on Build GRCh38
rs955084884 Chr.14: 68874853-68874853 on Build GRCh38
rs776311435 Chr.14: 68875108-68875108 on Build GRCh38
rs781108326 Chr.14: 68874921-68874921 on Build GRCh38
rs200240249 Chr.14: 68875127-68875128 on Build GRCh38
rs753963391 Chr.14: 68874969-68874969 on Build GRCh38
rs752368324 Chr.14: 68874899-68874899 on Build GRCh38
rs753470271 Chr.14: 68875021-68875022 on Build GRCh38
rs751884044 Chr.14: 68875078-68875078 on Build GRCh38
rs771638947 Chr.14: 68875101-68875101 on Build GRCh38
rs867352977 Chr.14: 68874959-68874959 on Build GRCh38
rs758714570 Chr.14: 68875015-68875015 on Build GRCh38
rs750243442 Chr.14: 68874998-68874998 on Build GRCh38
rs766265937 Chr.14: 68875096-68875096 on Build GRCh38
rs768184934 Chr.14: 68875032-68875032 on Build GRCh38
rs757670319 Chr.14: 68875079-68875079 on Build GRCh38
rs766220501 Chr.14: 68875040-68875040 on Build GRCh38
rs747468223 Chr.14: 68874985-68874985 on Build GRCh38
rs192640536 Chr.14: 68874876-68874876 on Build GRCh38
rs746713224 Chr.14: 68875104-68875104 on Build GRCh38
rs886391072 Chr.14: 68875006-68875006 on Build GRCh38
rs768105419 Chr.14: 68874991-68874991 on Build GRCh38
rs773833507 Chr.14: 68874942-68874942 on Build GRCh38
rs757078973 Chr.14: 68874917-68874917 on Build GRCh38
rs369475970 Chr.14: 68875025-68875025 on Build GRCh38
rs763097732 Chr.14: 68875070-68875070 on Build GRCh38
rs551047030 Chr.14: 68875039-68875039 on Build GRCh38
rs765089073 Chr.14: 68875116-68875116 on Build GRCh38
rs776909803 Chr.14: 68875038-68875038 on Build GRCh38
rs746348933 Chr.14: 68875088-68875088 on Build GRCh38
rs767345293 Chr.14: 68874960-68874960 on Build GRCh38
rs959254034 Chr.14: 68875120-68875120 on Build GRCh38
rs200346938 Chr.14: 68874978-68874978 on Build GRCh38
rs758624466 Chr.14: 68875063-68875063 on Build GRCh38
rs549088260 Chr.14: 68875110-68875110 on Build GRCh38
rs778223053 Chr.14: 68875016-68875016 on Build GRCh38
rs763066535 Chr.14: 68874934-68874934 on Build GRCh38
rs200879748 Chr.14: 68875026-68875026 on Build GRCh38
rs772449856 Chr.14: 68874980-68874980 on Build GRCh38
rs937291448 Chr.14: 68875227-68875227 on Build GRCh38
rs753785461 Chr.14: 68875009-68875009 on Build GRCh38
rs950156992 Chr.14: 68875291-68875291 on Build GRCh38
rs752352598 Chr.14: 68875043-68875043 on Build GRCh38

View Full Product Details