Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 68882731-68883273 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTN1
Entrez Gene ID
87
Gene Name
actinin alpha 1
Gene Aliases
BDPLT15
UniGene
Hs.235750
Gene Chromosome Location
Chr.14: 68874123-68979366 on Build GRCh38
Transcripts
NM_001102.3 NM_001130004.1 NM_001130005.1 XM_011537265.2 XM_011537266.2 XM_011537267.2 XM_011537268.2 XM_017021720.1 XM_017021721.1 XM_017021722.1 XM_017021723.1 XM_017021725.1 XM_017021726.1 XM_017021727.1 XM_017021728.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6592960 c.1760C>T p.A587V Chr.14: 68882931-68882931 on Build GRCh38
5832811 c.1641_1643delGAC p.T549delT Chr.14: 68883048-68883050 on Build GRCh38
4781134 c.1750G>A p.V584I Chr.14: 68882941-68882941 on Build GRCh38
4814172 c.1780A>G p.T594A Chr.14: 68882911-68882911 on Build GRCh38
5362653 c.1763G>A p.G588D Chr.14: 68882928-68882928 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773705825 Chr.14: 68883157-68883157 on Build GRCh38
rs760876057 Chr.14: 68882942-68882942 on Build GRCh38
rs565458454 Chr.14: 68882760-68882760 on Build GRCh38
rs745457004 Chr.14: 68882903-68882903 on Build GRCh38
rs766497394 Chr.14: 68882944-68882944 on Build GRCh38
rs1019812463 Chr.14: 68882788-68882788 on Build GRCh38
rs749614229 Chr.14: 68882853-68882853 on Build GRCh38
rs748969644 Chr.14: 68883017-68883017 on Build GRCh38
rs907173601 Chr.14: 68882937-68882937 on Build GRCh38
rs779417401 Chr.14: 68883099-68883099 on Build GRCh38
rs373146144 Chr.14: 68882930-68882930 on Build GRCh38
rs201840732 Chr.14: 68883102-68883102 on Build GRCh38
rs570942124 Chr.14: 68883112-68883112 on Build GRCh38
rs761673617 Chr.14: 68883094-68883094 on Build GRCh38
rs376776021 Chr.14: 68883221-68883221 on Build GRCh38
rs994449555 Chr.14: 68882829-68882829 on Build GRCh38
rs147824601 Chr.14: 68883007-68883007 on Build GRCh38
rs916185984 Chr.14: 68882915-68882915 on Build GRCh38
rs888711269 Chr.14: 68883160-68883160 on Build GRCh38
rs768390005 Chr.14: 68883037-68883037 on Build GRCh38
rs769325974 Chr.14: 68882904-68882904 on Build GRCh38
rs56060157 Chr.14: 68882811-68882811 on Build GRCh38
rs763913720 Chr.14: 68882927-68882927 on Build GRCh38
rs775271640 Chr.14: 68882917-68882917 on Build GRCh38
rs780402829 Chr.14: 68882852-68882852 on Build GRCh38
rs766524550 Chr.14: 68883104-68883104 on Build GRCh38
rs374366318 Chr.14: 68882984-68882984 on Build GRCh38
rs1027273023 Chr.14: 68883158-68883158 on Build GRCh38
rs940116853 Chr.14: 68882954-68882954 on Build GRCh38
rs776016415 Chr.14: 68882900-68882900 on Build GRCh38
rs569015131 Chr.14: 68882783-68882783 on Build GRCh38
rs755451797 Chr.14: 68882859-68882859 on Build GRCh38
rs774147265 Chr.14: 68883047-68883047 on Build GRCh38
rs184265425 Chr.14: 68883105-68883105 on Build GRCh38
rs748706936 Chr.14: 68882886-68882886 on Build GRCh38
rs749504247 Chr.14: 68882779-68882779 on Build GRCh38
rs538427814 Chr.14: 68883139-68883139 on Build GRCh38
rs754166842 Chr.14: 68882949-68882949 on Build GRCh38
rs562792371 Chr.14: 68882872-68882872 on Build GRCh38
rs977533079 Chr.14: 68882800-68882800 on Build GRCh38
rs201198540 Chr.14: 68882985-68882985 on Build GRCh38
rs759522292 Chr.14: 68882844-68882844 on Build GRCh38
rs957125710 Chr.14: 68883166-68883166 on Build GRCh38
rs745353238 Chr.14: 68882998-68882998 on Build GRCh38
rs1045728006 Chr.14: 68882932-68882932 on Build GRCh38
rs758830672 Chr.14: 68882997-68882997 on Build GRCh38
rs189124025 Chr.14: 68883111-68883111 on Build GRCh38
rs897351053 Chr.14: 68883154-68883154 on Build GRCh38
rs370531513 Chr.14: 68883100-68883100 on Build GRCh38
rs767579148 Chr.14: 68882935-68882935 on Build GRCh38
rs867564562 Chr.14: 68882921-68882921 on Build GRCh38
rs1050307837 Chr.14: 68882771-68882771 on Build GRCh38
rs750611707 Chr.14: 68882825-68882825 on Build GRCh38
rs916279997 Chr.14: 68882968-68882968 on Build GRCh38
rs750475462 Chr.14: 68882941-68882941 on Build GRCh38
rs779454401 Chr.14: 68882966-68882966 on Build GRCh38
rs995103995 Chr.14: 68883024-68883024 on Build GRCh38
rs771899490 Chr.14: 68883097-68883097 on Build GRCh38
rs116455820 Chr.14: 68883008-68883008 on Build GRCh38
rs944148866 Chr.14: 68883118-68883118 on Build GRCh38
rs1041220300 Chr.14: 68883145-68883145 on Build GRCh38
rs767238747 Chr.14: 68882983-68882991 on Build GRCh38
rs762709684 Chr.14: 68882922-68882922 on Build GRCh38
rs370990057 Chr.14: 68882827-68882827 on Build GRCh38
rs769394983 Chr.14: 68882999-68882999 on Build GRCh38
rs772539105 Chr.14: 68882764-68882764 on Build GRCh38
rs555037578 Chr.14: 68882931-68882931 on Build GRCh38
rs550781697 Chr.14: 68882841-68882841 on Build GRCh38
rs377453106 Chr.14: 68882989-68882989 on Build GRCh38
rs370983078 Chr.14: 68882990-68882990 on Build GRCh38
rs977106503 Chr.14: 68883146-68883146 on Build GRCh38
rs142151023 Chr.14: 68882957-68882957 on Build GRCh38

View Full Product Details