Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 70522281-70522779 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAM20
Entrez Gene ID
8748
Gene Name
ADAM metallopeptidase domain 20
Gene Aliases
-
UniGene
Hs.177984
Gene Chromosome Location
Chr.14: 70520817-70564742 on Build GRCh38
Transcripts
NM_003814.4 XM_005268151.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4565572 c.2251_2252CC>TT p.P751F Chr.14: 70522656-70522657 on Build GRCh38
6554495 c.2287C>A p.L763I Chr.14: 70522621-70522621 on Build GRCh38
5573927 c.2268_2269insT p.L757fs*9 Chr.14: 70522639-70522640 on Build GRCh38
4979926 c.2215G>T p.G739* Chr.14: 70522693-70522693 on Build GRCh38
957402 c.2274T>G p.F758L Chr.14: 70522634-70522634 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs999998694 Chr.14: 70522734-70522734 on Build GRCh38
rs764829431 Chr.14: 70522530-70522530 on Build GRCh38
rs1032899073 Chr.14: 70522460-70522460 on Build GRCh38
rs141912564 Chr.14: 70522755-70522755 on Build GRCh38
rs551271125 Chr.14: 70522729-70522729 on Build GRCh38
rs768158998 Chr.14: 70522750-70522750 on Build GRCh38
rs556054275 Chr.14: 70522453-70522453 on Build GRCh38
rs561183621 Chr.14: 70522496-70522496 on Build GRCh38
rs753081896 Chr.14: 70522693-70522693 on Build GRCh38
rs747551951 Chr.14: 70522633-70522633 on Build GRCh38
rs909128038 Chr.14: 70522699-70522699 on Build GRCh38
rs148102371 Chr.14: 70522676-70522676 on Build GRCh38
rs750467732 Chr.14: 70522581-70522581 on Build GRCh38
rs780204552 Chr.14: 70522590-70522590 on Build GRCh38
rs879189813 Chr.14: 70522732-70522732 on Build GRCh38
rs533739178 Chr.14: 70522319-70522319 on Build GRCh38
rs769141762 Chr.14: 70522651-70522651 on Build GRCh38
rs140037614 Chr.14: 70522608-70522608 on Build GRCh38
rs902998308 Chr.14: 70522424-70522424 on Build GRCh38
rs755231283 Chr.14: 70522600-70522600 on Build GRCh38
rs757171044 Chr.14: 70522567-70522567 on Build GRCh38
rs778308611 Chr.14: 70522713-70522713 on Build GRCh38
rs556553542 Chr.14: 70522547-70522547 on Build GRCh38
rs747464417 Chr.14: 70522717-70522717 on Build GRCh38
rs766302796 Chr.14: 70522688-70522688 on Build GRCh38
rs756143531 Chr.14: 70522588-70522588 on Build GRCh38
rs577041572 Chr.14: 70522376-70522376 on Build GRCh38
rs982514467 Chr.14: 70522519-70522519 on Build GRCh38
rs751377878 Chr.14: 70522555-70522555 on Build GRCh38
rs372675639 Chr.14: 70522551-70522551 on Build GRCh38
rs779245540 Chr.14: 70522692-70522692 on Build GRCh38
rs916461775 Chr.14: 70522334-70522334 on Build GRCh38
rs768425870 Chr.14: 70522668-70522668 on Build GRCh38
rs767490191 Chr.14: 70522580-70522580 on Build GRCh38
rs748875168 Chr.14: 70522733-70522733 on Build GRCh38
rs150693723 Chr.14: 70522368-70522368 on Build GRCh38
rs747853621 Chr.14: 70522455-70522455 on Build GRCh38
rs903029734 Chr.14: 70522723-70522723 on Build GRCh38
rs758796268 Chr.14: 70522708-70522708 on Build GRCh38
rs766671204 Chr.14: 70522558-70522560 on Build GRCh38
rs889351428 Chr.14: 70522529-70522529 on Build GRCh38
rs985185032 Chr.14: 70522388-70522388 on Build GRCh38
rs755140580 Chr.14: 70522691-70522691 on Build GRCh38
rs1008929524 Chr.14: 70522392-70522392 on Build GRCh38
rs763741227 Chr.14: 70522549-70522549 on Build GRCh38
rs377263674 Chr.14: 70522562-70522562 on Build GRCh38
rs958622933 Chr.14: 70522461-70522461 on Build GRCh38
rs746003526 Chr.14: 70522624-70522624 on Build GRCh38
rs779428789 Chr.14: 70522731-70522731 on Build GRCh38
rs112517334 Chr.14: 70522528-70522528 on Build GRCh38
rs201701584 Chr.14: 70522696-70522696 on Build GRCh38
rs1023974045 Chr.14: 70522497-70522497 on Build GRCh38
rs1059167 Chr.14: 70522399-70522399 on Build GRCh38
rs750308805 Chr.14: 70522684-70522684 on Build GRCh38
rs749412698 Chr.14: 70522595-70522595 on Build GRCh38
rs751789159 Chr.14: 70522635-70522637 on Build GRCh38
rs111718938 Chr.14: 70522586-70522586 on Build GRCh38
rs1023497640 Chr.14: 70522655-70522655 on Build GRCh38
rs61753549 Chr.14: 70522609-70522609 on Build GRCh38
rs542613215 Chr.14: 70522481-70522481 on Build GRCh38
rs1050474364 Chr.14: 70522354-70522354 on Build GRCh38
rs775083370 Chr.14: 70522652-70522652 on Build GRCh38
rs554659896 Chr.14: 70522307-70522307 on Build GRCh38
rs971168537 Chr.14: 70522498-70522498 on Build GRCh38
rs776735159 Chr.14: 70522529-70522529 on Build GRCh38
rs761634645 Chr.14: 70522679-70522679 on Build GRCh38
rs778786768 Chr.14: 70522403-70522403 on Build GRCh38
rs112530813 Chr.14: 70522683-70522683 on Build GRCh38
rs761473055 Chr.14: 70522758-70522758 on Build GRCh38
rs879137776 Chr.14: 70522517-70522517 on Build GRCh38
rs566572398 Chr.14: 70522309-70522309 on Build GRCh38
rs944817763 Chr.14: 70522703-70522703 on Build GRCh38
rs940640020 Chr.14: 70522726-70522726 on Build GRCh38
rs932235627 Chr.14: 70522631-70522631 on Build GRCh38
rs1012546789 Chr.14: 70522467-70522467 on Build GRCh38
rs17108204 Chr.14: 70522458-70522458 on Build GRCh38
rs544133946 Chr.14: 70522427-70522427 on Build GRCh38
rs754014760 Chr.14: 70522689-70522689 on Build GRCh38
rs760618847 Chr.14: 70522685-70522685 on Build GRCh38
rs144548693 Chr.14: 70522660-70522660 on Build GRCh38
rs778464321 Chr.14: 70522626-70522626 on Build GRCh38

View Full Product Details