Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 70524302-70524811 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAM20
Entrez Gene ID
8748
Gene Name
ADAM metallopeptidase domain 20
Gene Aliases
-
UniGene
Hs.177984
Gene Chromosome Location
Chr.14: 70520817-70564742 on Build GRCh38
Transcripts
NM_003814.4 XM_005268151.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1515849 c.270A>G p.P90P Chr.14: 70524638-70524638 on Build GRCh38
5382581 c.373C>A p.H125N Chr.14: 70524535-70524535 on Build GRCh38
3987766 c.479A>G p.Y160C Chr.14: 70524429-70524429 on Build GRCh38
698805 c.335C>T p.S112F Chr.14: 70524573-70524573 on Build GRCh38
698806 c.523C>T p.L175F Chr.14: 70524385-70524385 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775320440 Chr.14: 70524610-70524610 on Build GRCh38
rs751702950 Chr.14: 70524542-70524542 on Build GRCh38
rs187181836 Chr.14: 70524480-70524480 on Build GRCh38
rs752625355 Chr.14: 70524546-70524546 on Build GRCh38
rs754136240 Chr.14: 70524769-70524769 on Build GRCh38
rs769518959 Chr.14: 70524598-70524598 on Build GRCh38
rs935089066 Chr.14: 70524539-70524539 on Build GRCh38
rs376577554 Chr.14: 70524506-70524506 on Build GRCh38
rs777388468 Chr.14: 70524567-70524567 on Build GRCh38
rs746279398 Chr.14: 70524433-70524433 on Build GRCh38
rs759326079 Chr.14: 70524491-70524491 on Build GRCh38
rs769430491 Chr.14: 70524367-70524373 on Build GRCh38
rs773699050 Chr.14: 70524370-70524370 on Build GRCh38
rs755610724 Chr.14: 70524562-70524562 on Build GRCh38
rs749126085 Chr.14: 70524699-70524699 on Build GRCh38
rs756351593 Chr.14: 70524654-70524654 on Build GRCh38
rs747262200 Chr.14: 70524347-70524347 on Build GRCh38
rs560795989 Chr.14: 70524422-70524422 on Build GRCh38
rs773794388 Chr.14: 70524658-70524685 on Build GRCh38
rs760949231 Chr.14: 70524753-70524753 on Build GRCh38
rs199645906 Chr.14: 70524678-70524678 on Build GRCh38
rs749821155 Chr.14: 70524455-70524455 on Build GRCh38
rs923645029 Chr.14: 70524523-70524523 on Build GRCh38
rs890777558 Chr.14: 70524488-70524488 on Build GRCh38
rs757386320 Chr.14: 70524543-70524543 on Build GRCh38
rs764056762 Chr.14: 70524631-70524631 on Build GRCh38
rs750605319 Chr.14: 70524653-70524653 on Build GRCh38
rs759361256 Chr.14: 70524407-70524407 on Build GRCh38
rs201128252 Chr.14: 70524633-70524633 on Build GRCh38
rs905871218 Chr.14: 70524568-70524568 on Build GRCh38
rs766849076 Chr.14: 70524655-70524655 on Build GRCh38
rs373576636 Chr.14: 70524742-70524742 on Build GRCh38
rs563764959 Chr.14: 70524582-70524582 on Build GRCh38
rs754295950 Chr.14: 70524663-70524663 on Build GRCh38
rs965014835 Chr.14: 70524712-70524712 on Build GRCh38
rs752562055 Chr.14: 70524505-70524505 on Build GRCh38
rs947639771 Chr.14: 70524558-70524558 on Build GRCh38
rs772919007 Chr.14: 70524469-70524469 on Build GRCh38
rs866351370 Chr.14: 70524752-70524752 on Build GRCh38
rs372885164 Chr.14: 70524548-70524548 on Build GRCh38
rs752652118 Chr.14: 70524415-70524415 on Build GRCh38
rs61739470 Chr.14: 70524705-70524705 on Build GRCh38
rs761362349 Chr.14: 70524374-70524374 on Build GRCh38
rs760087634 Chr.14: 70524402-70524402 on Build GRCh38
rs1026811592 Chr.14: 70524404-70524404 on Build GRCh38
rs771652900 Chr.14: 70524463-70524463 on Build GRCh38
rs774811010 Chr.14: 70524354-70524354 on Build GRCh38
rs767624174 Chr.14: 70524741-70524741 on Build GRCh38
rs746577934 Chr.14: 70524573-70524573 on Build GRCh38
rs531288041 Chr.14: 70524624-70524624 on Build GRCh38
rs745519054 Chr.14: 70524682-70524682 on Build GRCh38
rs985353996 Chr.14: 70524697-70524697 on Build GRCh38
rs952658132 Chr.14: 70524416-70524416 on Build GRCh38
rs780671362 Chr.14: 70524452-70524452 on Build GRCh38
rs758597254 Chr.14: 70524337-70524337 on Build GRCh38
rs759941193 Chr.14: 70524773-70524773 on Build GRCh38
rs1024394504 Chr.14: 70524651-70524651 on Build GRCh38
rs374862551 Chr.14: 70524423-70524423 on Build GRCh38
rs770395441 Chr.14: 70524368-70524368 on Build GRCh38
rs758262878 Chr.14: 70524614-70524614 on Build GRCh38
rs952524221 Chr.14: 70524689-70524689 on Build GRCh38
rs761900874 Chr.14: 70524638-70524638 on Build GRCh38
rs765700772 Chr.14: 70524785-70524785 on Build GRCh38
rs770370530 Chr.14: 70524579-70524579 on Build GRCh38
rs117452523 Chr.14: 70524412-70524412 on Build GRCh38
rs140811379 Chr.14: 70524418-70524418 on Build GRCh38
rs769500504 Chr.14: 70524685-70524685 on Build GRCh38
rs758432789 Chr.14: 70524417-70524417 on Build GRCh38
rs1059166 Chr.14: 70524701-70524701 on Build GRCh38
rs878918875 Chr.14: 70524482-70524482 on Build GRCh38
rs768457876 Chr.14: 70524704-70524704 on Build GRCh38
rs777568501 Chr.14: 70524652-70524652 on Build GRCh38
rs575058707 Chr.14: 70524388-70524388 on Build GRCh38
rs770174303 Chr.14: 70524445-70524445 on Build GRCh38
rs745510452 Chr.14: 70524592-70524592 on Build GRCh38
rs138842334 Chr.14: 70524561-70524561 on Build GRCh38
rs777922288 Chr.14: 70524340-70524340 on Build GRCh38
rs779513861 Chr.14: 70524670-70524670 on Build GRCh38
rs867886971 Chr.14: 70524395-70524395 on Build GRCh38
rs746343890 Chr.14: 70524365-70524365 on Build GRCh38
rs867913616 Chr.14: 70524439-70524439 on Build GRCh38
rs766758251 Chr.14: 70524764-70524764 on Build GRCh38
rs764143143 Chr.14: 70524541-70524541 on Build GRCh38
rs370689994 Chr.14: 70524550-70524550 on Build GRCh38
rs760242363 Chr.14: 70524474-70524474 on Build GRCh38
rs374396103 Chr.14: 70524556-70524556 on Build GRCh38
rs764972924 Chr.14: 70524496-70524496 on Build GRCh38
rs144800201 Chr.14: 70524476-70524476 on Build GRCh38
rs891871785 Chr.14: 70524589-70524589 on Build GRCh38
rs750646329 Chr.14: 70524545-70524545 on Build GRCh38
rs771743089 Chr.14: 70524384-70524384 on Build GRCh38
rs756719590 Chr.14: 70524676-70524676 on Build GRCh38
rs369248948 Chr.14: 70524431-70524431 on Build GRCh38
rs142051635 Chr.14: 70524672-70524672 on Build GRCh38
rs755520106 Chr.14: 70524668-70524668 on Build GRCh38
rs138673978 Chr.14: 70524685-70524686 on Build GRCh38
rs1014329801 Chr.14: 70524617-70524617 on Build GRCh38
rs761674464 Chr.14: 70524719-70524719 on Build GRCh38
rs772935601 Chr.14: 70524386-70524386 on Build GRCh38
rs767502311 Chr.14: 70524640-70524640 on Build GRCh38
rs202217158 Chr.14: 70524698-70524698 on Build GRCh38
rs774282355 Chr.14: 70524709-70524709 on Build GRCh38
rs918960136 Chr.14: 70524734-70524734 on Build GRCh38
rs556710137 Chr.14: 70524355-70524355 on Build GRCh38
rs767826634 Chr.14: 70524544-70524544 on Build GRCh38
rs774388289 Chr.14: 70524636-70524636 on Build GRCh38
rs976861065 Chr.14: 70524727-70524727 on Build GRCh38

View Full Product Details