Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 75003414-75003909 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF2B2
Entrez Gene ID
8892
Gene Name
eukaryotic translation initiation factor 2B subunit beta
Gene Aliases
EIF-2Bbeta, EIF2B
UniGene
Hs.409137
Gene Chromosome Location
Chr.14: 75002909-75009591 on Build GRCh38
Transcripts
NM_014239.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1477773 c.331A>T p.K111* Chr.14: 75003597-75003597 on Build GRCh38
3793864 c.293G>A p.G98E Chr.14: 75003559-75003559 on Build GRCh38
433375 c.301G>A p.D101N Chr.14: 75003567-75003567 on Build GRCh38
5824579 c.367A>T p.S123C Chr.14: 75003633-75003633 on Build GRCh38
7313744 c.420G>T p.L140L Chr.14: 75003686-75003686 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777252700 Chr.14: 75003500-75003500 on Build GRCh38
rs762604029 Chr.14: 75003543-75003543 on Build GRCh38
rs372174903 Chr.14: 75003650-75003650 on Build GRCh38
rs368133564 Chr.14: 75003564-75003564 on Build GRCh38
rs759621968 Chr.14: 75003599-75003599 on Build GRCh38
rs746678040 Chr.14: 75003512-75003512 on Build GRCh38
rs1039929308 Chr.14: 75003643-75003643 on Build GRCh38
rs775335483 Chr.14: 75003537-75003537 on Build GRCh38
rs374195020 Chr.14: 75003620-75003620 on Build GRCh38
rs766487534 Chr.14: 75003714-75003714 on Build GRCh38
rs760706892 Chr.14: 75003706-75003706 on Build GRCh38
rs777077732 Chr.14: 75003666-75003668 on Build GRCh38
rs549982732 Chr.14: 75003855-75003855 on Build GRCh38
rs776342453 Chr.14: 75003522-75003522 on Build GRCh38
rs893220651 Chr.14: 75003817-75003817 on Build GRCh38
rs183116394 Chr.14: 75003437-75003437 on Build GRCh38
rs769894290 Chr.14: 75003532-75003532 on Build GRCh38
rs373906805 Chr.14: 75003550-75003550 on Build GRCh38
rs12432898 Chr.14: 75003834-75003834 on Build GRCh38
rs1020267453 Chr.14: 75003468-75003468 on Build GRCh38
rs768864027 Chr.14: 75003521-75003522 on Build GRCh38
rs1041742318 Chr.14: 75003856-75003856 on Build GRCh38
rs779349624 Chr.14: 75003611-75003611 on Build GRCh38
rs175041 Chr.14: 75003743-75003743 on Build GRCh38
rs768524274 Chr.14: 75003544-75003544 on Build GRCh38
rs751159755 Chr.14: 75003612-75003612 on Build GRCh38
rs997505668 Chr.14: 75003873-75003873 on Build GRCh38
rs200642949 Chr.14: 75003648-75003648 on Build GRCh38
rs201912801 Chr.14: 75003671-75003671 on Build GRCh38
rs748297529 Chr.14: 75003675-75003676 on Build GRCh38
rs755232669 Chr.14: 75003608-75003608 on Build GRCh38
rs754240366 Chr.14: 75003715-75003715 on Build GRCh38
rs373932168 Chr.14: 75003528-75003528 on Build GRCh38
rs745423619 Chr.14: 75003633-75003633 on Build GRCh38
rs760147450 Chr.14: 75003724-75003724 on Build GRCh38
rs770508411 Chr.14: 75003518-75003518 on Build GRCh38
rs367701960 Chr.14: 75003727-75003727 on Build GRCh38
rs772320060 Chr.14: 75003674-75003674 on Build GRCh38
rs967630584 Chr.14: 75003491-75003491 on Build GRCh38
rs370483997 Chr.14: 75003591-75003591 on Build GRCh38
rs755393155 Chr.14: 75003440-75003440 on Build GRCh38
rs1045660845 Chr.14: 75003708-75003708 on Build GRCh38
rs150617429 Chr.14: 75003646-75003646 on Build GRCh38
rs918318919 Chr.14: 75003464-75003464 on Build GRCh38
rs17849324 Chr.14: 75003565-75003565 on Build GRCh38
rs1030584724 Chr.14: 75003506-75003506 on Build GRCh38
rs997727777 Chr.14: 75003501-75003501 on Build GRCh38
rs1024990084 Chr.14: 75003794-75003794 on Build GRCh38
rs762017222 Chr.14: 75003673-75003673 on Build GRCh38
rs866004755 Chr.14: 75003859-75003859 on Build GRCh38
rs201622896 Chr.14: 75003667-75003667 on Build GRCh38
rs376824565 Chr.14: 75003707-75003707 on Build GRCh38
rs769455036 Chr.14: 75003638-75003638 on Build GRCh38
rs368343040 Chr.14: 75003621-75003621 on Build GRCh38
rs780945717 Chr.14: 75003745-75003745 on Build GRCh38
rs929643497 Chr.14: 75003669-75003669 on Build GRCh38
rs766843755 Chr.14: 75003577-75003577 on Build GRCh38
rs773252033 Chr.14: 75003689-75003689 on Build GRCh38
rs750568077 Chr.14: 75003557-75003557 on Build GRCh38
rs762068601 Chr.14: 75003547-75003547 on Build GRCh38
rs562944339 Chr.14: 75003860-75003860 on Build GRCh38
rs753157137 Chr.14: 75003739-75003739 on Build GRCh38
rs376049144 Chr.14: 75003549-75003549 on Build GRCh38

View Full Product Details