Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 76791215-76791716 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANGEL1
Entrez Gene ID
23357
Gene Name
angel homolog 1
Gene Aliases
Ccr4e, KIAA0759
UniGene
Hs.497448
Gene Chromosome Location
Chr.14: 76786473-76812947 on Build GRCh38
Transcripts
NM_015305.3 XM_017021115.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
958071 c.1622G>A p.R541Q Chr.14: 76791363-76791363 on Build GRCh38
6001427 c.1620G>A p.E540E Chr.14: 76791365-76791365 on Build GRCh38
374171 c.1631G>A p.G544D Chr.14: 76791354-76791354 on Build GRCh38
6141219 c.1651G>C p.E551Q Chr.14: 76791334-76791334 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs748876164 Chr.14: 76791249-76791249 on Build GRCh38
rs867597311 Chr.14: 76791460-76791460 on Build GRCh38
rs373832136 Chr.14: 76791312-76791312 on Build GRCh38
rs978770403 Chr.14: 76791621-76791621 on Build GRCh38
rs934406320 Chr.14: 76791271-76791271 on Build GRCh38
rs372803737 Chr.14: 76791422-76791422 on Build GRCh38
rs367994519 Chr.14: 76791320-76791320 on Build GRCh38
rs201471357 Chr.14: 76791293-76791293 on Build GRCh38
rs771569886 Chr.14: 76791380-76791380 on Build GRCh38
rs540272464 Chr.14: 76791658-76791658 on Build GRCh38
rs768223567 Chr.14: 76791251-76791251 on Build GRCh38
rs368211974 Chr.14: 76791378-76791378 on Build GRCh38
rs185784196 Chr.14: 76791622-76791622 on Build GRCh38
rs372887001 Chr.14: 76791406-76791406 on Build GRCh38
rs758529439 Chr.14: 76791323-76791323 on Build GRCh38
rs544353853 Chr.14: 76791621-76791621 on Build GRCh38
rs531908104 Chr.14: 76791285-76791285 on Build GRCh38
rs987990392 Chr.14: 76791262-76791262 on Build GRCh38
rs776251975 Chr.14: 76791396-76791396 on Build GRCh38
rs761767441 Chr.14: 76791266-76791266 on Build GRCh38
rs376487409 Chr.14: 76791531-76791531 on Build GRCh38
rs372266603 Chr.14: 76791326-76791326 on Build GRCh38
rs766077836 Chr.14: 76791289-76791289 on Build GRCh38
rs200052331 Chr.14: 76791401-76791401 on Build GRCh38
rs898036893 Chr.14: 76791569-76791569 on Build GRCh38
rs111350779 Chr.14: 76791359-76791359 on Build GRCh38
rs768201400 Chr.14: 76791403-76791406 on Build GRCh38
rs750365968 Chr.14: 76791514-76791514 on Build GRCh38
rs747699999 Chr.14: 76791363-76791363 on Build GRCh38
rs760475285 Chr.14: 76791393-76791393 on Build GRCh38
rs773779793 Chr.14: 76791253-76791253 on Build GRCh38
rs752776943 Chr.14: 76791321-76791321 on Build GRCh38
rs767494689 Chr.14: 76791275-76791275 on Build GRCh38
rs757697851 Chr.14: 76791347-76791347 on Build GRCh38
rs190244120 Chr.14: 76791647-76791647 on Build GRCh38
rs776075797 Chr.14: 76791413-76791414 on Build GRCh38
rs904943657 Chr.14: 76791631-76791631 on Build GRCh38
rs558788168 Chr.14: 76791597-76791597 on Build GRCh38
rs760533089 Chr.14: 76791287-76791287 on Build GRCh38
rs965630010 Chr.14: 76791620-76791621 on Build GRCh38
rs566461818 Chr.14: 76791441-76791441 on Build GRCh38
rs754400846 Chr.14: 76791356-76791356 on Build GRCh38
rs570309968 Chr.14: 76791465-76791465 on Build GRCh38
rs747513089 Chr.14: 76791335-76791335 on Build GRCh38
rs374258511 Chr.14: 76791357-76791357 on Build GRCh38
rs575364239 Chr.14: 76791617-76791617 on Build GRCh38
rs989849557 Chr.14: 76791636-76791636 on Build GRCh38
rs373023265 Chr.14: 76791621-76791622 on Build GRCh38
rs190537085 Chr.14: 76791248-76791248 on Build GRCh38
rs138253007 Chr.14: 76791545-76791545 on Build GRCh38
rs183050258 Chr.14: 76791394-76791394 on Build GRCh38
rs897129592 Chr.14: 76791454-76791454 on Build GRCh38
rs760371840 Chr.14: 76791287-76791289 on Build GRCh38

View Full Product Details