Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 77672009-77672549 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALKBH1
Entrez Gene ID
8846
Gene Name
alkB homolog 1, histone H2A dioxygenase
Gene Aliases
ABH, ABH1, ALKBH, alkB, hABH
UniGene
Hs.94542
Gene Chromosome Location
Chr.14: 77672404-77708038 on Build GRCh38
Transcripts
NM_006020.2

SNP Detail

SNP ID Chromosome Location
rs1030691140 Chr.14: 77672116-77672116 on Build GRCh38
rs549401629 Chr.14: 77672440-77672440 on Build GRCh38
rs555566592 Chr.14: 77672212-77672212 on Build GRCh38
rs555713371 Chr.14: 77672124-77672124 on Build GRCh38
rs184069289 Chr.14: 77672525-77672525 on Build GRCh38
rs111507552 Chr.14: 77672324-77672324 on Build GRCh38
rs908169880 Chr.14: 77672195-77672195 on Build GRCh38
rs923444505 Chr.14: 77672132-77672132 on Build GRCh38
rs756777330 Chr.14: 77672494-77672494 on Build GRCh38
rs759467326 Chr.14: 77672405-77672406 on Build GRCh38
rs778373999 Chr.14: 77672370-77672370 on Build GRCh38
rs369832370 Chr.14: 77672354-77672354 on Build GRCh38
rs966136138 Chr.14: 77672451-77672451 on Build GRCh38
rs986658516 Chr.14: 77672322-77672322 on Build GRCh38
rs1052941939 Chr.14: 77672515-77672515 on Build GRCh38
rs539710088 Chr.14: 77672431-77672431 on Build GRCh38
rs973683489 Chr.14: 77672430-77672430 on Build GRCh38
rs970002241 Chr.14: 77672187-77672187 on Build GRCh38
rs558041577 Chr.14: 77672390-77672390 on Build GRCh38
rs186066510 Chr.14: 77672245-77672245 on Build GRCh38
rs146593907 Chr.14: 77672286-77672286 on Build GRCh38
rs113809140 Chr.14: 77672150-77672150 on Build GRCh38
rs555957054 Chr.14: 77672457-77672457 on Build GRCh38
rs942476253 Chr.14: 77672326-77672326 on Build GRCh38
rs904950718 Chr.14: 77672388-77672388 on Build GRCh38
rs901066790 Chr.14: 77672350-77672350 on Build GRCh38
rs976553531 Chr.14: 77672460-77672460 on Build GRCh38
rs190105785 Chr.14: 77672164-77672164 on Build GRCh38
rs999153350 Chr.14: 77672107-77672107 on Build GRCh38
rs1002749814 Chr.14: 77672250-77672250 on Build GRCh38
rs1023840938 Chr.14: 77672387-77672387 on Build GRCh38
rs1011543542 Chr.14: 77672285-77672285 on Build GRCh38
rs539052232 Chr.14: 77672115-77672115 on Build GRCh38
rs1031584283 Chr.14: 77672256-77672256 on Build GRCh38
rs1038005615 Chr.14: 77672222-77672222 on Build GRCh38
rs868866464 Chr.14: 77672418-77672418 on Build GRCh38
rs1010653915 Chr.14: 77672367-77672370 on Build GRCh38
rs765439600 Chr.14: 77672086-77672086 on Build GRCh38
rs547096249 Chr.14: 77672085-77672085 on Build GRCh38
rs182934230 Chr.14: 77672240-77672240 on Build GRCh38
rs572142383 Chr.14: 77672220-77672220 on Build GRCh38
rs116272054 Chr.14: 77672365-77672365 on Build GRCh38
rs752701330 Chr.14: 77672310-77672310 on Build GRCh38
rs972311096 Chr.14: 77672419-77672419 on Build GRCh38
rs762109121 Chr.14: 77672049-77672049 on Build GRCh38
rs190970876 Chr.14: 77672283-77672283 on Build GRCh38
rs914624694 Chr.14: 77672074-77672074 on Build GRCh38
rs906986369 Chr.14: 77672500-77672500 on Build GRCh38
rs1052089405 Chr.14: 77672364-77672370 on Build GRCh38
rs564232747 Chr.14: 77672235-77672235 on Build GRCh38
rs964792628 Chr.14: 77672084-77672084 on Build GRCh38
rs1023788711 Chr.14: 77672178-77672178 on Build GRCh38
rs541355750 Chr.14: 77672224-77672224 on Build GRCh38
rs141349539 Chr.14: 77672333-77672333 on Build GRCh38
rs935249650 Chr.14: 77672361-77672362 on Build GRCh38
rs537643595 Chr.14: 77672426-77672426 on Build GRCh38
rs546239376 Chr.14: 77672242-77672242 on Build GRCh38
rs938478123 Chr.14: 77672501-77672501 on Build GRCh38
rs935218604 Chr.14: 77672521-77672521 on Build GRCh38
rs948489700 Chr.14: 77672230-77672230 on Build GRCh38

View Full Product Details