Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 87941256-87941786 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALC
Entrez Gene ID
2581
Gene Name
galactosylceramidase
Gene Aliases
-
UniGene
Hs.513439
Gene Chromosome Location
Chr.14: 87933014-87993665 on Build GRCh38
Transcripts
NM_000153.3 NM_001201401.1 NM_001201402.1 XM_011536618.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4578043 c.1717A>G p.T573A Chr.14: 87941512-87941512 on Build GRCh38
5614171 c.1819G>T p.V607F Chr.14: 87941410-87941410 on Build GRCh38
5614172 c.1741G>T p.V581F Chr.14: 87941410-87941410 on Build GRCh38
4815377 c.1677T>G p.G559G Chr.14: 87941474-87941474 on Build GRCh38
4869341 c.1691G>T p.R564I Chr.14: 87941460-87941460 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs912696793 Chr.14: 87941556-87941556 on Build GRCh38
rs540937002 Chr.14: 87941371-87941372 on Build GRCh38
rs754732860 Chr.14: 87941400-87941400 on Build GRCh38
rs780514845 Chr.14: 87941572-87941573 on Build GRCh38
rs371792183 Chr.14: 87941527-87941527 on Build GRCh38
rs387906953 Chr.14: 87941433-87941433 on Build GRCh38
rs553493589 Chr.14: 87941315-87941315 on Build GRCh38
rs189385102 Chr.14: 87941428-87941428 on Build GRCh38
rs755827044 Chr.14: 87941561-87941561 on Build GRCh38
rs762787332 Chr.14: 87941381-87941382 on Build GRCh38
rs773382913 Chr.14: 87941597-87941597 on Build GRCh38
rs764417158 Chr.14: 87941644-87941644 on Build GRCh38
rs181724240 Chr.14: 87941759-87941759 on Build GRCh38
rs903307258 Chr.14: 87941366-87941366 on Build GRCh38
rs767690197 Chr.14: 87941386-87941386 on Build GRCh38
rs910194407 Chr.14: 87941298-87941298 on Build GRCh38
rs754668472 Chr.14: 87941490-87941493 on Build GRCh38
rs148028728 Chr.14: 87941634-87941634 on Build GRCh38
rs1012511324 Chr.14: 87941737-87941737 on Build GRCh38
rs894465095 Chr.14: 87941457-87941457 on Build GRCh38
rs756709858 Chr.14: 87941462-87941462 on Build GRCh38
rs410116 Chr.14: 87941375-87941375 on Build GRCh38
rs771864129 Chr.14: 87941329-87941329 on Build GRCh38
rs774555128 Chr.14: 87941502-87941502 on Build GRCh38
rs372465848 Chr.14: 87941426-87941426 on Build GRCh38
rs398607 Chr.14: 87941544-87941544 on Build GRCh38
rs74073881 Chr.14: 87941713-87941713 on Build GRCh38
rs374999191 Chr.14: 87941589-87941589 on Build GRCh38
rs775654226 Chr.14: 87941373-87941373 on Build GRCh38
rs200019401 Chr.14: 87941524-87941524 on Build GRCh38
rs912788562 Chr.14: 87941323-87941323 on Build GRCh38
rs771256299 Chr.14: 87941353-87941353 on Build GRCh38
rs12432149 Chr.14: 87941573-87941573 on Build GRCh38
rs746189262 Chr.14: 87941471-87941471 on Build GRCh38
rs448805 Chr.14: 87941390-87941390 on Build GRCh38
rs756735161 Chr.14: 87941542-87941542 on Build GRCh38
rs575607707 Chr.14: 87941358-87941358 on Build GRCh38
rs186349286 Chr.14: 87941291-87941291 on Build GRCh38
rs1047706684 Chr.14: 87941611-87941611 on Build GRCh38
rs766646367 Chr.14: 87941603-87941603 on Build GRCh38
rs962346415 Chr.14: 87941513-87941513 on Build GRCh38
rs201560122 Chr.14: 87941498-87941498 on Build GRCh38
rs778850146 Chr.14: 87941413-87941413 on Build GRCh38
rs767710271 Chr.14: 87941519-87941519 on Build GRCh38
rs570766960 Chr.14: 87941566-87941566 on Build GRCh38
rs751283440 Chr.14: 87941466-87941468 on Build GRCh38
rs751725084 Chr.14: 87941364-87941364 on Build GRCh38
rs773814987 Chr.14: 87941719-87941719 on Build GRCh38
rs766007316 Chr.14: 87941414-87941415 on Build GRCh38
rs747869263 Chr.14: 87941280-87941280 on Build GRCh38
rs1057461259 Chr.14: 87941697-87941697 on Build GRCh38
rs749724769 Chr.14: 87941572-87941572 on Build GRCh38
rs774769711 Chr.14: 87941370-87941371 on Build GRCh38
rs410117 Chr.14: 87941378-87941378 on Build GRCh38
rs368818550 Chr.14: 87941469-87941469 on Build GRCh38
rs367926377 Chr.14: 87941372-87941372 on Build GRCh38
rs535956106 Chr.14: 87941337-87941337 on Build GRCh38
rs752664357 Chr.14: 87941422-87941422 on Build GRCh38
rs929215561 Chr.14: 87941410-87941410 on Build GRCh38
rs1002946173 Chr.14: 87941438-87941438 on Build GRCh38
rs421466 Chr.14: 87941531-87941531 on Build GRCh38
rs115018138 Chr.14: 87941441-87941441 on Build GRCh38
rs879825641 Chr.14: 87941661-87941661 on Build GRCh38
rs751008420 Chr.14: 87941533-87941533 on Build GRCh38
rs772064918 Chr.14: 87941595-87941595 on Build GRCh38
rs34752717 Chr.14: 87941372-87941373 on Build GRCh38
rs760787183 Chr.14: 87941599-87941599 on Build GRCh38
rs752537626 Chr.14: 87941529-87941529 on Build GRCh38
rs947379038 Chr.14: 87941625-87941625 on Build GRCh38
rs914540444 Chr.14: 87941341-87941341 on Build GRCh38
rs370579937 Chr.14: 87941356-87941356 on Build GRCh38
rs375602648 Chr.14: 87941357-87941357 on Build GRCh38
rs775002574 Chr.14: 87941579-87941579 on Build GRCh38
rs927764561 Chr.14: 87941621-87941621 on Build GRCh38
rs567016031 Chr.14: 87941316-87941316 on Build GRCh38
rs745534989 Chr.14: 87941551-87941551 on Build GRCh38
rs540808138 Chr.14: 87941520-87941520 on Build GRCh38
rs381069 Chr.14: 87941720-87941720 on Build GRCh38
rs777018799 Chr.14: 87941669-87941669 on Build GRCh38
rs201016488 Chr.14: 87941477-87941477 on Build GRCh38
rs987367229 Chr.14: 87941545-87941545 on Build GRCh38
rs200219480 Chr.14: 87941512-87941512 on Build GRCh38
rs386779713 Chr.14: 87941531-87941544 on Build GRCh38
rs776098499 Chr.14: 87941475-87941475 on Build GRCh38

View Full Product Details