Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 87950581-87951095 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALC
Entrez Gene ID
2581
Gene Name
galactosylceramidase
Gene Aliases
-
UniGene
Hs.513439
Gene Chromosome Location
Chr.14: 87933014-87993665 on Build GRCh38
Transcripts
NM_000153.3 NM_001201401.1 NM_001201402.1 XM_011536618.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4769102 c.1108C>T p.R370W Chr.14: 87950724-87950724 on Build GRCh38
3498440 c.1233T>C p.F411F Chr.14: 87950677-87950677 on Build GRCh38
252513 c.1211T>C p.V404A Chr.14: 87950699-87950699 on Build GRCh38
6141580 c.1165C>T p.H389Y Chr.14: 87950745-87950745 on Build GRCh38
4769101 c.1186C>T p.R396W Chr.14: 87950724-87950724 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs443019 Chr.14: 87950900-87950900 on Build GRCh38
rs569569879 Chr.14: 87950726-87950726 on Build GRCh38
rs769598460 Chr.14: 87950633-87950636 on Build GRCh38
rs200420321 Chr.14: 87950763-87950763 on Build GRCh38
rs774455523 Chr.14: 87950752-87950752 on Build GRCh38
rs148098570 Chr.14: 87950612-87950612 on Build GRCh38
rs370110535 Chr.14: 87950637-87950637 on Build GRCh38
rs962305731 Chr.14: 87950775-87950775 on Build GRCh38
rs990016510 Chr.14: 87950836-87950836 on Build GRCh38
rs771706821 Chr.14: 87950691-87950691 on Build GRCh38
rs763125933 Chr.14: 87950727-87950727 on Build GRCh38
rs753714214 Chr.14: 87950774-87950774 on Build GRCh38
rs534783618 Chr.14: 87950854-87950854 on Build GRCh38
rs762736061 Chr.14: 87950752-87950753 on Build GRCh38
rs113021000 Chr.14: 87950781-87950781 on Build GRCh38
rs431748 Chr.14: 87950817-87950817 on Build GRCh38
rs760476173 Chr.14: 87950768-87950768 on Build GRCh38
rs770360911 Chr.14: 87951001-87951001 on Build GRCh38
rs1036456402 Chr.14: 87950993-87950993 on Build GRCh38
rs997692673 Chr.14: 87951056-87951056 on Build GRCh38
rs200707535 Chr.14: 87950650-87950650 on Build GRCh38
rs763257931 Chr.14: 87950632-87950632 on Build GRCh38
rs181045225 Chr.14: 87951035-87951035 on Build GRCh38
rs928261786 Chr.14: 87950924-87950924 on Build GRCh38
rs889539646 Chr.14: 87950926-87950926 on Build GRCh38
rs957430002 Chr.14: 87950820-87950820 on Build GRCh38
rs11300320 Chr.14: 87950752-87950752 on Build GRCh38
rs375928141 Chr.14: 87950618-87950618 on Build GRCh38
rs921054145 Chr.14: 87950762-87950762 on Build GRCh38
rs375732556 Chr.14: 87950787-87950787 on Build GRCh38
rs773196564 Chr.14: 87950651-87950654 on Build GRCh38
rs754947654 Chr.14: 87950663-87950663 on Build GRCh38
rs755870379 Chr.14: 87950654-87950654 on Build GRCh38
rs981375868 Chr.14: 87950919-87950919 on Build GRCh38
rs766055483 Chr.14: 87950770-87950770 on Build GRCh38
rs376181222 Chr.14: 87950719-87950719 on Build GRCh38
rs937051809 Chr.14: 87950928-87950928 on Build GRCh38
rs368284541 Chr.14: 87950745-87950745 on Build GRCh38
rs764350127 Chr.14: 87950735-87950735 on Build GRCh38
rs745975978 Chr.14: 87950950-87950950 on Build GRCh38
rs765380291 Chr.14: 87950622-87950622 on Build GRCh38
rs188738895 Chr.14: 87950970-87950970 on Build GRCh38
rs746358292 Chr.14: 87950720-87950720 on Build GRCh38
rs746305477 Chr.14: 87950799-87950799 on Build GRCh38
rs752145215 Chr.14: 87950786-87950786 on Build GRCh38
rs759622420 Chr.14: 87950619-87950619 on Build GRCh38
rs200605515 Chr.14: 87950659-87950659 on Build GRCh38
rs369997150 Chr.14: 87950641-87950641 on Build GRCh38
rs752875388 Chr.14: 87950623-87950623 on Build GRCh38
rs764213916 Chr.14: 87951066-87951066 on Build GRCh38
rs887930208 Chr.14: 87950723-87950723 on Build GRCh38
rs771232832 Chr.14: 87950711-87950711 on Build GRCh38
rs777305549 Chr.14: 87950702-87950702 on Build GRCh38
rs781400623 Chr.14: 87950798-87950798 on Build GRCh38
rs777444526 Chr.14: 87950788-87950788 on Build GRCh38
rs779990751 Chr.14: 87950658-87950658 on Build GRCh38
rs942187653 Chr.14: 87950960-87950960 on Build GRCh38
rs913077002 Chr.14: 87950896-87950896 on Build GRCh38
rs428542 Chr.14: 87950945-87950945 on Build GRCh38
rs770485731 Chr.14: 87950724-87950724 on Build GRCh38
rs765225359 Chr.14: 87950784-87950784 on Build GRCh38
rs112531377 Chr.14: 87950674-87950674 on Build GRCh38
rs753747900 Chr.14: 87950661-87950661 on Build GRCh38
rs747714175 Chr.14: 87950689-87950689 on Build GRCh38
rs371724294 Chr.14: 87950703-87950703 on Build GRCh38
rs751373579 Chr.14: 87950751-87950752 on Build GRCh38
rs558293541 Chr.14: 87950793-87950793 on Build GRCh38

View Full Product Details