Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 91608143-91608641 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CATSPERB
Entrez Gene ID
79820
Gene Name
cation channel sperm associated auxiliary subunit beta
Gene Aliases
C14orf161, CatSper(beta)
UniGene
Hs.131755
Gene Chromosome Location
Chr.14: 91580774-91732086 on Build GRCh38
Transcripts
NM_024764.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4890703 c.2654T>A p.F885Y Chr.14: 91608349-91608349 on Build GRCh38
3361267 c.2690A>C p.N897T Chr.14: 91608313-91608313 on Build GRCh38
958764 c.2652T>G p.N884K Chr.14: 91608351-91608351 on Build GRCh38
6076570 c.2606A>G p.Y869C Chr.14: 91608397-91608397 on Build GRCh38
6463733 c.2622T>A p.N874K Chr.14: 91608381-91608381 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs182981119 Chr.14: 91608285-91608285 on Build GRCh38
rs548349773 Chr.14: 91608547-91608547 on Build GRCh38
rs567846199 Chr.14: 91608551-91608551 on Build GRCh38
rs757283239 Chr.14: 91608302-91608302 on Build GRCh38
rs746093232 Chr.14: 91608283-91608284 on Build GRCh38
rs753128464 Chr.14: 91608429-91608429 on Build GRCh38
rs776392881 Chr.14: 91608415-91608415 on Build GRCh38
rs759231595 Chr.14: 91608416-91608416 on Build GRCh38
rs764935077 Chr.14: 91608424-91608424 on Build GRCh38
rs1037644166 Chr.14: 91608505-91608505 on Build GRCh38
rs922948883 Chr.14: 91608589-91608589 on Build GRCh38
rs144932723 Chr.14: 91608320-91608320 on Build GRCh38
rs764765276 Chr.14: 91608431-91608431 on Build GRCh38
rs202047912 Chr.14: 91608412-91608412 on Build GRCh38
rs964824561 Chr.14: 91608559-91608559 on Build GRCh38
rs773124980 Chr.14: 91608554-91608554 on Build GRCh38
rs763343885 Chr.14: 91608430-91608430 on Build GRCh38
rs199590674 Chr.14: 91608297-91608297 on Build GRCh38
rs370762893 Chr.14: 91608310-91608310 on Build GRCh38
rs374038099 Chr.14: 91608248-91608248 on Build GRCh38
rs772318180 Chr.14: 91608309-91608309 on Build GRCh38
rs556726728 Chr.14: 91608616-91608616 on Build GRCh38
rs1743159 Chr.14: 91608201-91608201 on Build GRCh38
rs772952516 Chr.14: 91608381-91608381 on Build GRCh38
rs573082530 Chr.14: 91608209-91608209 on Build GRCh38
rs373076372 Chr.14: 91608243-91608243 on Build GRCh38
rs749950028 Chr.14: 91608257-91608257 on Build GRCh38
rs559323706 Chr.14: 91608202-91608202 on Build GRCh38
rs768482392 Chr.14: 91608367-91608367 on Build GRCh38
rs1743158 Chr.14: 91608512-91608512 on Build GRCh38
rs370063885 Chr.14: 91608446-91608446 on Build GRCh38
rs145756595 Chr.14: 91608591-91608591 on Build GRCh38
rs374427455 Chr.14: 91608380-91608380 on Build GRCh38
rs760298597 Chr.14: 91608394-91608394 on Build GRCh38
rs765674020 Chr.14: 91608254-91608254 on Build GRCh38
rs186257983 Chr.14: 91608458-91608458 on Build GRCh38
rs772346645 Chr.14: 91608310-91608311 on Build GRCh38
rs752000847 Chr.14: 91608438-91608438 on Build GRCh38
rs1025396970 Chr.14: 91608506-91608506 on Build GRCh38
rs950594664 Chr.14: 91608526-91608526 on Build GRCh38
rs528666799 Chr.14: 91608513-91608513 on Build GRCh38
rs753071709 Chr.14: 91608259-91608259 on Build GRCh38
rs747887662 Chr.14: 91608376-91608376 on Build GRCh38
rs907689420 Chr.14: 91608487-91608487 on Build GRCh38
rs375076511 Chr.14: 91608289-91608289 on Build GRCh38
rs543949772 Chr.14: 91608576-91608576 on Build GRCh38
rs774159492 Chr.14: 91608373-91608373 on Build GRCh38
rs552827013 Chr.14: 91608490-91608490 on Build GRCh38
rs902703489 Chr.14: 91608378-91608378 on Build GRCh38
rs189704096 Chr.14: 91608244-91608244 on Build GRCh38
rs550867012 Chr.14: 91608512-91608512 on Build GRCh38
rs781103559 Chr.14: 91608456-91608456 on Build GRCh38
rs891943751 Chr.14: 91608607-91608607 on Build GRCh38
rs772050007 Chr.14: 91608527-91608527 on Build GRCh38
rs749359208 Chr.14: 91608515-91608515 on Build GRCh38
rs933578412 Chr.14: 91608217-91608217 on Build GRCh38
rs974480410 Chr.14: 91608566-91608566 on Build GRCh38
rs943130752 Chr.14: 91608181-91608181 on Build GRCh38
rs755425864 Chr.14: 91608249-91608249 on Build GRCh38
rs909038827 Chr.14: 91608549-91608549 on Build GRCh38

View Full Product Details