Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 92130810-92131336 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPSF2
Entrez Gene ID
53981
Gene Name
cleavage and polyadenylation specific factor 2
Gene Aliases
CPSF100
UniGene
Hs.657632
Gene Chromosome Location
Chr.14: 92121954-92164199 on Build GRCh38
Transcripts
NM_001322270.1 NM_001322271.1 NM_001322272.1 NM_017437.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1371597 c.137A>C p.D46A Chr.14: 92131121-92131121 on Build GRCh38
4745692 c.147G>A p.R49R Chr.14: 92131131-92131131 on Build GRCh38
1371596 c.88delT p.L31fs*18 Chr.14: 92131072-92131072 on Build GRCh38
6634839 c.142C>A p.L48M Chr.14: 92131126-92131126 on Build GRCh38
6634847 c.24T>C p.T8T Chr.14: 92131008-92131008 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751718041 Chr.14: 92130942-92130942 on Build GRCh38
rs557411832 Chr.14: 92131203-92131203 on Build GRCh38
rs778665411 Chr.14: 92131302-92131302 on Build GRCh38
rs373779219 Chr.14: 92131062-92131062 on Build GRCh38
rs745971089 Chr.14: 92130978-92130978 on Build GRCh38
rs750505948 Chr.14: 92131100-92131100 on Build GRCh38
rs770435353 Chr.14: 92131012-92131012 on Build GRCh38
rs560501389 Chr.14: 92130970-92130970 on Build GRCh38
rs3759722 Chr.14: 92130884-92130884 on Build GRCh38
rs141334420 Chr.14: 92131004-92131004 on Build GRCh38
rs772147060 Chr.14: 92130977-92130978 on Build GRCh38
rs769414774 Chr.14: 92131055-92131055 on Build GRCh38
rs962618582 Chr.14: 92131309-92131309 on Build GRCh38
rs757295450 Chr.14: 92130951-92130951 on Build GRCh38
rs779584361 Chr.14: 92131178-92131178 on Build GRCh38
rs757138374 Chr.14: 92131154-92131154 on Build GRCh38
rs566265535 Chr.14: 92130990-92130990 on Build GRCh38
rs913632483 Chr.14: 92130930-92130930 on Build GRCh38
rs894468496 Chr.14: 92130958-92130958 on Build GRCh38
rs976255523 Chr.14: 92130907-92130907 on Build GRCh38
rs532887410 Chr.14: 92130887-92130887 on Build GRCh38
rs570093362 Chr.14: 92130885-92130885 on Build GRCh38
rs993614206 Chr.14: 92130872-92130872 on Build GRCh38
rs745544414 Chr.14: 92131046-92131046 on Build GRCh38
rs868242441 Chr.14: 92130854-92130854 on Build GRCh38
rs370678895 Chr.14: 92130952-92130952 on Build GRCh38
rs925227748 Chr.14: 92131039-92131039 on Build GRCh38
rs555135819 Chr.14: 92131155-92131155 on Build GRCh38
rs751484576 Chr.14: 92131151-92131151 on Build GRCh38
rs147996566 Chr.14: 92131126-92131126 on Build GRCh38
rs762366166 Chr.14: 92131093-92131093 on Build GRCh38
rs982603626 Chr.14: 92130929-92130929 on Build GRCh38
rs537715650 Chr.14: 92131173-92131173 on Build GRCh38
rs761015090 Chr.14: 92131108-92131108 on Build GRCh38
rs766707142 Chr.14: 92131112-92131112 on Build GRCh38
rs534953051 Chr.14: 92131101-92131101 on Build GRCh38
rs756352809 Chr.14: 92130966-92130966 on Build GRCh38
rs758145314 Chr.14: 92131148-92131148 on Build GRCh38
rs1045841047 Chr.14: 92131020-92131020 on Build GRCh38
rs543399495 Chr.14: 92130836-92130836 on Build GRCh38
rs192734765 Chr.14: 92131262-92131262 on Build GRCh38
rs117934581 Chr.14: 92130898-92130898 on Build GRCh38
rs922392749 Chr.14: 92130897-92130897 on Build GRCh38
rs777562704 Chr.14: 92130994-92130994 on Build GRCh38
rs776453284 Chr.14: 92131142-92131142 on Build GRCh38
rs1007779128 Chr.14: 92131249-92131249 on Build GRCh38
rs201839826 Chr.14: 92131168-92131168 on Build GRCh38
rs952296133 Chr.14: 92130886-92130886 on Build GRCh38
rs763212353 Chr.14: 92131072-92131072 on Build GRCh38
rs986307372 Chr.14: 92130888-92130888 on Build GRCh38
rs910226931 Chr.14: 92130894-92130894 on Build GRCh38
rs368745796 Chr.14: 92130973-92130973 on Build GRCh38
rs187443587 Chr.14: 92131246-92131246 on Build GRCh38
rs1017897121 Chr.14: 92131147-92131147 on Build GRCh38
rs776080823 Chr.14: 92131022-92131022 on Build GRCh38
rs962549091 Chr.14: 92130870-92130870 on Build GRCh38
rs893358614 Chr.14: 92131088-92131088 on Build GRCh38
rs963726811 Chr.14: 92130891-92130894 on Build GRCh38
rs966445446 Chr.14: 92131312-92131312 on Build GRCh38
rs750702090 Chr.14: 92130957-92130957 on Build GRCh38
rs748113258 Chr.14: 92131311-92131311 on Build GRCh38
rs1027782369 Chr.14: 92131217-92131217 on Build GRCh38
rs774724211 Chr.14: 92131079-92131079 on Build GRCh38
rs993667357 Chr.14: 92131204-92131204 on Build GRCh38
rs752598231 Chr.14: 92131124-92131124 on Build GRCh38
rs1017831507 Chr.14: 92131259-92131259 on Build GRCh38
rs372122742 Chr.14: 92131156-92131156 on Build GRCh38
rs887913035 Chr.14: 92131220-92131220 on Build GRCh38
rs367619373 Chr.14: 92131078-92131078 on Build GRCh38
rs767723845 Chr.14: 92131094-92131094 on Build GRCh38

View Full Product Details