Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 92135207-92135743 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPSF2
Entrez Gene ID
53981
Gene Name
cleavage and polyadenylation specific factor 2
Gene Aliases
CPSF100
UniGene
Hs.657632
Gene Chromosome Location
Chr.14: 92121954-92164199 on Build GRCh38
Transcripts
NM_001322270.1 NM_001322271.1 NM_001322272.1 NM_017437.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7247883 c.505G>C p.E169Q Chr.14: 92135456-92135456 on Build GRCh38
6634842 c.447G>A p.L149L Chr.14: 92135398-92135398 on Build GRCh38
958855 c.498_500delAGA p.E169delE Chr.14: 92135449-92135451 on Build GRCh38
4053031 c.463A>G p.I155V Chr.14: 92135414-92135414 on Build GRCh38
7178920 c.445C>T p.L149L Chr.14: 92135396-92135396 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1035940387 Chr.14: 92135683-92135683 on Build GRCh38
rs1015562303 Chr.14: 92135325-92135325 on Build GRCh38
rs576318811 Chr.14: 92135260-92135260 on Build GRCh38
rs905609157 Chr.14: 92135653-92135653 on Build GRCh38
rs371213615 Chr.14: 92135387-92135387 on Build GRCh38
rs935254233 Chr.14: 92135678-92135678 on Build GRCh38
rs141273633 Chr.14: 92135537-92135539 on Build GRCh38
rs747388084 Chr.14: 92135639-92135639 on Build GRCh38
rs562890303 Chr.14: 92135233-92135233 on Build GRCh38
rs769945857 Chr.14: 92135352-92135352 on Build GRCh38
rs773638216 Chr.14: 92135532-92135532 on Build GRCh38
rs377010120 Chr.14: 92135470-92135470 on Build GRCh38
rs961335639 Chr.14: 92135370-92135370 on Build GRCh38
rs1018347288 Chr.14: 92135284-92135284 on Build GRCh38
rs773205596 Chr.14: 92135477-92135478 on Build GRCh38
rs965559833 Chr.14: 92135304-92135307 on Build GRCh38
rs893799000 Chr.14: 92135720-92135720 on Build GRCh38
rs773053662 Chr.14: 92135398-92135398 on Build GRCh38
rs199953105 Chr.14: 92135540-92135540 on Build GRCh38
rs147491880 Chr.14: 92135625-92135625 on Build GRCh38
rs941711773 Chr.14: 92135475-92135475 on Build GRCh38
rs768832098 Chr.14: 92135396-92135396 on Build GRCh38
rs746292594 Chr.14: 92135347-92135347 on Build GRCh38
rs373766132 Chr.14: 92135596-92135596 on Build GRCh38
rs757655378 Chr.14: 92135545-92135545 on Build GRCh38
rs757773190 Chr.14: 92135334-92135334 on Build GRCh38
rs541132206 Chr.14: 92135395-92135395 on Build GRCh38
rs142973349 Chr.14: 92135318-92135318 on Build GRCh38
rs912693149 Chr.14: 92135589-92135589 on Build GRCh38
rs548888409 Chr.14: 92135314-92135314 on Build GRCh38
rs1001252580 Chr.14: 92135655-92135655 on Build GRCh38
rs369818860 Chr.14: 92135718-92135718 on Build GRCh38
rs1053334787 Chr.14: 92135620-92135620 on Build GRCh38
rs747345976 Chr.14: 92135319-92135319 on Build GRCh38
rs759355034 Chr.14: 92135461-92135461 on Build GRCh38
rs949225671 Chr.14: 92135584-92135584 on Build GRCh38
rs1035651447 Chr.14: 92135292-92135292 on Build GRCh38
rs138383589 Chr.14: 92135701-92135701 on Build GRCh38
rs770570776 Chr.14: 92135418-92135418 on Build GRCh38
rs762735556 Chr.14: 92135498-92135498 on Build GRCh38
rs559564189 Chr.14: 92135595-92135595 on Build GRCh38
rs959951197 Chr.14: 92135312-92135312 on Build GRCh38
rs545421179 Chr.14: 92135268-92135268 on Build GRCh38
rs752237287 Chr.14: 92135476-92135476 on Build GRCh38
rs374102493 Chr.14: 92135541-92135543 on Build GRCh38
rs377764376 Chr.14: 92135383-92135383 on Build GRCh38
rs760502958 Chr.14: 92135406-92135406 on Build GRCh38
rs564141531 Chr.14: 92135615-92135615 on Build GRCh38
rs891492388 Chr.14: 92135621-92135621 on Build GRCh38
rs1042848340 Chr.14: 92135641-92135641 on Build GRCh38
rs202153810 Chr.14: 92135437-92135437 on Build GRCh38
rs781716502 Chr.14: 92135340-92135340 on Build GRCh38
rs919990875 Chr.14: 92135491-92135491 on Build GRCh38
rs983081687 Chr.14: 92135389-92135389 on Build GRCh38
rs776639072 Chr.14: 92135419-92135419 on Build GRCh38
rs561075269 Chr.14: 92135567-92135567 on Build GRCh38
rs986026730 Chr.14: 92135552-92135552 on Build GRCh38

View Full Product Details