Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 94110654-94111134 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFI27
Entrez Gene ID
3429
Gene Name
interferon alpha inducible protein 27
Gene Aliases
FAM14D, ISG12, ISG12A, P27
UniGene
Hs.532634
Gene Chromosome Location
Chr.14: 94110733-94116699 on Build GRCh38
Transcripts
NM_001130080.2 NM_001288952.1 NM_001288954.1 NM_001288956.1 NM_001288957.1 NM_001288958.1 NM_001288959.1 NM_001288960.1 NM_001288995.1 NM_005532.4

SNP Detail

SNP ID Chromosome Location
rs941351803 Chr.14: 94110842-94110842 on Build GRCh38
rs956633445 Chr.14: 94110937-94110937 on Build GRCh38
rs914976186 Chr.14: 94110788-94110788 on Build GRCh38
rs755587716 Chr.14: 94110845-94110845 on Build GRCh38
rs573122011 Chr.14: 94110949-94110949 on Build GRCh38
rs899903570 Chr.14: 94110873-94110873 on Build GRCh38
rs185177063 Chr.14: 94110906-94110906 on Build GRCh38
rs980934630 Chr.14: 94111055-94111055 on Build GRCh38
rs962708675 Chr.14: 94110844-94110844 on Build GRCh38
rs758808779 Chr.14: 94111031-94111031 on Build GRCh38
rs997351957 Chr.14: 94110882-94110882 on Build GRCh38
rs1029826158 Chr.14: 94110829-94110829 on Build GRCh38
rs1008765610 Chr.14: 94110900-94110900 on Build GRCh38
rs948687458 Chr.14: 94110883-94110883 on Build GRCh38
rs779417810 Chr.14: 94110899-94110899 on Build GRCh38
rs1052540655 Chr.14: 94110950-94110950 on Build GRCh38
rs1043758077 Chr.14: 94110973-94110973 on Build GRCh38
rs908779511 Chr.14: 94111085-94111085 on Build GRCh38
rs777246259 Chr.14: 94111079-94111079 on Build GRCh38
rs756098003 Chr.14: 94110755-94110755 on Build GRCh38
rs989174378 Chr.14: 94110776-94110776 on Build GRCh38
rs955119475 Chr.14: 94110838-94110838 on Build GRCh38
rs947804700 Chr.14: 94110789-94110789 on Build GRCh38
rs927776591 Chr.14: 94111061-94111061 on Build GRCh38
rs1011134315 Chr.14: 94110786-94110786 on Build GRCh38
rs1047757220 Chr.14: 94110828-94110828 on Build GRCh38
rs981269577 Chr.14: 94110901-94110901 on Build GRCh38
rs74617881 Chr.14: 94110841-94110841 on Build GRCh38
rs531639994 Chr.14: 94110848-94110858 on Build GRCh38
rs375776183 Chr.14: 94110804-94110804 on Build GRCh38
rs879525307 Chr.14: 94110849-94110849 on Build GRCh38
rs181658839 Chr.14: 94110868-94110868 on Build GRCh38
rs900083081 Chr.14: 94110993-94110993 on Build GRCh38
rs575193576 Chr.14: 94110834-94110834 on Build GRCh38
rs560614555 Chr.14: 94110727-94110727 on Build GRCh38
rs939582914 Chr.14: 94110826-94110826 on Build GRCh38
rs892465551 Chr.14: 94110756-94110756 on Build GRCh38
rs368767195 Chr.14: 94110807-94110807 on Build GRCh38
rs1050235549 Chr.14: 94110893-94110893 on Build GRCh38
rs980935918 Chr.14: 94110792-94110792 on Build GRCh38
rs542621119 Chr.14: 94110968-94110968 on Build GRCh38
rs946696039 Chr.14: 94110970-94110970 on Build GRCh38
rs550054199 Chr.14: 94111089-94111089 on Build GRCh38
rs564084600 Chr.14: 94110680-94110680 on Build GRCh38
rs554609222 Chr.14: 94111062-94111062 on Build GRCh38
rs750670515 Chr.14: 94110704-94110704 on Build GRCh38
rs565412086 Chr.14: 94110989-94110989 on Build GRCh38
rs746706565 Chr.14: 94111088-94111088 on Build GRCh38
rs1030632266 Chr.14: 94110731-94110731 on Build GRCh38
rs1038303569 Chr.14: 94111111-94111111 on Build GRCh38
rs766508023 Chr.14: 94110770-94110770 on Build GRCh38
rs969132467 Chr.14: 94111030-94111030 on Build GRCh38

View Full Product Details