Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 94111547-94112056 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFI27
Entrez Gene ID
3429
Gene Name
interferon alpha inducible protein 27
Gene Aliases
FAM14D, ISG12, ISG12A, P27
UniGene
Hs.532634
Gene Chromosome Location
Chr.14: 94110733-94116699 on Build GRCh38
Transcripts
NM_001130080.2 NM_001288952.1 NM_001288954.1 NM_001288956.1 NM_001288957.1 NM_001288958.1 NM_001288959.1 NM_001288960.1 NM_001288995.1 NM_005532.4

SNP Detail

SNP ID Chromosome Location
rs539898593 Chr.14: 94112013-94112013 on Build GRCh38
rs117317777 Chr.14: 94111874-94111874 on Build GRCh38
rs937884655 Chr.14: 94111825-94111825 on Build GRCh38
rs759450912 Chr.14: 94111653-94111653 on Build GRCh38
rs765212537 Chr.14: 94111742-94111742 on Build GRCh38
rs745383972 Chr.14: 94111681-94111681 on Build GRCh38
rs201077623 Chr.14: 94111785-94111785 on Build GRCh38
rs574728148 Chr.14: 94111984-94111984 on Build GRCh38
rs749624037 Chr.14: 94111699-94111699 on Build GRCh38
rs774718963 Chr.14: 94111702-94111702 on Build GRCh38
rs1021558447 Chr.14: 94111972-94111972 on Build GRCh38
rs760720351 Chr.14: 94111740-94111740 on Build GRCh38
rs533941006 Chr.14: 94111849-94111849 on Build GRCh38
rs956301004 Chr.14: 94111969-94111969 on Build GRCh38
rs767134074 Chr.14: 94111797-94111797 on Build GRCh38
rs749744817 Chr.14: 94111808-94111808 on Build GRCh38
rs780690968 Chr.14: 94111677-94111677 on Build GRCh38
rs1243080 Chr.14: 94111842-94111842 on Build GRCh38
rs773280959 Chr.14: 94111737-94111737 on Build GRCh38
rs755620880 Chr.14: 94111804-94111804 on Build GRCh38
rs898275134 Chr.14: 94111664-94111664 on Build GRCh38
rs527985978 Chr.14: 94112030-94112030 on Build GRCh38
rs372591348 Chr.14: 94111771-94111771 on Build GRCh38
rs751211229 Chr.14: 94111675-94111675 on Build GRCh38
rs777534846 Chr.14: 94111674-94111674 on Build GRCh38
rs545942791 Chr.14: 94112033-94112033 on Build GRCh38
rs767583436 Chr.14: 94111633-94111633 on Build GRCh38
rs1028820700 Chr.14: 94112009-94112009 on Build GRCh38
rs556660609 Chr.14: 94111955-94111955 on Build GRCh38
rs939676300 Chr.14: 94111837-94111837 on Build GRCh38
rs752523703 Chr.14: 94111659-94111659 on Build GRCh38
rs961418489 Chr.14: 94112016-94112018 on Build GRCh38
rs147373518 Chr.14: 94111815-94111815 on Build GRCh38
rs141542083 Chr.14: 94111619-94111619 on Build GRCh38
rs779912529 Chr.14: 94111791-94111791 on Build GRCh38
rs748328072 Chr.14: 94111705-94111705 on Build GRCh38
rs114731937 Chr.14: 94111875-94111875 on Build GRCh38
rs954119100 Chr.14: 94112012-94112012 on Build GRCh38
rs537339688 Chr.14: 94111620-94111620 on Build GRCh38
rs1056499103 Chr.14: 94111597-94111597 on Build GRCh38
rs755509389 Chr.14: 94111810-94111810 on Build GRCh38
rs189690100 Chr.14: 94111826-94111826 on Build GRCh38
rs756994085 Chr.14: 94112032-94112032 on Build GRCh38
rs751149794 Chr.14: 94112018-94112018 on Build GRCh38
rs747077148 Chr.14: 94111818-94111818 on Build GRCh38
rs1010548521 Chr.14: 94111971-94111971 on Build GRCh38
rs904465396 Chr.14: 94111973-94111973 on Build GRCh38
rs562746213 Chr.14: 94111612-94111612 on Build GRCh38
rs1028505310 Chr.14: 94111693-94111693 on Build GRCh38
rs1008848067 Chr.14: 94111946-94111946 on Build GRCh38
rs937954359 Chr.14: 94111578-94111578 on Build GRCh38
rs750498326 Chr.14: 94111641-94111641 on Build GRCh38
rs765095423 Chr.14: 94111658-94111658 on Build GRCh38
rs756894747 Chr.14: 94111676-94111676 on Build GRCh38
rs866755649 Chr.14: 94111698-94111698 on Build GRCh38
rs2239644 Chr.14: 94111816-94111816 on Build GRCh38
rs987075349 Chr.14: 94111779-94111779 on Build GRCh38
rs112353168 Chr.14: 94111828-94111828 on Build GRCh38
rs1056785471 Chr.14: 94111884-94111884 on Build GRCh38
rs773370002 Chr.14: 94111817-94111817 on Build GRCh38
rs568750988 Chr.14: 94111833-94111833 on Build GRCh38
rs370109756 Chr.14: 94111632-94111632 on Build GRCh38
rs536277784 Chr.14: 94111841-94111841 on Build GRCh38
rs749946085 Chr.14: 94111801-94111801 on Build GRCh38
rs1050312640 Chr.14: 94111914-94111914 on Build GRCh38
rs772187601 Chr.14: 94111726-94111726 on Build GRCh38
rs374884974 Chr.14: 94111663-94111663 on Build GRCh38
rs544379897 Chr.14: 94111945-94111945 on Build GRCh38
rs560377866 Chr.14: 94112003-94112003 on Build GRCh38
rs138062603 Chr.14: 94111829-94111829 on Build GRCh38
rs968601715 Chr.14: 94111639-94111639 on Build GRCh38
rs780518012 Chr.14: 94111807-94111807 on Build GRCh38
rs770963065 Chr.14: 94111820-94111820 on Build GRCh38
rs769055213 Chr.14: 94111700-94111700 on Build GRCh38
rs541851181 Chr.14: 94111990-94111990 on Build GRCh38
rs775606082 Chr.14: 94111758-94111758 on Build GRCh38
rs1010164822 Chr.14: 94111598-94111598 on Build GRCh38
rs780494242 Chr.14: 94111696-94111696 on Build GRCh38
rs763009344 Chr.14: 94111767-94111767 on Build GRCh38
rs1028349542 Chr.14: 94111680-94111680 on Build GRCh38
rs548465908 Chr.14: 94111689-94111689 on Build GRCh38
rs956020204 Chr.14: 94111796-94111796 on Build GRCh38
rs891472678 Chr.14: 94111947-94111947 on Build GRCh38

View Full Product Details