Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 104734932-104735438 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADSSL1
Entrez Gene ID
122622
Gene Name
adenylosuccinate synthase like 1
Gene Aliases
-
UniGene
Hs.592327
Gene Chromosome Location
Chr.14: 104724197-104747325 on Build GRCh38
Transcripts
NM_001320424.1 NM_152328.4 NM_199165.2 XM_006720026.3 XM_011536412.1 XM_011536414.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6275961 c.322-7G>T p.? Chr.14: 104735013-104735013 on Build GRCh38
4967962 c.369C>A p.Y123* Chr.14: 104735067-104735067 on Build GRCh38
6859194 c.241G>A p.D81N Chr.14: 104735068-104735068 on Build GRCh38
5725576 c.399C>T p.I133I Chr.14: 104735097-104735097 on Build GRCh38
6595649 c.193-2_193-1insG p.? Chr.14: 104735018-104735019 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs576279934 Chr.14: 104735289-104735289 on Build GRCh38
rs66802280 Chr.14: 104735409-104735415 on Build GRCh38
rs761771211 Chr.14: 104735083-104735083 on Build GRCh38
rs555324958 Chr.14: 104734953-104734953 on Build GRCh38
rs759035711 Chr.14: 104734973-104734973 on Build GRCh38
rs1056362164 Chr.14: 104734982-104734982 on Build GRCh38
rs543621707 Chr.14: 104735206-104735206 on Build GRCh38
rs776595051 Chr.14: 104735091-104735093 on Build GRCh38
rs1053129877 Chr.14: 104735235-104735235 on Build GRCh38
rs368198688 Chr.14: 104735138-104735138 on Build GRCh38
rs970250330 Chr.14: 104735340-104735340 on Build GRCh38
rs763024737 Chr.14: 104735172-104735172 on Build GRCh38
rs569820135 Chr.14: 104735315-104735315 on Build GRCh38
rs925678568 Chr.14: 104735258-104735258 on Build GRCh38
rs985205024 Chr.14: 104735046-104735046 on Build GRCh38
rs1019288115 Chr.14: 104735287-104735287 on Build GRCh38
rs947274564 Chr.14: 104735376-104735376 on Build GRCh38
rs751442286 Chr.14: 104735002-104735002 on Build GRCh38
rs770565985 Chr.14: 104735042-104735042 on Build GRCh38
rs149077259 Chr.14: 104735075-104735075 on Build GRCh38
rs774438398 Chr.14: 104735152-104735152 on Build GRCh38
rs577790277 Chr.14: 104735133-104735133 on Build GRCh38
rs752768457 Chr.14: 104735171-104735171 on Build GRCh38
rs1011506740 Chr.14: 104735252-104735252 on Build GRCh38
rs779422330 Chr.14: 104735029-104735029 on Build GRCh38
rs755022185 Chr.14: 104735023-104735023 on Build GRCh38
rs913261169 Chr.14: 104735370-104735370 on Build GRCh38
rs775459928 Chr.14: 104735068-104735068 on Build GRCh38
rs991214992 Chr.14: 104735271-104735271 on Build GRCh38
rs145376192 Chr.14: 104735301-104735301 on Build GRCh38
rs938632385 Chr.14: 104735224-104735224 on Build GRCh38
rs765551699 Chr.14: 104735169-104735169 on Build GRCh38
rs766570941 Chr.14: 104735013-104735013 on Build GRCh38
rs948846262 Chr.14: 104734967-104734967 on Build GRCh38
rs932192901 Chr.14: 104735244-104735244 on Build GRCh38
rs938767711 Chr.14: 104735368-104735368 on Build GRCh38
rs73360636 Chr.14: 104734970-104734970 on Build GRCh38
rs774425831 Chr.14: 104735018-104735018 on Build GRCh38
rs745538776 Chr.14: 104735115-104735115 on Build GRCh38
rs7149183 Chr.14: 104735259-104735259 on Build GRCh38
rs377452057 Chr.14: 104735164-104735164 on Build GRCh38
rs766559463 Chr.14: 104735084-104735084 on Build GRCh38
rs772660842 Chr.14: 104735031-104735031 on Build GRCh38
rs747307365 Chr.14: 104735109-104735109 on Build GRCh38
rs563386751 Chr.14: 104735188-104735188 on Build GRCh38
rs185034782 Chr.14: 104735303-104735303 on Build GRCh38
rs141635495 Chr.14: 104735156-104735156 on Build GRCh38
rs371105407 Chr.14: 104735166-104735166 on Build GRCh38
rs528035794 Chr.14: 104735290-104735290 on Build GRCh38
rs765197637 Chr.14: 104735093-104735093 on Build GRCh38
rs367904065 Chr.14: 104735005-104735005 on Build GRCh38
rs768713180 Chr.14: 104735146-104735146 on Build GRCh38
rs753952847 Chr.14: 104735020-104735020 on Build GRCh38
rs866110938 Chr.14: 104735279-104735279 on Build GRCh38
rs200594013 Chr.14: 104735316-104735316 on Build GRCh38
rs541319189 Chr.14: 104735392-104735398 on Build GRCh38
rs765154977 Chr.14: 104734977-104734977 on Build GRCh38
rs755041450 Chr.14: 104735089-104735089 on Build GRCh38
rs762788472 Chr.14: 104734987-104734987 on Build GRCh38
rs764014811 Chr.14: 104734990-104734990 on Build GRCh38
rs965101613 Chr.14: 104735291-104735291 on Build GRCh38
rs891718199 Chr.14: 104735006-104735006 on Build GRCh38
rs1043012922 Chr.14: 104735377-104735377 on Build GRCh38
rs889666220 Chr.14: 104735260-104735260 on Build GRCh38
rs752819745 Chr.14: 104735095-104735095 on Build GRCh38
rs749516224 Chr.14: 104734980-104734980 on Build GRCh38
rs775304833 Chr.14: 104734980-104734980 on Build GRCh38
rs12885496 Chr.14: 104735276-104735276 on Build GRCh38
rs74089207 Chr.14: 104735043-104735043 on Build GRCh38
rs760706338 Chr.14: 104735018-104735019 on Build GRCh38
rs1053501668 Chr.14: 104735369-104735369 on Build GRCh38
rs143122131 Chr.14: 104735088-104735088 on Build GRCh38
rs558056310 Chr.14: 104735008-104735008 on Build GRCh38
rs773096183 Chr.14: 104735165-104735165 on Build GRCh38
rs181011783 Chr.14: 104735189-104735189 on Build GRCh38
rs994820123 Chr.14: 104735056-104735056 on Build GRCh38
rs564376552 Chr.14: 104735299-104735299 on Build GRCh38
rs889706927 Chr.14: 104735384-104735384 on Build GRCh38
rs771791003 Chr.14: 104735160-104735160 on Build GRCh38
rs764178731 Chr.14: 104735078-104735078 on Build GRCh38
rs777473775 Chr.14: 104735367-104735367 on Build GRCh38
rs768661926 Chr.14: 104735073-104735073 on Build GRCh38
rs149534850 Chr.14: 104735385-104735391 on Build GRCh38
rs148331541 Chr.14: 104735118-104735118 on Build GRCh38
rs532947317 Chr.14: 104735385-104735398 on Build GRCh38
rs916995355 Chr.14: 104735274-104735274 on Build GRCh38
rs1031625393 Chr.14: 104734995-104734995 on Build GRCh38
rs12589474 Chr.14: 104735288-104735288 on Build GRCh38
rs748866704 Chr.14: 104735139-104735139 on Build GRCh38
rs761935811 Chr.14: 104735034-104735034 on Build GRCh38
rs778086673 Chr.14: 104735035-104735035 on Build GRCh38
rs887836970 Chr.14: 104734959-104734959 on Build GRCh38
rs752358851 Chr.14: 104735302-104735302 on Build GRCh38
rs778317397 Chr.14: 104735108-104735108 on Build GRCh38
rs929431751 Chr.14: 104734952-104734952 on Build GRCh38
rs780850246 Chr.14: 104735111-104735111 on Build GRCh38
rs747429946 Chr.14: 104735036-104735036 on Build GRCh38
rs79940581 Chr.14: 104734968-104734968 on Build GRCh38
rs763476936 Chr.14: 104735222-104735222 on Build GRCh38
rs891684571 Chr.14: 104735246-104735246 on Build GRCh38
rs758891870 Chr.14: 104735106-104735106 on Build GRCh38
rs980276069 Chr.14: 104735343-104735343 on Build GRCh38
rs774276997 Chr.14: 104735079-104735079 on Build GRCh38
rs201199083 Chr.14: 104735052-104735052 on Build GRCh38
rs12885343 Chr.14: 104735298-104735298 on Build GRCh38
rs554970016 Chr.14: 104735110-104735110 on Build GRCh38
rs1006638858 Chr.14: 104735391-104735391 on Build GRCh38

View Full Product Details