Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 104739668-104740174 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADSSL1
Entrez Gene ID
122622
Gene Name
adenylosuccinate synthase like 1
Gene Aliases
-
UniGene
Hs.592327
Gene Chromosome Location
Chr.14: 104724197-104747325 on Build GRCh38
Transcripts
NM_001320424.1 NM_152328.4 NM_199165.2 XM_006720026.3 XM_011536412.1 XM_011536414.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1707915 c.571C>T p.Q191* Chr.14: 104739782-104739782 on Build GRCh38
366170 c.575A>T p.E192V Chr.14: 104739786-104739786 on Build GRCh38
7263908 c.539-6C>T p.? Chr.14: 104739744-104739744 on Build GRCh38
5659101 c.562G>C p.D188H Chr.14: 104739773-104739773 on Build GRCh38
7263909 c.410-6C>T p.? Chr.14: 104739744-104739744 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373708293 Chr.14: 104739819-104739819 on Build GRCh38
rs778986361 Chr.14: 104739776-104739776 on Build GRCh38
rs539452449 Chr.14: 104739968-104739968 on Build GRCh38
rs763665743 Chr.14: 104739732-104739732 on Build GRCh38
rs976673402 Chr.14: 104739996-104739996 on Build GRCh38
rs781372625 Chr.14: 104739844-104739844 on Build GRCh38
rs781694223 Chr.14: 104739738-104739738 on Build GRCh38
rs1014988246 Chr.14: 104739740-104739740 on Build GRCh38
rs762617781 Chr.14: 104739729-104739729 on Build GRCh38
rs56932269 Chr.14: 104739904-104739904 on Build GRCh38
rs569975694 Chr.14: 104739982-104739982 on Build GRCh38
rs1031744313 Chr.14: 104739810-104739810 on Build GRCh38
rs769295625 Chr.14: 104739951-104739951 on Build GRCh38
rs1053377979 Chr.14: 104740070-104740070 on Build GRCh38
rs376631598 Chr.14: 104739700-104739700 on Build GRCh38
rs1049069618 Chr.14: 104740033-104740033 on Build GRCh38
rs910704361 Chr.14: 104739983-104739983 on Build GRCh38
rs925679327 Chr.14: 104740071-104740071 on Build GRCh38
rs371322534 Chr.14: 104739830-104739830 on Build GRCh38
rs966706765 Chr.14: 104740135-104740135 on Build GRCh38
rs781756056 Chr.14: 104739782-104739782 on Build GRCh38
rs967566368 Chr.14: 104739850-104739850 on Build GRCh38
rs978922525 Chr.14: 104739859-104739859 on Build GRCh38
rs889486088 Chr.14: 104740130-104740130 on Build GRCh38
rs1021274682 Chr.14: 104739978-104739978 on Build GRCh38
rs375515624 Chr.14: 104739834-104739834 on Build GRCh38
rs567737049 Chr.14: 104739901-104739903 on Build GRCh38
rs919342835 Chr.14: 104739905-104739905 on Build GRCh38
rs534054807 Chr.14: 104739836-104739836 on Build GRCh38
rs1003125464 Chr.14: 104739946-104739946 on Build GRCh38
rs775758604 Chr.14: 104739721-104739721 on Build GRCh38
rs753181410 Chr.14: 104739793-104739794 on Build GRCh38
rs907118371 Chr.14: 104740055-104740055 on Build GRCh38
rs778197813 Chr.14: 104739838-104739841 on Build GRCh38
rs547501351 Chr.14: 104739845-104739845 on Build GRCh38
rs370357731 Chr.14: 104739747-104739747 on Build GRCh38
rs535290699 Chr.14: 104740076-104740076 on Build GRCh38
rs763230248 Chr.14: 104739802-104739802 on Build GRCh38
rs761377362 Chr.14: 104739741-104739741 on Build GRCh38
rs756576069 Chr.14: 104739862-104739862 on Build GRCh38
rs746337932 Chr.14: 104739789-104739789 on Build GRCh38
rs756530101 Chr.14: 104739804-104739805 on Build GRCh38
rs770225592 Chr.14: 104739714-104739714 on Build GRCh38
rs757584829 Chr.14: 104739842-104739842 on Build GRCh38
rs551294668 Chr.14: 104739889-104739889 on Build GRCh38
rs772588185 Chr.14: 104739909-104739909 on Build GRCh38
rs887680950 Chr.14: 104739926-104739926 on Build GRCh38
rs952548594 Chr.14: 104739806-104739806 on Build GRCh38
rs950312247 Chr.14: 104740126-104740126 on Build GRCh38
rs140388878 Chr.14: 104740131-104740131 on Build GRCh38
rs767426853 Chr.14: 104739744-104739744 on Build GRCh38
rs750406939 Chr.14: 104739745-104739745 on Build GRCh38
rs143062280 Chr.14: 104739773-104739773 on Build GRCh38
rs753593522 Chr.14: 104739772-104739772 on Build GRCh38
rs751871726 Chr.14: 104739838-104739838 on Build GRCh38
rs953120395 Chr.14: 104739977-104739977 on Build GRCh38
rs199615439 Chr.14: 104739753-104739753 on Build GRCh38
rs200636761 Chr.14: 104739703-104739703 on Build GRCh38
rs1044284593 Chr.14: 104740050-104740050 on Build GRCh38
rs948635259 Chr.14: 104740046-104740046 on Build GRCh38
rs377351431 Chr.14: 104739868-104739868 on Build GRCh38
rs770225306 Chr.14: 104739794-104739794 on Build GRCh38
rs867698726 Chr.14: 104740045-104740045 on Build GRCh38
rs189475831 Chr.14: 104739973-104739973 on Build GRCh38
rs536872881 Chr.14: 104739966-104739966 on Build GRCh38
rs771185141 Chr.14: 104739781-104739781 on Build GRCh38
rs146144477 Chr.14: 104739775-104739775 on Build GRCh38
rs751013239 Chr.14: 104739736-104739736 on Build GRCh38
rs907067828 Chr.14: 104739937-104739937 on Build GRCh38
rs555567061 Chr.14: 104740094-104740094 on Build GRCh38
rs994004646 Chr.14: 104739761-104739761 on Build GRCh38
rs370388695 Chr.14: 104739997-104739997 on Build GRCh38
rs1002687772 Chr.14: 104740060-104740060 on Build GRCh38
rs377420068 Chr.14: 104739828-104739828 on Build GRCh38
rs1017829808 Chr.14: 104740133-104740133 on Build GRCh38
rs1012860029 Chr.14: 104739932-104739932 on Build GRCh38
rs967066788 Chr.14: 104740044-104740044 on Build GRCh38
rs531771346 Chr.14: 104739690-104739690 on Build GRCh38
rs968253882 Chr.14: 104739980-104739980 on Build GRCh38
rs909248963 Chr.14: 104740043-104740043 on Build GRCh38
rs1049104223 Chr.14: 104739916-104739916 on Build GRCh38
rs555152004 Chr.14: 104739906-104739906 on Build GRCh38
rs1027532445 Chr.14: 104739755-104739755 on Build GRCh38
rs966197510 Chr.14: 104739987-104739987 on Build GRCh38
rs926071811 Chr.14: 104739869-104739869 on Build GRCh38
rs1044616946 Chr.14: 104739934-104739934 on Build GRCh38
rs768348409 Chr.14: 104739808-104739808 on Build GRCh38
rs929404329 Chr.14: 104740025-104740025 on Build GRCh38
rs57597030 Chr.14: 104740139-104740139 on Build GRCh38
rs777289653 Chr.14: 104739693-104739693 on Build GRCh38
rs71421880 Chr.14: 104739835-104739835 on Build GRCh38
rs1026505510 Chr.14: 104740140-104740140 on Build GRCh38
rs775866496 Chr.14: 104739795-104739795 on Build GRCh38
rs761287400 Chr.14: 104739827-104739827 on Build GRCh38
rs991631152 Chr.14: 104740085-104740085 on Build GRCh38
rs780490455 Chr.14: 104739863-104739863 on Build GRCh38
rs151233085 Chr.14: 104740113-104740113 on Build GRCh38

View Full Product Details