Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 104868343-104868848 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP170B
Entrez Gene ID
283638
Gene Name
centrosomal protein 170B
Gene Aliases
CEP170R, FAM68C, KIAA0284
UniGene
Hs.533721
Gene Chromosome Location
Chr.14: 104865313-104896770 on Build GRCh38
Transcripts
NM_001112726.2 NM_015005.2 XM_005267550.3 XM_011536665.2 XM_011536666.2 XM_017021227.1 XM_017021228.1 XM_017021229.1 XM_017021230.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5437075 c.50G>A p.R17Q Chr.14: 104868500-104868500 on Build GRCh38
953793 c.80G>A p.R27H Chr.14: 104868530-104868530 on Build GRCh38
4506801 c.72C>T p.F24F Chr.14: 104868522-104868522 on Build GRCh38
3753823 c.12G>A p.T4T Chr.14: 104868462-104868462 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs111847403 Chr.14: 104868782-104868782 on Build GRCh38
rs1007590218 Chr.14: 104868786-104868786 on Build GRCh38
rs776055514 Chr.14: 104868769-104868769 on Build GRCh38
rs916550587 Chr.14: 104868709-104868709 on Build GRCh38
rs918655632 Chr.14: 104868666-104868666 on Build GRCh38
rs1002224319 Chr.14: 104868703-104868703 on Build GRCh38
rs780455254 Chr.14: 104868516-104868516 on Build GRCh38
rs946919175 Chr.14: 104868796-104868796 on Build GRCh38
rs771693991 Chr.14: 104868597-104868597 on Build GRCh38
rs999716754 Chr.14: 104868432-104868432 on Build GRCh38
rs953256552 Chr.14: 104868742-104868742 on Build GRCh38
rs866749086 Chr.14: 104868632-104868632 on Build GRCh38
rs905416535 Chr.14: 104868685-104868685 on Build GRCh38
rs773985581 Chr.14: 104868565-104868565 on Build GRCh38
rs961888955 Chr.14: 104868738-104868738 on Build GRCh38
rs183217990 Chr.14: 104868462-104868462 on Build GRCh38
rs1014751713 Chr.14: 104868458-104868458 on Build GRCh38
rs528315851 Chr.14: 104868374-104868374 on Build GRCh38
rs984133858 Chr.14: 104868704-104868704 on Build GRCh38
rs985734954 Chr.14: 104868512-104868512 on Build GRCh38
rs954451460 Chr.14: 104868501-104868501 on Build GRCh38
rs1019849430 Chr.14: 104868794-104868794 on Build GRCh38
rs893571679 Chr.14: 104868768-104868768 on Build GRCh38
rs1046666907 Chr.14: 104868398-104868398 on Build GRCh38
rs1046307892 Chr.14: 104868723-104868723 on Build GRCh38
rs1015194831 Chr.14: 104868737-104868737 on Build GRCh38
rs770013504 Chr.14: 104868489-104868489 on Build GRCh38
rs758100184 Chr.14: 104868402-104868402 on Build GRCh38
rs576696102 Chr.14: 104868459-104868459 on Build GRCh38
rs1002317653 Chr.14: 104868368-104868368 on Build GRCh38
rs56752713 Chr.14: 104868630-104868630 on Build GRCh38
rs972838602 Chr.14: 104868665-104868665 on Build GRCh38
rs1012136704 Chr.14: 104868387-104868387 on Build GRCh38
rs12431850 Chr.14: 104868712-104868712 on Build GRCh38
rs550176070 Chr.14: 104868626-104868626 on Build GRCh38
rs764197790 Chr.14: 104868739-104868739 on Build GRCh38
rs531918050 Chr.14: 104868447-104868447 on Build GRCh38
rs906327648 Chr.14: 104868425-104868425 on Build GRCh38
rs1004296871 Chr.14: 104868731-104868731 on Build GRCh38
rs1030695546 Chr.14: 104868439-104868439 on Build GRCh38
rs1035117904 Chr.14: 104868419-104868419 on Build GRCh38
rs756650412 Chr.14: 104868490-104868490 on Build GRCh38
rs749355724 Chr.14: 104868609-104868609 on Build GRCh38
rs757265284 Chr.14: 104868461-104868461 on Build GRCh38
rs768914940 Chr.14: 104868561-104868561 on Build GRCh38
rs920287635 Chr.14: 104868486-104868486 on Build GRCh38
rs571039825 Chr.14: 104868530-104868530 on Build GRCh38
rs892635161 Chr.14: 104868654-104868654 on Build GRCh38
rs911555129 Chr.14: 104868781-104868781 on Build GRCh38
rs1052198253 Chr.14: 104868756-104868756 on Build GRCh38
rs747876545 Chr.14: 104868584-104868584 on Build GRCh38
rs555831447 Chr.14: 104868785-104868785 on Build GRCh38
rs533598431 Chr.14: 104868730-104868730 on Build GRCh38
rs368068386 Chr.14: 104868569-104868569 on Build GRCh38
rs774697488 Chr.14: 104868680-104868680 on Build GRCh38

View Full Product Details