Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 104878354-104878859 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP170B
Entrez Gene ID
283638
Gene Name
centrosomal protein 170B
Gene Aliases
CEP170R, FAM68C, KIAA0284
UniGene
Hs.533721
Gene Chromosome Location
Chr.14: 104865313-104896770 on Build GRCh38
Transcripts
NM_001112726.2 NM_015005.2 XM_005267550.3 XM_011536665.2 XM_011536666.2 XM_017021227.1 XM_017021228.1 XM_017021229.1 XM_017021230.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2026460 c.107C>T p.P36L Chr.14: 104878485-104878485 on Build GRCh38
6626915 c.68C>A p.S23Y Chr.14: 104878446-104878446 on Build GRCh38
696952 c.315C>T p.V105V Chr.14: 104878483-104878483 on Build GRCh38
4759771 c.77A>G p.Y26C Chr.14: 104878455-104878455 on Build GRCh38
4487310 c.106C>T p.P36S Chr.14: 104878484-104878484 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs528104117 Chr.14: 104878582-104878582 on Build GRCh38
rs41317300 Chr.14: 104878424-104878424 on Build GRCh38
rs904436286 Chr.14: 104878782-104878782 on Build GRCh38
rs376709112 Chr.14: 104878532-104878532 on Build GRCh38
rs932488233 Chr.14: 104878825-104878825 on Build GRCh38
rs541418376 Chr.14: 104878479-104878479 on Build GRCh38
rs920819451 Chr.14: 104878680-104878680 on Build GRCh38
rs1001178670 Chr.14: 104878783-104878783 on Build GRCh38
rs971849114 Chr.14: 104878746-104878746 on Build GRCh38
rs558096507 Chr.14: 104878806-104878806 on Build GRCh38
rs962302569 Chr.14: 104878675-104878675 on Build GRCh38
rs747849575 Chr.14: 104878395-104878395 on Build GRCh38
rs993695191 Chr.14: 104878679-104878679 on Build GRCh38
rs774713847 Chr.14: 104878509-104878509 on Build GRCh38
rs74637178 Chr.14: 104878441-104878441 on Build GRCh38
rs774932129 Chr.14: 104878433-104878433 on Build GRCh38
rs1033972300 Chr.14: 104878800-104878800 on Build GRCh38
rs535037964 Chr.14: 104878803-104878803 on Build GRCh38
rs757126815 Chr.14: 104878547-104878547 on Build GRCh38
rs571077045 Chr.14: 104878613-104878613 on Build GRCh38
rs991515234 Chr.14: 104878501-104878501 on Build GRCh38
rs372520258 Chr.14: 104878452-104878452 on Build GRCh38
rs374026981 Chr.14: 104878745-104878745 on Build GRCh38
rs762438192 Chr.14: 104878517-104878517 on Build GRCh38
rs563455681 Chr.14: 104878536-104878536 on Build GRCh38
rs981504533 Chr.14: 104878754-104878754 on Build GRCh38
rs1021508639 Chr.14: 104878777-104878777 on Build GRCh38
rs1034090219 Chr.14: 104878608-104878608 on Build GRCh38
rs377003510 Chr.14: 104878453-104878453 on Build GRCh38
rs184504308 Chr.14: 104878434-104878434 on Build GRCh38
rs756080658 Chr.14: 104878427-104878427 on Build GRCh38
rs774519664 Chr.14: 104878412-104878414 on Build GRCh38
rs754506872 Chr.14: 104878457-104878457 on Build GRCh38
rs1028094579 Chr.14: 104878629-104878629 on Build GRCh38
rs571103838 Chr.14: 104878402-104878402 on Build GRCh38
rs1054324365 Chr.14: 104878566-104878566 on Build GRCh38
rs972242656 Chr.14: 104878470-104878470 on Build GRCh38
rs749199842 Chr.14: 104878393-104878393 on Build GRCh38
rs7150561 Chr.14: 104878698-104878698 on Build GRCh38
rs145003340 Chr.14: 104878609-104878609 on Build GRCh38
rs776652931 Chr.14: 104878407-104878407 on Build GRCh38
rs367553168 Chr.14: 104878394-104878394 on Build GRCh38
rs202220553 Chr.14: 104878414-104878414 on Build GRCh38
rs768781898 Chr.14: 104878499-104878499 on Build GRCh38
rs554079568 Chr.14: 104878382-104878382 on Build GRCh38
rs187437760 Chr.14: 104878435-104878435 on Build GRCh38
rs931948378 Chr.14: 104878438-104878438 on Build GRCh38
rs58276039 Chr.14: 104878731-104878731 on Build GRCh38
rs890584150 Chr.14: 104878730-104878731 on Build GRCh38
rs373748193 Chr.14: 104878381-104878381 on Build GRCh38
rs372967151 Chr.14: 104878572-104878572 on Build GRCh38
rs993049435 Chr.14: 104878621-104878621 on Build GRCh38
rs548710255 Chr.14: 104878546-104878546 on Build GRCh38
rs924600501 Chr.14: 104878805-104878805 on Build GRCh38
rs1052497368 Chr.14: 104878739-104878739 on Build GRCh38
rs755811975 Chr.14: 104878450-104878450 on Build GRCh38
rs758126565 Chr.14: 104878478-104878478 on Build GRCh38
rs914469690 Chr.14: 104878815-104878815 on Build GRCh38
rs996591128 Chr.14: 104878383-104878383 on Build GRCh38
rs540088503 Chr.14: 104878668-104878668 on Build GRCh38
rs369953985 Chr.14: 104878460-104878460 on Build GRCh38
rs1014634519 Chr.14: 104878813-104878813 on Build GRCh38
rs1050401238 Chr.14: 104878706-104878706 on Build GRCh38
rs528958469 Chr.14: 104878542-104878542 on Build GRCh38
rs368570320 Chr.14: 104878425-104878425 on Build GRCh38
rs1015671099 Chr.14: 104878672-104878672 on Build GRCh38
rs1054358105 Chr.14: 104878812-104878812 on Build GRCh38
rs565423799 Chr.14: 104878569-104878569 on Build GRCh38
rs749582881 Chr.14: 104878551-104878551 on Build GRCh38
rs761058425 Chr.14: 104878521-104878521 on Build GRCh38
rs749318862 Chr.14: 104878498-104878498 on Build GRCh38
rs531511093 Chr.14: 104878722-104878723 on Build GRCh38
rs769984048 Chr.14: 104878419-104878419 on Build GRCh38
rs530686082 Chr.14: 104878744-104878744 on Build GRCh38
rs41314519 Chr.14: 104878486-104878486 on Build GRCh38
rs777402414 Chr.14: 104878543-104878543 on Build GRCh38
rs748122914 Chr.14: 104878475-104878475 on Build GRCh38
rs61995998 Chr.14: 104878535-104878535 on Build GRCh38
rs771149807 Chr.14: 104878592-104878595 on Build GRCh38
rs533226742 Chr.14: 104878520-104878520 on Build GRCh38
rs765966813 Chr.14: 104878522-104878522 on Build GRCh38
rs759824241 Chr.14: 104878552-104878552 on Build GRCh38
rs555930502 Chr.14: 104878747-104878747 on Build GRCh38
rs753502863 Chr.14: 104878526-104878526 on Build GRCh38
rs1007903366 Chr.14: 104878670-104878670 on Build GRCh38
rs780369145 Chr.14: 104878548-104878548 on Build GRCh38
rs572548629 Chr.14: 104878787-104878787 on Build GRCh38
rs962537716 Chr.14: 104878827-104878827 on Build GRCh38

View Full Product Details