Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 104883959-104884492 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP170B
Entrez Gene ID
283638
Gene Name
centrosomal protein 170B
Gene Aliases
CEP170R, FAM68C, KIAA0284
UniGene
Hs.533721
Gene Chromosome Location
Chr.14: 104865313-104896770 on Build GRCh38
Transcripts
NM_001112726.2 NM_015005.2 XM_005267550.3 XM_011536665.2 XM_011536666.2 XM_011536667.2 XM_017021227.1 XM_017021228.1 XM_017021229.1 XM_017021230.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1188926 c.1406G>A p.R469Q Chr.14: 104884185-104884185 on Build GRCh38
4621293 c.1450C>T p.R484* Chr.14: 104884229-104884229 on Build GRCh38
3711465 c.1147C>T p.Q383* Chr.14: 104884136-104884136 on Build GRCh38
6838712 c.1313C>G p.P438R Chr.14: 104884092-104884092 on Build GRCh38
6031675 c.1094G>A p.R365H Chr.14: 104884083-104884083 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367897923 Chr.14: 104884172-104884172 on Build GRCh38
rs761311635 Chr.14: 104884113-104884113 on Build GRCh38
rs772670141 Chr.14: 104884203-104884203 on Build GRCh38
rs765873690 Chr.14: 104884298-104884298 on Build GRCh38
rs770032638 Chr.14: 104884096-104884096 on Build GRCh38
rs750638092 Chr.14: 104884415-104884415 on Build GRCh38
rs537857490 Chr.14: 104884042-104884042 on Build GRCh38
rs947442535 Chr.14: 104884022-104884022 on Build GRCh38
rs763673496 Chr.14: 104884057-104884057 on Build GRCh38
rs929961846 Chr.14: 104884272-104884272 on Build GRCh38
rs368618168 Chr.14: 104884061-104884061 on Build GRCh38
rs372568174 Chr.14: 104884147-104884147 on Build GRCh38
rs760760618 Chr.14: 104884437-104884437 on Build GRCh38
rs765284336 Chr.14: 104884314-104884314 on Build GRCh38
rs765452990 Chr.14: 104884062-104884062 on Build GRCh38
rs756159243 Chr.14: 104884241-104884241 on Build GRCh38
rs752451112 Chr.14: 104883993-104883993 on Build GRCh38
rs760159993 Chr.14: 104884095-104884095 on Build GRCh38
rs769636798 Chr.14: 104884015-104884015 on Build GRCh38
rs546614335 Chr.14: 104884256-104884256 on Build GRCh38
rs760869423 Chr.14: 104884025-104884025 on Build GRCh38
rs202028986 Chr.14: 104884067-104884067 on Build GRCh38
rs763733382 Chr.14: 104884103-104884103 on Build GRCh38
rs773965520 Chr.14: 104884185-104884185 on Build GRCh38
rs369365096 Chr.14: 104884223-104884223 on Build GRCh38
rs372246157 Chr.14: 104884031-104884031 on Build GRCh38
rs777479560 Chr.14: 104884391-104884391 on Build GRCh38
rs755400640 Chr.14: 104883980-104883980 on Build GRCh38
rs753273180 Chr.14: 104884456-104884456 on Build GRCh38
rs554565193 Chr.14: 104884056-104884056 on Build GRCh38
rs997233687 Chr.14: 104884360-104884360 on Build GRCh38
rs753975341 Chr.14: 104884338-104884338 on Build GRCh38
rs989378605 Chr.14: 104884010-104884010 on Build GRCh38
rs749802920 Chr.14: 104884250-104884250 on Build GRCh38
rs906998074 Chr.14: 104884107-104884107 on Build GRCh38
rs964420919 Chr.14: 104884176-104884176 on Build GRCh38
rs370195295 Chr.14: 104884249-104884249 on Build GRCh38
rs757379746 Chr.14: 104884151-104884151 on Build GRCh38
rs777381966 Chr.14: 104883998-104883998 on Build GRCh38
rs9788593 Chr.14: 104884462-104884462 on Build GRCh38
rs1040944330 Chr.14: 104884041-104884041 on Build GRCh38
rs904969215 Chr.14: 104884321-104884321 on Build GRCh38
rs2582554 Chr.14: 104884322-104884322 on Build GRCh38
rs748445998 Chr.14: 104884017-104884017 on Build GRCh38
rs779002476 Chr.14: 104884016-104884016 on Build GRCh38
rs1009197594 Chr.14: 104884361-104884361 on Build GRCh38
rs568196034 Chr.14: 104884138-104884138 on Build GRCh38
rs747300597 Chr.14: 104884150-104884150 on Build GRCh38
rs201713998 Chr.14: 104884075-104884075 on Build GRCh38
rs779382893 Chr.14: 104884454-104884454 on Build GRCh38
rs982768113 Chr.14: 104884423-104884423 on Build GRCh38
rs745593453 Chr.14: 104884012-104884012 on Build GRCh38
rs373443985 Chr.14: 104884202-104884202 on Build GRCh38
rs752496892 Chr.14: 104884317-104884317 on Build GRCh38
rs745392077 Chr.14: 104884009-104884009 on Build GRCh38
rs951461065 Chr.14: 104884422-104884422 on Build GRCh38
rs991441590 Chr.14: 104884436-104884436 on Build GRCh38
rs550561242 Chr.14: 104884155-104884155 on Build GRCh38
rs538480548 Chr.14: 104884351-104884351 on Build GRCh38
rs748548390 Chr.14: 104884263-104884263 on Build GRCh38
rs867876630 Chr.14: 104884421-104884421 on Build GRCh38
rs770450389 Chr.14: 104884402-104884402 on Build GRCh38
rs776598460 Chr.14: 104884305-104884305 on Build GRCh38
rs564371749 Chr.14: 104884318-104884318 on Build GRCh38
rs377006406 Chr.14: 104884076-104884076 on Build GRCh38
rs768970440 Chr.14: 104884178-104884178 on Build GRCh38
rs568983781 Chr.14: 104884411-104884411 on Build GRCh38
rs755068276 Chr.14: 104884452-104884452 on Build GRCh38
rs745890787 Chr.14: 104884232-104884232 on Build GRCh38
rs1020511941 Chr.14: 104884242-104884242 on Build GRCh38
rs1000358416 Chr.14: 104884401-104884401 on Build GRCh38
rs750341823 Chr.14: 104884114-104884114 on Build GRCh38
rs373943144 Chr.14: 104884060-104884060 on Build GRCh38
rs754795299 Chr.14: 104884222-104884222 on Build GRCh38
rs1029720652 Chr.14: 104884418-104884418 on Build GRCh38
rs911268393 Chr.14: 104884139-104884139 on Build GRCh38
rs775870934 Chr.14: 104884099-104884099 on Build GRCh38
rs779401454 Chr.14: 104884378-104884378 on Build GRCh38
rs750666951 Chr.14: 104884326-104884326 on Build GRCh38
rs1030024880 Chr.14: 104884386-104884386 on Build GRCh38
rs756189724 Chr.14: 104884336-104884336 on Build GRCh38
rs778147308 Chr.14: 104884082-104884082 on Build GRCh38
rs761506010 Chr.14: 104884186-104884186 on Build GRCh38
rs748458503 Chr.14: 104884387-104884387 on Build GRCh38
rs983120860 Chr.14: 104884285-104884285 on Build GRCh38
rs775951197 Chr.14: 104884403-104884403 on Build GRCh38
rs751471747 Chr.14: 104884006-104884006 on Build GRCh38
rs766655499 Chr.14: 104884445-104884445 on Build GRCh38
rs771647102 Chr.14: 104884265-104884265 on Build GRCh38
rs752260545 Chr.14: 104884048-104884048 on Build GRCh38
rs1047547734 Chr.14: 104884471-104884471 on Build GRCh38
rs770446169 Chr.14: 104884465-104884465 on Build GRCh38
rs867273753 Chr.14: 104884374-104884374 on Build GRCh38
rs367947710 Chr.14: 104884218-104884218 on Build GRCh38
rs868330788 Chr.14: 104884463-104884463 on Build GRCh38
rs560991102 Chr.14: 104884177-104884177 on Build GRCh38
rs9788592 Chr.14: 104884143-104884143 on Build GRCh38
rs764045653 Chr.14: 104884414-104884414 on Build GRCh38
rs200188010 Chr.14: 104884032-104884032 on Build GRCh38
rs768989634 Chr.14: 104884255-104884255 on Build GRCh38
rs746678714 Chr.14: 104884396-104884396 on Build GRCh38
rs2841236 Chr.14: 104884018-104884018 on Build GRCh38
rs759144632 Chr.14: 104884408-104884408 on Build GRCh38
rs781591615 Chr.14: 104884230-104884230 on Build GRCh38
rs759437141 Chr.14: 104884307-104884307 on Build GRCh38
rs745952726 Chr.14: 104884156-104884156 on Build GRCh38
rs756033867 Chr.14: 104884116-104884116 on Build GRCh38
rs762826566 Chr.14: 104884412-104884412 on Build GRCh38
rs1012791679 Chr.14: 104884098-104884098 on Build GRCh38
rs771191178 Chr.14: 104884085-104884085 on Build GRCh38
rs573383969 Chr.14: 104884093-104884093 on Build GRCh38
rs751026610 Chr.14: 104884058-104884058 on Build GRCh38
rs755837654 Chr.14: 104884063-104884063 on Build GRCh38
rs538814557 Chr.14: 104884007-104884007 on Build GRCh38
rs772754598 Chr.14: 104884266-104884266 on Build GRCh38
rs747030766 Chr.14: 104884083-104884083 on Build GRCh38
rs772486458 Chr.14: 104884390-104884390 on Build GRCh38
rs201167874 Chr.14: 104883986-104883986 on Build GRCh38
rs368962009 Chr.14: 104884171-104884171 on Build GRCh38
rs753793628 Chr.14: 104884446-104884446 on Build GRCh38
rs780048597 Chr.14: 104884337-104884337 on Build GRCh38
rs537894408 Chr.14: 104884426-104884426 on Build GRCh38
rs751224174 Chr.14: 104884112-104884112 on Build GRCh38
rs1017278315 Chr.14: 104884343-104884343 on Build GRCh38
rs373744807 Chr.14: 104883997-104883997 on Build GRCh38
rs922583437 Chr.14: 104884167-104884167 on Build GRCh38
rs376115742 Chr.14: 104884410-104884410 on Build GRCh38
rs1043067523 Chr.14: 104884029-104884029 on Build GRCh38
rs780897550 Chr.14: 104884472-104884472 on Build GRCh38
rs376693361 Chr.14: 104884148-104884148 on Build GRCh38
rs765699127 Chr.14: 104884047-104884047 on Build GRCh38
rs143836703 Chr.14: 104884102-104884102 on Build GRCh38
rs758734296 Chr.14: 104884460-104884460 on Build GRCh38
rs918655591 Chr.14: 104884448-104884448 on Build GRCh38
rs777271652 Chr.14: 104884086-104884086 on Build GRCh38
rs112723134 Chr.14: 104884324-104884324 on Build GRCh38
rs900137573 Chr.14: 104884350-104884350 on Build GRCh38
rs760220838 Chr.14: 104884271-104884271 on Build GRCh38
rs753778950 Chr.14: 104884219-104884219 on Build GRCh38
rs773293486 Chr.14: 104884023-104884023 on Build GRCh38

View Full Product Details