Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 104885251-104885746 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP170B
Entrez Gene ID
283638
Gene Name
centrosomal protein 170B
Gene Aliases
CEP170R, FAM68C, KIAA0284
UniGene
Hs.533721
Gene Chromosome Location
Chr.14: 104865313-104896770 on Build GRCh38
Transcripts
NM_001112726.2 NM_015005.2 XM_005267550.3 XM_011536665.2 XM_011536666.2 XM_011536667.2 XM_017021227.1 XM_017021228.1 XM_017021229.1 XM_017021230.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6053070 c.1944+1G>T p.? Chr.14: 104885543-104885543 on Build GRCh38
4614734 c.1715delC p.Q574fs*179 Chr.14: 104885523-104885523 on Build GRCh38
6423466 c.1826G>A p.R609H Chr.14: 104885424-104885424 on Build GRCh38
1368570 c.1566T>C p.F522F Chr.14: 104885374-104885374 on Build GRCh38
4614733 c.1925delC p.Q644fs*179 Chr.14: 104885523-104885523 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766894227 Chr.14: 104885537-104885537 on Build GRCh38
rs974410956 Chr.14: 104885696-104885696 on Build GRCh38
rs768846975 Chr.14: 104885447-104885447 on Build GRCh38
rs375753389 Chr.14: 104885468-104885468 on Build GRCh38
rs921733652 Chr.14: 104885318-104885318 on Build GRCh38
rs538066245 Chr.14: 104885703-104885703 on Build GRCh38
rs770766563 Chr.14: 104885510-104885510 on Build GRCh38
rs563105820 Chr.14: 104885656-104885656 on Build GRCh38
rs754578462 Chr.14: 104885366-104885366 on Build GRCh38
rs200020098 Chr.14: 104885526-104885526 on Build GRCh38
rs964425871 Chr.14: 104885691-104885691 on Build GRCh38
rs868326344 Chr.14: 104885375-104885375 on Build GRCh38
rs370708323 Chr.14: 104885424-104885424 on Build GRCh38
rs1013515101 Chr.14: 104885604-104885604 on Build GRCh38
rs755789070 Chr.14: 104885341-104885341 on Build GRCh38
rs767627411 Chr.14: 104885565-104885565 on Build GRCh38
rs199847377 Chr.14: 104885361-104885361 on Build GRCh38
rs773755154 Chr.14: 104885528-104885528 on Build GRCh38
rs939487092 Chr.14: 104885625-104885625 on Build GRCh38
rs761805553 Chr.14: 104885523-104885523 on Build GRCh38
rs775974302 Chr.14: 104885618-104885618 on Build GRCh38
rs375255381 Chr.14: 104885485-104885485 on Build GRCh38
rs923838625 Chr.14: 104885568-104885568 on Build GRCh38
rs761135554 Chr.14: 104885474-104885474 on Build GRCh38
rs369486100 Chr.14: 104885557-104885557 on Build GRCh38
rs979200040 Chr.14: 104885662-104885662 on Build GRCh38
rs894038 Chr.14: 104885336-104885336 on Build GRCh38
rs974540093 Chr.14: 104885314-104885314 on Build GRCh38
rs772939384 Chr.14: 104885407-104885407 on Build GRCh38
rs750842766 Chr.14: 104885340-104885340 on Build GRCh38
rs749563676 Chr.14: 104885516-104885516 on Build GRCh38
rs922976500 Chr.14: 104885702-104885702 on Build GRCh38
rs779857675 Chr.14: 104885348-104885348 on Build GRCh38
rs963280649 Chr.14: 104885313-104885313 on Build GRCh38
rs984235265 Chr.14: 104885292-104885292 on Build GRCh38
rs552482779 Chr.14: 104885622-104885622 on Build GRCh38
rs887220810 Chr.14: 104885271-104885271 on Build GRCh38
rs764825244 Chr.14: 104885567-104885567 on Build GRCh38
rs761230284 Chr.14: 104885619-104885619 on Build GRCh38
rs766116440 Chr.14: 104885477-104885477 on Build GRCh38
rs752166832 Chr.14: 104885332-104885332 on Build GRCh38
rs867713193 Chr.14: 104885532-104885532 on Build GRCh38
rs774451983 Chr.14: 104885521-104885521 on Build GRCh38
rs748749918 Chr.14: 104885351-104885351 on Build GRCh38
rs1047348749 Chr.14: 104885634-104885634 on Build GRCh38
rs780249886 Chr.14: 104885513-104885513 on Build GRCh38
rs2582549 Chr.14: 104885300-104885300 on Build GRCh38
rs906713005 Chr.14: 104885620-104885620 on Build GRCh38
rs748159843 Chr.14: 104885396-104885396 on Build GRCh38
rs898576541 Chr.14: 104885560-104885560 on Build GRCh38
rs762927867 Chr.14: 104885427-104885427 on Build GRCh38
rs921784675 Chr.14: 104885547-104885547 on Build GRCh38
rs923641172 Chr.14: 104885401-104885401 on Build GRCh38
rs369239687 Chr.14: 104885462-104885462 on Build GRCh38
rs745492301 Chr.14: 104885317-104885317 on Build GRCh38
rs773915120 Chr.14: 104885530-104885530 on Build GRCh38
rs200493207 Chr.14: 104885508-104885508 on Build GRCh38
rs374120520 Chr.14: 104885473-104885473 on Build GRCh38
rs1053857470 Chr.14: 104885579-104885579 on Build GRCh38
rs960654929 Chr.14: 104885399-104885399 on Build GRCh38
rs568048526 Chr.14: 104885533-104885533 on Build GRCh38
rs752183142 Chr.14: 104885572-104885572 on Build GRCh38
rs777689043 Chr.14: 104885490-104885490 on Build GRCh38
rs371465096 Chr.14: 104885467-104885467 on Build GRCh38
rs746835504 Chr.14: 104885409-104885409 on Build GRCh38
rs754419769 Chr.14: 104885483-104885483 on Build GRCh38
rs1005963269 Chr.14: 104885495-104885495 on Build GRCh38
rs1002173558 Chr.14: 104885354-104885354 on Build GRCh38
rs532547844 Chr.14: 104885294-104885294 on Build GRCh38
rs778714638 Chr.14: 104885388-104885388 on Build GRCh38
rs369304576 Chr.14: 104885478-104885478 on Build GRCh38
rs763088776 Chr.14: 104885319-104885319 on Build GRCh38
rs771744040 Chr.14: 104885402-104885402 on Build GRCh38
rs764447204 Chr.14: 104885326-104885326 on Build GRCh38
rs746743939 Chr.14: 104885507-104885507 on Build GRCh38
rs966572118 Chr.14: 104885661-104885661 on Build GRCh38
rs940145107 Chr.14: 104885545-104885545 on Build GRCh38
rs1032084970 Chr.14: 104885665-104885665 on Build GRCh38
rs899818950 Chr.14: 104885722-104885722 on Build GRCh38
rs570464617 Chr.14: 104885520-104885520 on Build GRCh38
rs769933746 Chr.14: 104885413-104885413 on Build GRCh38
rs906662182 Chr.14: 104885480-104885480 on Build GRCh38
rs201922775 Chr.14: 104885484-104885484 on Build GRCh38
rs896421545 Chr.14: 104885603-104885603 on Build GRCh38
rs1044814004 Chr.14: 104885605-104885605 on Build GRCh38
rs1012456285 Chr.14: 104885471-104885471 on Build GRCh38
rs757556773 Chr.14: 104885334-104885334 on Build GRCh38
rs377213074 Chr.14: 104885437-104885437 on Build GRCh38

View Full Product Details