Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 34230422-34230936 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC12A6
Entrez Gene ID
9990
Gene Name
solute carrier family 12 member 6
Gene Aliases
ACCPN, KCC3, KCC3A, KCC3B
UniGene
Hs.510939
Gene Chromosome Location
Chr.15: 34229996-34338064 on Build GRCh38
Transcripts
NM_001042494.1 NM_001042495.1 NM_001042496.1 NM_001042497.1 NM_005135.2 NM_133647.1 XM_006720793.3

SNP Detail

SNP ID Chromosome Location
rs1055649737 Chr.15: 34230491-34230491 on Build GRCh38
rs1016405131 Chr.15: 34230458-34230458 on Build GRCh38
rs566619994 Chr.15: 34230572-34230572 on Build GRCh38
rs75128875 Chr.15: 34230520-34230520 on Build GRCh38
rs1009025986 Chr.15: 34230809-34230809 on Build GRCh38
rs555407270 Chr.15: 34230527-34230527 on Build GRCh38
rs954955121 Chr.15: 34230744-34230744 on Build GRCh38
rs558474157 Chr.15: 34230505-34230505 on Build GRCh38
rs886193153 Chr.15: 34230618-34230618 on Build GRCh38
rs961982793 Chr.15: 34230580-34230580 on Build GRCh38
rs4530104 Chr.15: 34230478-34230478 on Build GRCh38
rs1041398130 Chr.15: 34230498-34230498 on Build GRCh38
rs149756268 Chr.15: 34230826-34230827 on Build GRCh38
rs899564303 Chr.15: 34230789-34230789 on Build GRCh38
rs973388837 Chr.15: 34230634-34230635 on Build GRCh38
rs754832097 Chr.15: 34230842-34230842 on Build GRCh38
rs937465064 Chr.15: 34230673-34230673 on Build GRCh38
rs907913403 Chr.15: 34230480-34230480 on Build GRCh38
rs193075132 Chr.15: 34230801-34230801 on Build GRCh38
rs937141344 Chr.15: 34230489-34230489 on Build GRCh38
rs917125166 Chr.15: 34230717-34230718 on Build GRCh38
rs537054621 Chr.15: 34230523-34230523 on Build GRCh38
rs974988902 Chr.15: 34230465-34230465 on Build GRCh38
rs386782880 Chr.15: 34230572-34230574 on Build GRCh38
rs4238571 Chr.15: 34230784-34230784 on Build GRCh38
rs79634593 Chr.15: 34230481-34230481 on Build GRCh38
rs989227860 Chr.15: 34230746-34230746 on Build GRCh38
rs1013686083 Chr.15: 34230597-34230597 on Build GRCh38
rs545180509 Chr.15: 34230767-34230767 on Build GRCh38
rs74759842 Chr.15: 34230725-34230725 on Build GRCh38
rs996639834 Chr.15: 34230791-34230791 on Build GRCh38
rs1006008341 Chr.15: 34230904-34230904 on Build GRCh38
rs955119979 Chr.15: 34230570-34230570 on Build GRCh38
rs996458564 Chr.15: 34230681-34230681 on Build GRCh38
rs886051038 Chr.15: 34230719-34230719 on Build GRCh38
rs531291849 Chr.15: 34230821-34230821 on Build GRCh38
rs1029187609 Chr.15: 34230569-34230569 on Build GRCh38
rs963803396 Chr.15: 34230532-34230532 on Build GRCh38
rs759137272 Chr.15: 34230763-34230763 on Build GRCh38
rs560073124 Chr.15: 34230589-34230589 on Build GRCh38
rs549820750 Chr.15: 34230841-34230841 on Build GRCh38
rs991629008 Chr.15: 34230845-34230845 on Build GRCh38
rs777445922 Chr.15: 34230455-34230455 on Build GRCh38
rs964959746 Chr.15: 34230464-34230464 on Build GRCh38
rs573818109 Chr.15: 34230573-34230573 on Build GRCh38
rs969248179 Chr.15: 34230800-34230800 on Build GRCh38
rs996726509 Chr.15: 34230554-34230554 on Build GRCh38
rs544542527 Chr.15: 34230453-34230453 on Build GRCh38
rs949294510 Chr.15: 34230883-34230883 on Build GRCh38
rs923301056 Chr.15: 34230466-34230466 on Build GRCh38
rs143386876 Chr.15: 34230735-34230738 on Build GRCh38
rs1016480279 Chr.15: 34230643-34230643 on Build GRCh38
rs886102032 Chr.15: 34230902-34230902 on Build GRCh38
rs767424760 Chr.15: 34230510-34230510 on Build GRCh38
rs933303602 Chr.15: 34230467-34230467 on Build GRCh38
rs548190451 Chr.15: 34230509-34230513 on Build GRCh38
rs748705504 Chr.15: 34230855-34230855 on Build GRCh38
rs937966600 Chr.15: 34230550-34230550 on Build GRCh38
rs964866602 Chr.15: 34230665-34230665 on Build GRCh38
rs1056020638 Chr.15: 34230679-34230679 on Build GRCh38
rs765437783 Chr.15: 34230842-34230842 on Build GRCh38
rs1047851348 Chr.15: 34230887-34230887 on Build GRCh38
rs999836107 Chr.15: 34230758-34230758 on Build GRCh38
rs561741632 Chr.15: 34230859-34230859 on Build GRCh38
rs541154916 Chr.15: 34230574-34230574 on Build GRCh38
rs145421659 Chr.15: 34230841-34230842 on Build GRCh38
rs920686890 Chr.15: 34230487-34230487 on Build GRCh38
rs759659424 Chr.15: 34230825-34230825 on Build GRCh38
rs1025949476 Chr.15: 34230604-34230604 on Build GRCh38
rs1005928439 Chr.15: 34230637-34230637 on Build GRCh38
rs189231487 Chr.15: 34230493-34230493 on Build GRCh38

View Full Product Details