Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 34233393-34233912 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC12A6
Entrez Gene ID
9990
Gene Name
solute carrier family 12 member 6
Gene Aliases
ACCPN, KCC3, KCC3A, KCC3B
UniGene
Hs.510939
Gene Chromosome Location
Chr.15: 34229996-34338064 on Build GRCh38
Transcripts
NM_001042494.1 NM_001042495.1 NM_001042496.1 NM_001042497.1 NM_005135.2 NM_133647.1 XM_006720793.3

SNP Detail

SNP ID Chromosome Location
rs969900949 Chr.15: 34233843-34233843 on Build GRCh38
rs184521227 Chr.15: 34233880-34233880 on Build GRCh38
rs199625435 Chr.15: 34233834-34233834 on Build GRCh38
rs187536136 Chr.15: 34233675-34233675 on Build GRCh38
rs567450515 Chr.15: 34233815-34233815 on Build GRCh38
rs770725914 Chr.15: 34233717-34233717 on Build GRCh38
rs767632019 Chr.15: 34233856-34233856 on Build GRCh38
rs946379670 Chr.15: 34233613-34233613 on Build GRCh38
rs867239207 Chr.15: 34233450-34233450 on Build GRCh38
rs867412270 Chr.15: 34233620-34233620 on Build GRCh38
rs117540484 Chr.15: 34233787-34233787 on Build GRCh38
rs192377599 Chr.15: 34233632-34233632 on Build GRCh38
rs760605080 Chr.15: 34233857-34233857 on Build GRCh38
rs965883968 Chr.15: 34233575-34233575 on Build GRCh38
rs998431162 Chr.15: 34233770-34233770 on Build GRCh38
rs901423099 Chr.15: 34233740-34233740 on Build GRCh38
rs1018424714 Chr.15: 34233733-34233733 on Build GRCh38
rs183696226 Chr.15: 34233637-34233637 on Build GRCh38
rs540920981 Chr.15: 34233464-34233464 on Build GRCh38
rs34987939 Chr.15: 34233825-34233826 on Build GRCh38
rs914499094 Chr.15: 34233691-34233691 on Build GRCh38
rs561736527 Chr.15: 34233532-34233532 on Build GRCh38
rs778536148 Chr.15: 34233465-34233465 on Build GRCh38
rs985872003 Chr.15: 34233544-34233544 on Build GRCh38
rs201915253 Chr.15: 34233835-34233835 on Build GRCh38
rs912524811 Chr.15: 34233558-34233558 on Build GRCh38
rs909964316 Chr.15: 34233448-34233449 on Build GRCh38
rs934513827 Chr.15: 34233644-34233644 on Build GRCh38
rs56372103 Chr.15: 34233678-34233678 on Build GRCh38
rs978192828 Chr.15: 34233594-34233594 on Build GRCh38
rs892177326 Chr.15: 34233604-34233604 on Build GRCh38
rs746999362 Chr.15: 34233710-34233710 on Build GRCh38
rs372202037 Chr.15: 34233865-34233865 on Build GRCh38
rs530201839 Chr.15: 34233746-34233746 on Build GRCh38
rs771870110 Chr.15: 34233641-34233641 on Build GRCh38
rs1044495746 Chr.15: 34233748-34233748 on Build GRCh38
rs759376614 Chr.15: 34233851-34233851 on Build GRCh38
rs191955067 Chr.15: 34233850-34233850 on Build GRCh38
rs547741348 Chr.15: 34233678-34233678 on Build GRCh38
rs927847964 Chr.15: 34233515-34233515 on Build GRCh38
rs939279183 Chr.15: 34233533-34233533 on Build GRCh38
rs759449271 Chr.15: 34233418-34233418 on Build GRCh38
rs764086996 Chr.15: 34233864-34233864 on Build GRCh38
rs373099224 Chr.15: 34233651-34233651 on Build GRCh38
rs187535372 Chr.15: 34233425-34233425 on Build GRCh38

View Full Product Details