Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 34234905-34235407 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC12A6
Entrez Gene ID
9990
Gene Name
solute carrier family 12 member 6
Gene Aliases
ACCPN, KCC3, KCC3A, KCC3B
UniGene
Hs.510939
Gene Chromosome Location
Chr.15: 34229996-34338064 on Build GRCh38
Transcripts
NM_001042494.1 NM_001042495.1 NM_001042496.1 NM_001042497.1 NM_005135.2 NM_133647.1 XM_006720793.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7333345 c.3174A>T p.P1058P Chr.15: 34235215-34235215 on Build GRCh38
4946088 c.3183-1G>A p.? Chr.15: 34235315-34235315 on Build GRCh38
6342334 c.3246C>A p.N1082K Chr.15: 34235269-34235269 on Build GRCh38
5660477 c.3315C>T p.N1105N Chr.15: 34235200-34235200 on Build GRCh38
960805 c.3311G>A p.R1104Q Chr.15: 34235204-34235204 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770967705 Chr.15: 34235076-34235076 on Build GRCh38
rs768483323 Chr.15: 34235357-34235357 on Build GRCh38
rs779725347 Chr.15: 34235356-34235356 on Build GRCh38
rs918285694 Chr.15: 34234990-34234990 on Build GRCh38
rs755368320 Chr.15: 34235313-34235313 on Build GRCh38
rs1021037238 Chr.15: 34234973-34234973 on Build GRCh38
rs376575991 Chr.15: 34235329-34235329 on Build GRCh38
rs768514327 Chr.15: 34235205-34235205 on Build GRCh38
rs149640638 Chr.15: 34235293-34235293 on Build GRCh38
rs751908075 Chr.15: 34235136-34235136 on Build GRCh38
rs972638765 Chr.15: 34234954-34234954 on Build GRCh38
rs746758533 Chr.15: 34235058-34235058 on Build GRCh38
rs745740236 Chr.15: 34235200-34235200 on Build GRCh38
rs748818909 Chr.15: 34235160-34235160 on Build GRCh38
rs746796598 Chr.15: 34235204-34235204 on Build GRCh38
rs938212238 Chr.15: 34235319-34235319 on Build GRCh38
rs765311588 Chr.15: 34235265-34235265 on Build GRCh38
rs756624500 Chr.15: 34235337-34235337 on Build GRCh38
rs781705649 Chr.15: 34235152-34235152 on Build GRCh38
rs763190677 Chr.15: 34235276-34235276 on Build GRCh38
rs567361041 Chr.15: 34235294-34235294 on Build GRCh38
rs773205027 Chr.15: 34235269-34235269 on Build GRCh38
rs775446736 Chr.15: 34235194-34235194 on Build GRCh38
rs1029919719 Chr.15: 34235109-34235109 on Build GRCh38
rs758173206 Chr.15: 34235346-34235346 on Build GRCh38
rs866878301 Chr.15: 34235343-34235343 on Build GRCh38
rs767970540 Chr.15: 34235317-34235317 on Build GRCh38
rs905281040 Chr.15: 34235374-34235374 on Build GRCh38
rs756781352 Chr.15: 34235161-34235161 on Build GRCh38
rs776625055 Chr.15: 34235236-34235236 on Build GRCh38
rs748661070 Chr.15: 34235336-34235338 on Build GRCh38
rs1017964172 Chr.15: 34235006-34235006 on Build GRCh38
rs781655653 Chr.15: 34235143-34235144 on Build GRCh38
rs541875091 Chr.15: 34235057-34235057 on Build GRCh38
rs751913210 Chr.15: 34235302-34235302 on Build GRCh38
rs1042896680 Chr.15: 34235382-34235383 on Build GRCh38
rs778509280 Chr.15: 34235162-34235162 on Build GRCh38
rs560608306 Chr.15: 34235110-34235110 on Build GRCh38
rs1010877216 Chr.15: 34235348-34235348 on Build GRCh38
rs776229241 Chr.15: 34235365-34235365 on Build GRCh38
rs745521643 Chr.15: 34235175-34235175 on Build GRCh38
rs1057516752 Chr.15: 34235234-34235238 on Build GRCh38
rs778597105 Chr.15: 34235345-34235345 on Build GRCh38
rs1009618927 Chr.15: 34234956-34234956 on Build GRCh38
rs145628806 Chr.15: 34235001-34235001 on Build GRCh38
rs903720468 Chr.15: 34234997-34234997 on Build GRCh38
rs560558267 Chr.15: 34235151-34235151 on Build GRCh38
rs771810437 Chr.15: 34235188-34235188 on Build GRCh38
rs761854359 Chr.15: 34235262-34235262 on Build GRCh38
rs564731517 Chr.15: 34235373-34235373 on Build GRCh38
rs1056483234 Chr.15: 34235322-34235322 on Build GRCh38
rs983645707 Chr.15: 34235115-34235115 on Build GRCh38
rs776418777 Chr.15: 34235092-34235092 on Build GRCh38

View Full Product Details