Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 39250651-39251194 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C15orf54
Entrez Gene ID
400360
Gene Name
chromosome 15 open reading frame 54
Gene Aliases
-
UniGene
Hs.376109
Gene Chromosome Location
Chr.15: 39250669-39254845 on Build GRCh38
Transcripts
NM_001302797.1 NM_207445.3

SNP Detail

SNP ID Chromosome Location
rs896333926 Chr.15: 39251112-39251112 on Build GRCh38
rs535110343 Chr.15: 39250917-39250917 on Build GRCh38
rs549790889 Chr.15: 39250776-39250776 on Build GRCh38
rs749758834 Chr.15: 39250848-39250848 on Build GRCh38
rs971527027 Chr.15: 39250863-39250863 on Build GRCh38
rs944552871 Chr.15: 39250784-39250784 on Build GRCh38
rs974052404 Chr.15: 39250748-39250748 on Build GRCh38
rs1045959459 Chr.15: 39251053-39251053 on Build GRCh38
rs1010833038 Chr.15: 39250786-39250786 on Build GRCh38
rs943131482 Chr.15: 39250924-39250924 on Build GRCh38
rs987186078 Chr.15: 39251149-39251149 on Build GRCh38
rs1002269166 Chr.15: 39251093-39251093 on Build GRCh38
rs964354827 Chr.15: 39251143-39251143 on Build GRCh38
rs557250985 Chr.15: 39250933-39250933 on Build GRCh38
rs1026159756 Chr.15: 39251145-39251145 on Build GRCh38
rs921800984 Chr.15: 39250970-39250970 on Build GRCh38
rs530334922 Chr.15: 39250884-39250884 on Build GRCh38
rs1001816871 Chr.15: 39251079-39251079 on Build GRCh38
rs983356655 Chr.15: 39250942-39250942 on Build GRCh38
rs910272377 Chr.15: 39250726-39250726 on Build GRCh38
rs188643269 Chr.15: 39251158-39251158 on Build GRCh38
rs574300511 Chr.15: 39251117-39251117 on Build GRCh38
rs148790697 Chr.15: 39250730-39250730 on Build GRCh38
rs142440594 Chr.15: 39250751-39250751 on Build GRCh38
rs555800001 Chr.15: 39250834-39250834 on Build GRCh38
rs566070128 Chr.15: 39250809-39250809 on Build GRCh38
rs572529057 Chr.15: 39250912-39250912 on Build GRCh38
rs972025559 Chr.15: 39250843-39250843 on Build GRCh38
rs755131607 Chr.15: 39250683-39250683 on Build GRCh38
rs563796507 Chr.15: 39251099-39251099 on Build GRCh38
rs78974568 Chr.15: 39251138-39251138 on Build GRCh38
rs17613185 Chr.15: 39250718-39250718 on Build GRCh38
rs907435048 Chr.15: 39251074-39251074 on Build GRCh38
rs78280861 Chr.15: 39250761-39250761 on Build GRCh38
rs892676594 Chr.15: 39250951-39250951 on Build GRCh38
rs758016776 Chr.15: 39250859-39250859 on Build GRCh38
rs1034540659 Chr.15: 39250900-39250900 on Build GRCh38
rs2852077 Chr.15: 39250752-39250752 on Build GRCh38
rs16968537 Chr.15: 39250829-39250829 on Build GRCh38
rs762892137 Chr.15: 39251168-39251168 on Build GRCh38
rs569946597 Chr.15: 39250794-39250794 on Build GRCh38
rs1032669736 Chr.15: 39250836-39250836 on Build GRCh38
rs111982835 Chr.15: 39250988-39250988 on Build GRCh38
rs562658338 Chr.15: 39251021-39251021 on Build GRCh38
rs891493714 Chr.15: 39250837-39250837 on Build GRCh38
rs1001722116 Chr.15: 39250881-39250881 on Build GRCh38
rs760266363 Chr.15: 39250967-39250967 on Build GRCh38
rs998474353 Chr.15: 39250756-39250756 on Build GRCh38
rs765044535 Chr.15: 39251142-39251142 on Build GRCh38
rs796171913 Chr.15: 39250918-39250918 on Build GRCh38
rs891426354 Chr.15: 39251029-39251029 on Build GRCh38
rs1024473181 Chr.15: 39250682-39250682 on Build GRCh38
rs935749168 Chr.15: 39250973-39250973 on Build GRCh38
rs951386159 Chr.15: 39251148-39251148 on Build GRCh38
rs905325727 Chr.15: 39250800-39250800 on Build GRCh38
rs761413280 Chr.15: 39251060-39251060 on Build GRCh38
rs971975089 Chr.15: 39250715-39250715 on Build GRCh38
rs944647611 Chr.15: 39250968-39250968 on Build GRCh38
rs1052714184 Chr.15: 39250950-39250950 on Build GRCh38
rs775082227 Chr.15: 39250949-39250949 on Build GRCh38
rs151147213 Chr.15: 39250941-39250941 on Build GRCh38
rs1035492342 Chr.15: 39251103-39251103 on Build GRCh38
rs1010329511 Chr.15: 39251037-39251037 on Build GRCh38
rs922816705 Chr.15: 39250943-39250943 on Build GRCh38
rs776408775 Chr.15: 39250990-39250990 on Build GRCh38
rs951984298 Chr.15: 39251118-39251118 on Build GRCh38
rs984696121 Chr.15: 39251133-39251133 on Build GRCh38
rs912589101 Chr.15: 39251150-39251150 on Build GRCh38
rs561157528 Chr.15: 39250677-39250677 on Build GRCh38
rs1024985515 Chr.15: 39250849-39250849 on Build GRCh38
rs993704537 Chr.15: 39251114-39251114 on Build GRCh38
rs74010024 Chr.15: 39250919-39250919 on Build GRCh38
rs372703176 Chr.15: 39250892-39250892 on Build GRCh38
rs930283542 Chr.15: 39250693-39250693 on Build GRCh38

View Full Product Details