Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 48335954-48336456 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUT
Entrez Gene ID
1854
Gene Name
deoxyuridine triphosphatase
Gene Aliases
dUTPase
UniGene
Hs.527980
Gene Chromosome Location
Chr.15: 48331018-48343373 on Build GRCh38
Transcripts
NM_001025248.1 NM_001025249.1 NM_001948.3 XM_011521315.2 XM_017021988.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5017110 c.254G>T p.R85L Chr.15: 48336052-48336052 on Build GRCh38
6257363 c.556+1G>T p.? Chr.15: 48336091-48336091 on Build GRCh38
5017109 c.518G>T p.R173L Chr.15: 48336052-48336052 on Build GRCh38
6257364 c.292+1G>T p.? Chr.15: 48336091-48336091 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs758678340 Chr.15: 48336094-48336094 on Build GRCh38
rs139161403 Chr.15: 48336038-48336038 on Build GRCh38
rs766035223 Chr.15: 48336030-48336030 on Build GRCh38
rs201845064 Chr.15: 48336125-48336125 on Build GRCh38
rs752686917 Chr.15: 48336062-48336062 on Build GRCh38
rs942121289 Chr.15: 48336168-48336168 on Build GRCh38
rs781192278 Chr.15: 48336283-48336285 on Build GRCh38
rs956016757 Chr.15: 48336396-48336396 on Build GRCh38
rs781764281 Chr.15: 48336139-48336139 on Build GRCh38
rs918422585 Chr.15: 48336296-48336299 on Build GRCh38
rs1021751246 Chr.15: 48336418-48336418 on Build GRCh38
rs981279405 Chr.15: 48336334-48336337 on Build GRCh38
rs778967652 Chr.15: 48336069-48336069 on Build GRCh38
rs759363930 Chr.15: 48336040-48336040 on Build GRCh38
rs559298847 Chr.15: 48336164-48336164 on Build GRCh38
rs373308210 Chr.15: 48336058-48336058 on Build GRCh38
rs749514959 Chr.15: 48336217-48336219 on Build GRCh38
rs570075863 Chr.15: 48336403-48336403 on Build GRCh38
rs1011332107 Chr.15: 48336405-48336405 on Build GRCh38
rs769498074 Chr.15: 48336007-48336007 on Build GRCh38
rs937258803 Chr.15: 48336357-48336357 on Build GRCh38
rs568755619 Chr.15: 48336326-48336329 on Build GRCh38
rs780180352 Chr.15: 48336102-48336102 on Build GRCh38
rs202180121 Chr.15: 48336048-48336048 on Build GRCh38
rs747435087 Chr.15: 48336001-48336001 on Build GRCh38
rs927164826 Chr.15: 48336346-48336346 on Build GRCh38
rs764683220 Chr.15: 48336422-48336422 on Build GRCh38
rs7178868 Chr.15: 48336193-48336193 on Build GRCh38
rs1022851629 Chr.15: 48336280-48336280 on Build GRCh38
rs767416846 Chr.15: 48336051-48336051 on Build GRCh38
rs28381115 Chr.15: 48336329-48336332 on Build GRCh38
rs942993547 Chr.15: 48335995-48335995 on Build GRCh38
rs761397753 Chr.15: 48336006-48336006 on Build GRCh38
rs747776521 Chr.15: 48336037-48336038 on Build GRCh38
rs188372724 Chr.15: 48336309-48336309 on Build GRCh38
rs368983307 Chr.15: 48335994-48335994 on Build GRCh38
rs751396653 Chr.15: 48336032-48336032 on Build GRCh38
rs907114798 Chr.15: 48336333-48336333 on Build GRCh38
rs1047909162 Chr.15: 48336103-48336103 on Build GRCh38
rs757322113 Chr.15: 48336066-48336066 on Build GRCh38
rs1029162676 Chr.15: 48336132-48336136 on Build GRCh38
rs1054103452 Chr.15: 48336379-48336379 on Build GRCh38
rs530703944 Chr.15: 48336252-48336252 on Build GRCh38
rs780480318 Chr.15: 48335998-48335998 on Build GRCh38
rs7175137 Chr.15: 48336004-48336004 on Build GRCh38
rs200067199 Chr.15: 48336023-48336023 on Build GRCh38
rs750553095 Chr.15: 48336077-48336077 on Build GRCh38
rs28381116 Chr.15: 48336370-48336370 on Build GRCh38
rs1031501226 Chr.15: 48336393-48336393 on Build GRCh38
rs563632753 Chr.15: 48336225-48336225 on Build GRCh38
rs762683508 Chr.15: 48336026-48336026 on Build GRCh38
rs529869253 Chr.15: 48336187-48336187 on Build GRCh38
rs755891790 Chr.15: 48336136-48336137 on Build GRCh38

View Full Product Details