Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 49125510-49126021 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COPS2
Entrez Gene ID
9318
Gene Name
COP9 signalosome subunit 2
Gene Aliases
ALIEN, CSN2, SGN2, TRIP15
UniGene
Hs.369614
Gene Chromosome Location
Chr.15: 49125274-49155657 on Build GRCh38
Transcripts
NM_001143887.1 NM_004236.3

SNP Detail

SNP ID Chromosome Location
rs1052843470 Chr.15: 49125657-49125657 on Build GRCh38
rs891093375 Chr.15: 49125571-49125571 on Build GRCh38
rs968487807 Chr.15: 49125809-49125809 on Build GRCh38
rs951550935 Chr.15: 49125766-49125766 on Build GRCh38
rs1044542229 Chr.15: 49125566-49125566 on Build GRCh38
rs987783249 Chr.15: 49125850-49125850 on Build GRCh38
rs931357326 Chr.15: 49125909-49125909 on Build GRCh38
rs899953969 Chr.15: 49125727-49125727 on Build GRCh38
rs150263491 Chr.15: 49125957-49125957 on Build GRCh38
rs766835675 Chr.15: 49125865-49125865 on Build GRCh38
rs995624665 Chr.15: 49125736-49125736 on Build GRCh38
rs1039741460 Chr.15: 49125876-49125876 on Build GRCh38
rs530572728 Chr.15: 49125956-49125956 on Build GRCh38
rs138494934 Chr.15: 49125549-49125549 on Build GRCh38
rs559375629 Chr.15: 49125785-49125785 on Build GRCh38
rs954685986 Chr.15: 49125837-49125837 on Build GRCh38
rs777894606 Chr.15: 49125678-49125678 on Build GRCh38
rs891505575 Chr.15: 49125684-49125684 on Build GRCh38
rs529928384 Chr.15: 49125786-49125786 on Build GRCh38
rs906966045 Chr.15: 49125588-49125588 on Build GRCh38
rs773300966 Chr.15: 49125828-49125828 on Build GRCh38
rs541888064 Chr.15: 49125847-49125847 on Build GRCh38
rs899851702 Chr.15: 49125891-49125891 on Build GRCh38
rs570501867 Chr.15: 49125987-49125987 on Build GRCh38
rs938469508 Chr.15: 49125589-49125589 on Build GRCh38
rs187443287 Chr.15: 49125859-49125859 on Build GRCh38
rs1035957528 Chr.15: 49125768-49125768 on Build GRCh38
rs887246680 Chr.15: 49125993-49125993 on Build GRCh38
rs920140543 Chr.15: 49125546-49125546 on Build GRCh38
rs1021560405 Chr.15: 49125767-49125767 on Build GRCh38
rs1008258284 Chr.15: 49125579-49125579 on Build GRCh38
rs752101633 Chr.15: 49125873-49125873 on Build GRCh38
rs1051702148 Chr.15: 49125568-49125568 on Build GRCh38
rs116194550 Chr.15: 49125754-49125754 on Build GRCh38
rs944034882 Chr.15: 49125874-49125874 on Build GRCh38
rs974012394 Chr.15: 49125871-49125871 on Build GRCh38
rs1040175061 Chr.15: 49125625-49125625 on Build GRCh38
rs948975957 Chr.15: 49125972-49125972 on Build GRCh38
rs900388604 Chr.15: 49125770-49125770 on Build GRCh38
rs143037590 Chr.15: 49125558-49125558 on Build GRCh38
rs986879394 Chr.15: 49125840-49125840 on Build GRCh38
rs991192098 Chr.15: 49125774-49125774 on Build GRCh38
rs569749170 Chr.15: 49125864-49125864 on Build GRCh38

View Full Product Details