Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 60347012-60347531 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANXA2
Entrez Gene ID
302
Gene Name
annexin A2
Gene Aliases
ANX2, ANX2L4, CAL1H, HEL-S-270, LIP2, LPC2, LPC2D, P36, PAP-IV
UniGene
Hs.511605
Gene Chromosome Location
Chr.15: 60347151-60398025 on Build GRCh38
Transcripts
NM_001002857.1 NM_001002858.2 NM_001136015.2 NM_004039.2 XM_011521477.2 XM_017022090.1 XM_017022091.1 XM_017022092.1 XM_017022093.1

SNP Detail

SNP ID Chromosome Location
rs986971712 Chr.15: 60347132-60347132 on Build GRCh38
rs967501112 Chr.15: 60347203-60347203 on Build GRCh38
rs941320843 Chr.15: 60347343-60347343 on Build GRCh38
rs377090405 Chr.15: 60347184-60347184 on Build GRCh38
rs894362610 Chr.15: 60347361-60347361 on Build GRCh38
rs140887396 Chr.15: 60347391-60347391 on Build GRCh38
rs965210712 Chr.15: 60347106-60347106 on Build GRCh38
rs35828699 Chr.15: 60347057-60347057 on Build GRCh38
rs11553798 Chr.15: 60347485-60347485 on Build GRCh38
rs41315062 Chr.15: 60347401-60347401 on Build GRCh38
rs747362761 Chr.15: 60347044-60347044 on Build GRCh38
rs555423795 Chr.15: 60347314-60347314 on Build GRCh38
rs948401113 Chr.15: 60347296-60347296 on Build GRCh38
rs1024532528 Chr.15: 60347386-60347386 on Build GRCh38
rs942888435 Chr.15: 60347288-60347288 on Build GRCh38
rs546079795 Chr.15: 60347430-60347430 on Build GRCh38
rs566236506 Chr.15: 60347191-60347191 on Build GRCh38
rs928390694 Chr.15: 60347305-60347305 on Build GRCh38
rs11553797 Chr.15: 60347411-60347411 on Build GRCh38
rs867195434 Chr.15: 60347359-60347359 on Build GRCh38
rs184828726 Chr.15: 60347463-60347463 on Build GRCh38
rs1008351300 Chr.15: 60347466-60347466 on Build GRCh38
rs151000652 Chr.15: 60347284-60347284 on Build GRCh38
rs990829513 Chr.15: 60347259-60347259 on Build GRCh38
rs561764153 Chr.15: 60347490-60347490 on Build GRCh38
rs866242568 Chr.15: 60347452-60347452 on Build GRCh38
rs979795701 Chr.15: 60347301-60347301 on Build GRCh38
rs955502019 Chr.15: 60347104-60347104 on Build GRCh38
rs879159858 Chr.15: 60347335-60347335 on Build GRCh38
rs996928278 Chr.15: 60347075-60347075 on Build GRCh38
rs576093157 Chr.15: 60347439-60347439 on Build GRCh38
rs1018547934 Chr.15: 60347504-60347504 on Build GRCh38
rs150128007 Chr.15: 60347404-60347404 on Build GRCh38
rs1047545289 Chr.15: 60347326-60347326 on Build GRCh38
rs1014099597 Chr.15: 60347379-60347379 on Build GRCh38
rs143592697 Chr.15: 60347195-60347195 on Build GRCh38
rs925589277 Chr.15: 60347221-60347221 on Build GRCh38
rs138873688 Chr.15: 60347436-60347436 on Build GRCh38
rs879886894 Chr.15: 60347289-60347289 on Build GRCh38
rs989725509 Chr.15: 60347253-60347253 on Build GRCh38
rs770639800 Chr.15: 60347055-60347055 on Build GRCh38
rs115565448 Chr.15: 60347076-60347076 on Build GRCh38
rs116928563 Chr.15: 60347249-60347249 on Build GRCh38
rs535453430 Chr.15: 60347479-60347479 on Build GRCh38
rs888382926 Chr.15: 60347124-60347124 on Build GRCh38
rs930028362 Chr.15: 60347329-60347329 on Build GRCh38
rs769141986 Chr.15: 60347265-60347265 on Build GRCh38
rs867865719 Chr.15: 60347038-60347038 on Build GRCh38
rs938483800 Chr.15: 60347306-60347306 on Build GRCh38
rs958528720 Chr.15: 60347152-60347152 on Build GRCh38
rs569738535 Chr.15: 60347251-60347251 on Build GRCh38
rs888435252 Chr.15: 60347339-60347339 on Build GRCh38
rs11553799 Chr.15: 60347454-60347454 on Build GRCh38
rs566396858 Chr.15: 60347414-60347414 on Build GRCh38
rs1047477497 Chr.15: 60347088-60347088 on Build GRCh38
rs1016933008 Chr.15: 60347045-60347045 on Build GRCh38
rs572205706 Chr.15: 60347413-60347413 on Build GRCh38
rs977130777 Chr.15: 60347215-60347215 on Build GRCh38
rs868457426 Chr.15: 60347328-60347328 on Build GRCh38
rs555028596 Chr.15: 60347489-60347489 on Build GRCh38
rs1034178781 Chr.15: 60347150-60347150 on Build GRCh38
rs533037904 Chr.15: 60347153-60347157 on Build GRCh38
rs3582 Chr.15: 60347469-60347469 on Build GRCh38

View Full Product Details