Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 63307033-63307542 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APH1B
Entrez Gene ID
83464
Gene Name
aph-1 homolog B, gamma-secretase subunit
Gene Aliases
APH-1B, PRO1328, PSFL, TAAV688
UniGene
Hs.511703
Gene Chromosome Location
Chr.15: 63277519-63309126 on Build GRCh38
Transcripts
NM_001145646.1 NM_031301.3

SNP Detail

SNP ID Chromosome Location
rs568127938 Chr.15: 63307467-63307467 on Build GRCh38
rs1016735801 Chr.15: 63307407-63307407 on Build GRCh38
rs138792718 Chr.15: 63307363-63307373 on Build GRCh38
rs117880351 Chr.15: 63307193-63307193 on Build GRCh38
rs755721488 Chr.15: 63307341-63307341 on Build GRCh38
rs545708655 Chr.15: 63307095-63307095 on Build GRCh38
rs142392559 Chr.15: 63307323-63307323 on Build GRCh38
rs942799629 Chr.15: 63307202-63307202 on Build GRCh38
rs79106040 Chr.15: 63307476-63307476 on Build GRCh38
rs911229410 Chr.15: 63307457-63307457 on Build GRCh38
rs548146789 Chr.15: 63307266-63307266 on Build GRCh38
rs116314964 Chr.15: 63307436-63307436 on Build GRCh38
rs369215207 Chr.15: 63307374-63307374 on Build GRCh38
rs560439293 Chr.15: 63307308-63307308 on Build GRCh38
rs909729750 Chr.15: 63307092-63307092 on Build GRCh38
rs558901603 Chr.15: 63307321-63307321 on Build GRCh38
rs918590206 Chr.15: 63307128-63307128 on Build GRCh38
rs866475523 Chr.15: 63307299-63307299 on Build GRCh38
rs1019392346 Chr.15: 63307363-63307382 on Build GRCh38
rs776601515 Chr.15: 63307487-63307487 on Build GRCh38
rs12899650 Chr.15: 63307372-63307372 on Build GRCh38
rs562601436 Chr.15: 63307189-63307189 on Build GRCh38
rs1051603271 Chr.15: 63307172-63307172 on Build GRCh38
rs986797556 Chr.15: 63307455-63307455 on Build GRCh38
rs1003231916 Chr.15: 63307358-63307358 on Build GRCh38
rs914733025 Chr.15: 63307488-63307488 on Build GRCh38
rs552538587 Chr.15: 63307324-63307324 on Build GRCh38
rs934042348 Chr.15: 63307155-63307155 on Build GRCh38
rs902162176 Chr.15: 63307340-63307340 on Build GRCh38
rs771433839 Chr.15: 63307223-63307225 on Build GRCh38
rs549859192 Chr.15: 63307437-63307437 on Build GRCh38
rs962577275 Chr.15: 63307414-63307414 on Build GRCh38
rs990629443 Chr.15: 63307084-63307084 on Build GRCh38
rs946246039 Chr.15: 63307493-63307493 on Build GRCh38
rs942755041 Chr.15: 63307466-63307466 on Build GRCh38
rs749024348 Chr.15: 63307390-63307394 on Build GRCh38
rs564295808 Chr.15: 63307107-63307107 on Build GRCh38
rs1004107357 Chr.15: 63307362-63307372 on Build GRCh38
rs545417753 Chr.15: 63307313-63307313 on Build GRCh38
rs571379195 Chr.15: 63307470-63307470 on Build GRCh38
rs140956620 Chr.15: 63307121-63307121 on Build GRCh38
rs747156036 Chr.15: 63307404-63307404 on Build GRCh38
rs571023917 Chr.15: 63307405-63307405 on Build GRCh38
rs544695386 Chr.15: 63307122-63307122 on Build GRCh38
rs1041930924 Chr.15: 63307515-63307515 on Build GRCh38
rs990575565 Chr.15: 63307389-63307389 on Build GRCh38
rs76119745 Chr.15: 63307215-63307215 on Build GRCh38

View Full Product Details