Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 63307398-63307902 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APH1B
Entrez Gene ID
83464
Gene Name
aph-1 homolog B, gamma-secretase subunit
Gene Aliases
APH-1B, PRO1328, PSFL, TAAV688
UniGene
Hs.511703
Gene Chromosome Location
Chr.15: 63277519-63309126 on Build GRCh38
Transcripts
NM_001145646.1 NM_031301.3

SNP Detail

SNP ID Chromosome Location
rs568127938 Chr.15: 63307467-63307467 on Build GRCh38
rs928803237 Chr.15: 63307851-63307854 on Build GRCh38
rs373121586 Chr.15: 63307544-63307544 on Build GRCh38
rs568607378 Chr.15: 63307552-63307552 on Build GRCh38
rs946151230 Chr.15: 63307831-63307831 on Build GRCh38
rs79106040 Chr.15: 63307476-63307476 on Build GRCh38
rs911229410 Chr.15: 63307457-63307457 on Build GRCh38
rs914680810 Chr.15: 63307825-63307825 on Build GRCh38
rs116314964 Chr.15: 63307436-63307436 on Build GRCh38
rs762602066 Chr.15: 63307778-63307778 on Build GRCh38
rs4984240 Chr.15: 63307738-63307738 on Build GRCh38
rs180926473 Chr.15: 63307792-63307792 on Build GRCh38
rs765048455 Chr.15: 63307676-63307676 on Build GRCh38
rs759542921 Chr.15: 63307638-63307638 on Build GRCh38
rs543215084 Chr.15: 63307740-63307740 on Build GRCh38
rs539716475 Chr.15: 63307624-63307624 on Build GRCh38
rs1041860732 Chr.15: 63307850-63307850 on Build GRCh38
rs1056489091 Chr.15: 63307877-63307877 on Build GRCh38
rs776601515 Chr.15: 63307487-63307487 on Build GRCh38
rs987132234 Chr.15: 63307748-63307748 on Build GRCh38
rs1056099096 Chr.15: 63307575-63307575 on Build GRCh38
rs986797556 Chr.15: 63307455-63307455 on Build GRCh38
rs139511778 Chr.15: 63307753-63307753 on Build GRCh38
rs914733025 Chr.15: 63307488-63307488 on Build GRCh38
rs1012000167 Chr.15: 63307630-63307630 on Build GRCh38
rs549859192 Chr.15: 63307437-63307437 on Build GRCh38
rs572404890 Chr.15: 63307591-63307591 on Build GRCh38
rs376522140 Chr.15: 63307632-63307632 on Build GRCh38
rs946246039 Chr.15: 63307493-63307493 on Build GRCh38
rs938906874 Chr.15: 63307874-63307874 on Build GRCh38
rs942755041 Chr.15: 63307466-63307466 on Build GRCh38
rs1003549727 Chr.15: 63307563-63307563 on Build GRCh38
rs994025501 Chr.15: 63307685-63307685 on Build GRCh38
rs894805622 Chr.15: 63307625-63307625 on Build GRCh38
rs571379195 Chr.15: 63307470-63307470 on Build GRCh38
rs907425983 Chr.15: 63307536-63307536 on Build GRCh38
rs377712202 Chr.15: 63307543-63307543 on Build GRCh38
rs962524795 Chr.15: 63307649-63307649 on Build GRCh38
rs58984049 Chr.15: 63307727-63307727 on Build GRCh38
rs771579805 Chr.15: 63307738-63307739 on Build GRCh38
rs1041930924 Chr.15: 63307515-63307515 on Build GRCh38
rs911187382 Chr.15: 63307763-63307763 on Build GRCh38
rs1025917436 Chr.15: 63307686-63307686 on Build GRCh38
rs4984263 Chr.15: 63307789-63307789 on Build GRCh38

View Full Product Details