Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 89628404-89628893 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF7
Entrez Gene ID
374654
Gene Name
kinesin family member 7
Gene Aliases
ACLS, AGBK, HLS2, JBTS12, UNQ340
UniGene
Hs.513134
Gene Chromosome Location
Chr.15: 89627970-89663086 on Build GRCh38
Transcripts
NM_198525.2 XM_011521531.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4057811 c.2346G>A p.Q782Q Chr.15: 89628566-89628566 on Build GRCh38
4696218 c.4016G>A p.R1339Q Chr.15: 89628435-89628435 on Build GRCh38
4696222 c.3760_3761insC p.L1254fs*3 Chr.15: 89628690-89628691 on Build GRCh38
272868 c.2347C>T p.R783W Chr.15: 89628565-89628565 on Build GRCh38
5148593 c.3939C>T p.G1313G Chr.15: 89628512-89628512 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779149527 Chr.15: 89628550-89628550 on Build GRCh38
rs778654148 Chr.15: 89628432-89628432 on Build GRCh38
rs149585159 Chr.15: 89628623-89628623 on Build GRCh38
rs775576101 Chr.15: 89628679-89628680 on Build GRCh38
rs141028210 Chr.15: 89628491-89628491 on Build GRCh38
rs751068207 Chr.15: 89628423-89628423 on Build GRCh38
rs371645372 Chr.15: 89628693-89628693 on Build GRCh38
rs749221779 Chr.15: 89628616-89628616 on Build GRCh38
rs749235273 Chr.15: 89628662-89628662 on Build GRCh38
rs370123540 Chr.15: 89628463-89628463 on Build GRCh38
rs923542256 Chr.15: 89628512-89628512 on Build GRCh38
rs769211531 Chr.15: 89628795-89628795 on Build GRCh38
rs781336592 Chr.15: 89628702-89628702 on Build GRCh38
rs750907041 Chr.15: 89628540-89628540 on Build GRCh38
rs761421026 Chr.15: 89628581-89628581 on Build GRCh38
rs375983330 Chr.15: 89628489-89628489 on Build GRCh38
rs768596111 Chr.15: 89628732-89628734 on Build GRCh38
rs767091542 Chr.15: 89628631-89628631 on Build GRCh38
rs145726393 Chr.15: 89628453-89628453 on Build GRCh38
rs367833833 Chr.15: 89628678-89628678 on Build GRCh38
rs768543012 Chr.15: 89628663-89628663 on Build GRCh38
rs905968852 Chr.15: 89628700-89628700 on Build GRCh38
rs770804978 Chr.15: 89628678-89628679 on Build GRCh38
rs747829509 Chr.15: 89628435-89628435 on Build GRCh38
rs1044478234 Chr.15: 89628577-89628577 on Build GRCh38
rs759086470 Chr.15: 89628690-89628690 on Build GRCh38
rs770165381 Chr.15: 89628455-89628455 on Build GRCh38
rs757991046 Chr.15: 89628426-89628426 on Build GRCh38
rs778763577 Chr.15: 89628708-89628708 on Build GRCh38
rs143516080 Chr.15: 89628484-89628484 on Build GRCh38
rs776965394 Chr.15: 89628771-89628773 on Build GRCh38
rs769455694 Chr.15: 89628677-89628677 on Build GRCh38
rs747733266 Chr.15: 89628473-89628475 on Build GRCh38
rs372979006 Chr.15: 89628475-89628475 on Build GRCh38
rs79532879 Chr.15: 89628791-89628791 on Build GRCh38
rs147772494 Chr.15: 89628466-89628466 on Build GRCh38
rs544791294 Chr.15: 89628600-89628600 on Build GRCh38
rs761932434 Chr.15: 89628795-89628796 on Build GRCh38
rs1026093138 Chr.15: 89628767-89628767 on Build GRCh38
rs138993311 Chr.15: 89628537-89628537 on Build GRCh38
rs761522868 Chr.15: 89628629-89628629 on Build GRCh38
rs757946358 Chr.15: 89628436-89628436 on Build GRCh38
rs756750505 Chr.15: 89628538-89628538 on Build GRCh38
rs149883801 Chr.15: 89628712-89628712 on Build GRCh38
rs755333464 Chr.15: 89628598-89628598 on Build GRCh38
rs749499441 Chr.15: 89628496-89628496 on Build GRCh38
rs759591288 Chr.15: 89628513-89628513 on Build GRCh38
rs73477443 Chr.15: 89628477-89628477 on Build GRCh38
rs148749994 Chr.15: 89628656-89628656 on Build GRCh38
rs964521354 Chr.15: 89628864-89628864 on Build GRCh38
rs751191936 Chr.15: 89628580-89628580 on Build GRCh38
rs747131428 Chr.15: 89628690-89628691 on Build GRCh38
rs374693639 Chr.15: 89628715-89628715 on Build GRCh38
rs759021362 Chr.15: 89628719-89628719 on Build GRCh38
rs546772749 Chr.15: 89628696-89628696 on Build GRCh38
rs778142943 Chr.15: 89628606-89628606 on Build GRCh38
rs773655787 Chr.15: 89628828-89628828 on Build GRCh38
rs202214398 Chr.15: 89628465-89628465 on Build GRCh38
rs749230267 Chr.15: 89628673-89628690 on Build GRCh38
rs754774163 Chr.15: 89628473-89628473 on Build GRCh38
rs147767277 Chr.15: 89628692-89628692 on Build GRCh38
rs745496315 Chr.15: 89628793-89628793 on Build GRCh38
rs775649452 Chr.15: 89628460-89628460 on Build GRCh38
rs530450447 Chr.15: 89628611-89628611 on Build GRCh38
rs774442900 Chr.15: 89628501-89628501 on Build GRCh38
rs777426943 Chr.15: 89628672-89628673 on Build GRCh38
rs140562404 Chr.15: 89628673-89628673 on Build GRCh38
rs148134313 Chr.15: 89628485-89628485 on Build GRCh38
rs769319252 Chr.15: 89628722-89628722 on Build GRCh38
rs752651717 Chr.15: 89628764-89628764 on Build GRCh38
rs774071217 Chr.15: 89628624-89628624 on Build GRCh38
rs137993810 Chr.15: 89628452-89628452 on Build GRCh38
rs371960047 Chr.15: 89628738-89628738 on Build GRCh38
rs561205514 Chr.15: 89628666-89628666 on Build GRCh38
rs754341930 Chr.15: 89628640-89628640 on Build GRCh38
rs746868507 Chr.15: 89628779-89628779 on Build GRCh38
rs376563309 Chr.15: 89628761-89628761 on Build GRCh38
rs775882284 Chr.15: 89628492-89628492 on Build GRCh38
rs78862745 Chr.15: 89628804-89628804 on Build GRCh38
rs199526156 Chr.15: 89628429-89628429 on Build GRCh38
rs765893045 Chr.15: 89628675-89628675 on Build GRCh38
rs577637542 Chr.15: 89628563-89628563 on Build GRCh38
rs558106454 Chr.15: 89628458-89628458 on Build GRCh38
rs202197486 Chr.15: 89628641-89628641 on Build GRCh38
rs750635928 Chr.15: 89628753-89628753 on Build GRCh38
rs150248985 Chr.15: 89628507-89628507 on Build GRCh38
rs780942335 Chr.15: 89628454-89628465 on Build GRCh38
rs763741856 Chr.15: 89628536-89628536 on Build GRCh38
rs773479675 Chr.15: 89628560-89628560 on Build GRCh38
rs760900374 Chr.15: 89628838-89628838 on Build GRCh38
rs201519024 Chr.15: 89628709-89628709 on Build GRCh38
rs775084299 Chr.15: 89628803-89628803 on Build GRCh38
rs776674506 Chr.15: 89628509-89628509 on Build GRCh38
rs549257070 Chr.15: 89628825-89628825 on Build GRCh38
rs763506306 Chr.15: 89628737-89628737 on Build GRCh38
rs749589340 Chr.15: 89628548-89628548 on Build GRCh38
rs143877028 Chr.15: 89628609-89628609 on Build GRCh38
rs144301755 Chr.15: 89628654-89628654 on Build GRCh38
rs769783876 Chr.15: 89628565-89628565 on Build GRCh38
rs774986061 Chr.15: 89628824-89628824 on Build GRCh38
rs760277701 Chr.15: 89628674-89628674 on Build GRCh38
rs768917178 Chr.15: 89628549-89628549 on Build GRCh38
rs765636110 Chr.15: 89628816-89628817 on Build GRCh38
rs771532005 Chr.15: 89628447-89628447 on Build GRCh38
rs747263865 Chr.15: 89628655-89628655 on Build GRCh38
rs765625703 Chr.15: 89628644-89628644 on Build GRCh38
rs868862477 Chr.15: 89628556-89628556 on Build GRCh38
rs144929293 Chr.15: 89628713-89628713 on Build GRCh38
rs756204120 Chr.15: 89628707-89628707 on Build GRCh38
rs777140020 Chr.15: 89628478-89628478 on Build GRCh38
rs528407611 Chr.15: 89628691-89628691 on Build GRCh38
rs371596537 Chr.15: 89628650-89628650 on Build GRCh38
rs763825284 Chr.15: 89628578-89628578 on Build GRCh38
rs775526351 Chr.15: 89628612-89628612 on Build GRCh38
rs750639239 Chr.15: 89628833-89628833 on Build GRCh38
rs767690124 Chr.15: 89628747-89628747 on Build GRCh38
rs770740912 Chr.15: 89628781-89628781 on Build GRCh38
rs780483011 Chr.15: 89628587-89628587 on Build GRCh38
rs200891085 Chr.15: 89628487-89628487 on Build GRCh38
rs886051529 Chr.15: 89628494-89628494 on Build GRCh38
rs750002097 Chr.15: 89628585-89628585 on Build GRCh38
rs772244933 Chr.15: 89628605-89628605 on Build GRCh38
rs142598777 Chr.15: 89628667-89628667 on Build GRCh38
rs765907353 Chr.15: 89628636-89628636 on Build GRCh38
rs866142810 Chr.15: 89628619-89628619 on Build GRCh38
rs142246932 Chr.15: 89628545-89628545 on Build GRCh38
rs777387974 Chr.15: 89628441-89628441 on Build GRCh38
rs754326027 Chr.15: 89628597-89628597 on Build GRCh38
rs754712742 Chr.15: 89628438-89628439 on Build GRCh38
rs370333561 Chr.15: 89628557-89628557 on Build GRCh38
rs149448369 Chr.15: 89628508-89628508 on Build GRCh38
rs769354808 Chr.15: 89628808-89628808 on Build GRCh38
rs551619127 Chr.15: 89628774-89628774 on Build GRCh38
rs767890596 Chr.15: 89628830-89628830 on Build GRCh38
rs772866453 Chr.15: 89628635-89628635 on Build GRCh38
rs775942651 Chr.15: 89628688-89628688 on Build GRCh38
rs780139363 Chr.15: 89628762-89628762 on Build GRCh38
rs146300227 Chr.15: 89628558-89628558 on Build GRCh38
rs976233649 Chr.15: 89628665-89628665 on Build GRCh38
rs775579416 Chr.15: 89628574-89628574 on Build GRCh38
rs746002125 Chr.15: 89628706-89628706 on Build GRCh38
rs747632760 Chr.15: 89628657-89628657 on Build GRCh38
rs201363681 Chr.15: 89628564-89628564 on Build GRCh38
rs752933212 Chr.15: 89628645-89628645 on Build GRCh38
rs1017659112 Chr.15: 89628855-89628855 on Build GRCh38
rs771882581 Chr.15: 89628710-89628710 on Build GRCh38
rs377066237 Chr.15: 89628462-89628462 on Build GRCh38
rs537735484 Chr.15: 89628483-89628483 on Build GRCh38
rs775895621 Chr.15: 89628728-89628728 on Build GRCh38
rs147195322 Chr.15: 89628669-89628669 on Build GRCh38
rs769689199 Chr.15: 89628668-89628685 on Build GRCh38
rs963145880 Chr.15: 89628638-89628638 on Build GRCh38
rs762263951 Chr.15: 89628746-89628746 on Build GRCh38
rs768990207 Chr.15: 89628498-89628498 on Build GRCh38
rs1039815039 Chr.15: 89628554-89628554 on Build GRCh38
rs149061204 Chr.15: 89628773-89628773 on Build GRCh38
rs141225908 Chr.15: 89628474-89628474 on Build GRCh38
rs756672880 Chr.15: 89628544-89628544 on Build GRCh38
rs762657379 Chr.15: 89628524-89628524 on Build GRCh38
rs752603278 Chr.15: 89628519-89628519 on Build GRCh38
rs747455856 Chr.15: 89628444-89628444 on Build GRCh38
rs140121479 Chr.15: 89628698-89628698 on Build GRCh38

View Full Product Details