Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 89630124-89630623 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF7
Entrez Gene ID
374654
Gene Name
kinesin family member 7
Gene Aliases
ACLS, AGBK, HLS2, JBTS12, UNQ340
UniGene
Hs.513134
Gene Chromosome Location
Chr.15: 89627970-89663086 on Build GRCh38
Transcripts
NM_198525.2 XM_011521531.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6695707 c.3250C>A p.L1084I Chr.15: 89630355-89630355 on Build GRCh38
4166381 c.3218C>T p.S1073L Chr.15: 89630387-89630387 on Build GRCh38
1608811 c.3150G>A p.E1050E Chr.15: 89630455-89630455 on Build GRCh38
6143161 c.3208C>T p.L1070F Chr.15: 89630397-89630397 on Build GRCh38
4166382 c.1679C>T p.S560L Chr.15: 89630387-89630387 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770485780 Chr.15: 89630302-89630302 on Build GRCh38
rs541580399 Chr.15: 89630422-89630422 on Build GRCh38
rs762077405 Chr.15: 89630401-89630401 on Build GRCh38
rs77474187 Chr.15: 89630409-89630409 on Build GRCh38
rs530364548 Chr.15: 89630290-89630290 on Build GRCh38
rs925622958 Chr.15: 89630213-89630213 on Build GRCh38
rs77896636 Chr.15: 89630353-89630353 on Build GRCh38
rs200260697 Chr.15: 89630248-89630248 on Build GRCh38
rs372323903 Chr.15: 89630271-89630271 on Build GRCh38
rs370525086 Chr.15: 89630602-89630602 on Build GRCh38
rs748595084 Chr.15: 89630486-89630486 on Build GRCh38
rs140549999 Chr.15: 89630488-89630488 on Build GRCh38
rs1015151833 Chr.15: 89630299-89630299 on Build GRCh38
rs754842380 Chr.15: 89630261-89630261 on Build GRCh38
rs143669426 Chr.15: 89630200-89630200 on Build GRCh38
rs375802364 Chr.15: 89630380-89630380 on Build GRCh38
rs370392455 Chr.15: 89630247-89630247 on Build GRCh38
rs367627150 Chr.15: 89630321-89630321 on Build GRCh38
rs976170562 Chr.15: 89630163-89630163 on Build GRCh38
rs368047247 Chr.15: 89630541-89630541 on Build GRCh38
rs146808726 Chr.15: 89630482-89630482 on Build GRCh38
rs565305401 Chr.15: 89630233-89630233 on Build GRCh38
rs971160449 Chr.15: 89630582-89630582 on Build GRCh38
rs770366525 Chr.15: 89630284-89630284 on Build GRCh38
rs372151159 Chr.15: 89630512-89630512 on Build GRCh38
rs747994640 Chr.15: 89630267-89630267 on Build GRCh38
rs765207161 Chr.15: 89630343-89630343 on Build GRCh38
rs555924068 Chr.15: 89630171-89630171 on Build GRCh38
rs758215908 Chr.15: 89630348-89630348 on Build GRCh38
rs147191956 Chr.15: 89630403-89630403 on Build GRCh38
rs780698440 Chr.15: 89630372-89630372 on Build GRCh38
rs1036922746 Chr.15: 89630559-89630559 on Build GRCh38
rs1000402274 Chr.15: 89630560-89630560 on Build GRCh38
rs752092734 Chr.15: 89630322-89630323 on Build GRCh38
rs886039282 Chr.15: 89630426-89630426 on Build GRCh38
rs566534275 Chr.15: 89630344-89630344 on Build GRCh38
rs775530686 Chr.15: 89630225-89630225 on Build GRCh38
rs149378390 Chr.15: 89630381-89630381 on Build GRCh38
rs72522688 Chr.15: 89630502-89630503 on Build GRCh38
rs991445665 Chr.15: 89630434-89630434 on Build GRCh38
rs779273024 Chr.15: 89630483-89630483 on Build GRCh38
rs574319954 Chr.15: 89630184-89630184 on Build GRCh38
rs1029711579 Chr.15: 89630340-89630340 on Build GRCh38
rs983384787 Chr.15: 89630457-89630457 on Build GRCh38
rs1032931470 Chr.15: 89630561-89630561 on Build GRCh38
rs542706994 Chr.15: 89630191-89630191 on Build GRCh38
rs368905885 Chr.15: 89630518-89630518 on Build GRCh38
rs751775036 Chr.15: 89630245-89630245 on Build GRCh38
rs776622132 Chr.15: 89630510-89630510 on Build GRCh38
rs768917109 Chr.15: 89630393-89630393 on Build GRCh38
rs570460369 Chr.15: 89630394-89630394 on Build GRCh38
rs932658769 Chr.15: 89630511-89630511 on Build GRCh38
rs772293531 Chr.15: 89630487-89630487 on Build GRCh38
rs903008085 Chr.15: 89630543-89630543 on Build GRCh38
rs1038802503 Chr.15: 89630275-89630275 on Build GRCh38
rs888752945 Chr.15: 89630583-89630583 on Build GRCh38
rs138764398 Chr.15: 89630341-89630341 on Build GRCh38
rs763451105 Chr.15: 89630311-89630311 on Build GRCh38
rs202129004 Chr.15: 89630385-89630385 on Build GRCh38
rs977333722 Chr.15: 89630216-89630216 on Build GRCh38
rs767497946 Chr.15: 89630333-89630333 on Build GRCh38
rs564914280 Chr.15: 89630575-89630575 on Build GRCh38
rs757004181 Chr.15: 89630365-89630365 on Build GRCh38
rs913157453 Chr.15: 89630199-89630199 on Build GRCh38
rs746652178 Chr.15: 89630357-89630357 on Build GRCh38
rs540213363 Chr.15: 89630229-89630229 on Build GRCh38
rs750580913 Chr.15: 89630335-89630335 on Build GRCh38
rs916572967 Chr.15: 89630472-89630472 on Build GRCh38
rs964505763 Chr.15: 89630161-89630161 on Build GRCh38
rs545150173 Chr.15: 89630170-89630170 on Build GRCh38
rs940838929 Chr.15: 89630526-89630526 on Build GRCh38
rs767837351 Chr.15: 89630402-89630402 on Build GRCh38
rs78984448 Chr.15: 89630238-89630238 on Build GRCh38
rs767060184 Chr.15: 89630276-89630276 on Build GRCh38
rs996871530 Chr.15: 89630164-89630164 on Build GRCh38
rs774461779 Chr.15: 89630320-89630320 on Build GRCh38
rs764513802 Chr.15: 89630312-89630312 on Build GRCh38
rs759958018 Chr.15: 89630261-89630262 on Build GRCh38
rs565121041 Chr.15: 89630556-89630556 on Build GRCh38
rs1020101136 Chr.15: 89630350-89630350 on Build GRCh38
rs574281511 Chr.15: 89630413-89630413 on Build GRCh38
rs982551157 Chr.15: 89630585-89630585 on Build GRCh38
rs899831488 Chr.15: 89630296-89630296 on Build GRCh38
rs771009677 Chr.15: 89630509-89630509 on Build GRCh38
rs376433874 Chr.15: 89630501-89630501 on Build GRCh38
rs201335315 Chr.15: 89630242-89630242 on Build GRCh38
rs758270931 Chr.15: 89630407-89630407 on Build GRCh38
rs750772799 Chr.15: 89630479-89630479 on Build GRCh38
rs958588270 Chr.15: 89630562-89630562 on Build GRCh38
rs753466002 Chr.15: 89630517-89630518 on Build GRCh38
rs745367865 Chr.15: 89630300-89630300 on Build GRCh38
rs201342316 Chr.15: 89630404-89630404 on Build GRCh38
rs528572567 Chr.15: 89630208-89630208 on Build GRCh38
rs372699643 Chr.15: 89630304-89630304 on Build GRCh38
rs914407609 Chr.15: 89630201-89630201 on Build GRCh38
rs991166275 Chr.15: 89630574-89630574 on Build GRCh38
rs756984615 Chr.15: 89630411-89630411 on Build GRCh38
rs778818088 Chr.15: 89630265-89630265 on Build GRCh38
rs761702891 Chr.15: 89630530-89630530 on Build GRCh38
rs1011252791 Chr.15: 89630598-89630598 on Build GRCh38
rs35820949 Chr.15: 89630503-89630504 on Build GRCh38
rs948136740 Chr.15: 89630263-89630263 on Build GRCh38
rs764460692 Chr.15: 89630243-89630243 on Build GRCh38
rs370548253 Chr.15: 89630503-89630503 on Build GRCh38
rs763937653 Chr.15: 89630408-89630408 on Build GRCh38
rs779655277 Chr.15: 89630432-89630432 on Build GRCh38
rs373864613 Chr.15: 89630504-89630504 on Build GRCh38
rs746630755 Chr.15: 89630273-89630273 on Build GRCh38
rs66606640 Chr.15: 89630504-89630505 on Build GRCh38
rs573225695 Chr.15: 89630506-89630506 on Build GRCh38
rs749819007 Chr.15: 89630451-89630451 on Build GRCh38
rs544509331 Chr.15: 89630168-89630168 on Build GRCh38
rs745423935 Chr.15: 89630379-89630379 on Build GRCh38
rs751425504 Chr.15: 89630517-89630517 on Build GRCh38
rs200625779 Chr.15: 89630501-89630502 on Build GRCh38
rs112107761 Chr.15: 89630256-89630256 on Build GRCh38
rs762038154 Chr.15: 89630327-89630327 on Build GRCh38
rs577033655 Chr.15: 89630149-89630149 on Build GRCh38
rs374801877 Chr.15: 89630514-89630514 on Build GRCh38
rs757314765 Chr.15: 89630246-89630246 on Build GRCh38
rs1020573842 Chr.15: 89630524-89630524 on Build GRCh38

View Full Product Details