Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 2597021-2597518 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDPK1
Entrez Gene ID
5170
Gene Name
3-phosphoinositide dependent protein kinase 1
Gene Aliases
PDK1, PDPK2, PDPK2P, PRO0461
UniGene
Hs.459691
Gene Chromosome Location
Chr.16: 2537964-2603190 on Build GRCh38
Transcripts
NM_001261816.1 NM_002613.4 NM_031268.5 XM_011522521.2 XM_011522523.2 XM_017023271.1 XM_017023272.1 XM_017023273.1 XM_017023274.1 XM_017023275.1 XM_017023276.1 XM_017023277.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
332681 c.1554+1G>T p.? Chr.16: 2597276-2597276 on Build GRCh38
6961974 c.1500G>A p.W500* Chr.16: 2597221-2597221 on Build GRCh38
702846 c.1477G>C p.V493L Chr.16: 2597198-2597198 on Build GRCh38
6850435 c.1496C>T p.P499L Chr.16: 2597217-2597217 on Build GRCh38
6946654 c.1442G>T p.G481V Chr.16: 2597163-2597163 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs896701150 Chr.16: 2597419-2597419 on Build GRCh38
rs147946576 Chr.16: 2597412-2597412 on Build GRCh38
rs866640912 Chr.16: 2597224-2597224 on Build GRCh38
rs576677552 Chr.16: 2597441-2597441 on Build GRCh38
rs900601956 Chr.16: 2597384-2597384 on Build GRCh38
rs761277571 Chr.16: 2597210-2597210 on Build GRCh38
rs189326146 Chr.16: 2597077-2597077 on Build GRCh38
rs780180372 Chr.16: 2597332-2597332 on Build GRCh38
rs563055958 Chr.16: 2597414-2597414 on Build GRCh38
rs778508775 Chr.16: 2597089-2597089 on Build GRCh38
rs765324347 Chr.16: 2597099-2597101 on Build GRCh38
rs144926872 Chr.16: 2597369-2597369 on Build GRCh38
rs1001507233 Chr.16: 2597201-2597201 on Build GRCh38
rs535332245 Chr.16: 2597134-2597134 on Build GRCh38
rs1013748027 Chr.16: 2597101-2597101 on Build GRCh38
rs757954456 Chr.16: 2597482-2597483 on Build GRCh38
rs969132480 Chr.16: 2597260-2597260 on Build GRCh38
rs1007676520 Chr.16: 2597487-2597487 on Build GRCh38
rs781060849 Chr.16: 2597290-2597290 on Build GRCh38
rs1002309639 Chr.16: 2597065-2597065 on Build GRCh38
rs929908862 Chr.16: 2597331-2597331 on Build GRCh38
rs766091042 Chr.16: 2597262-2597262 on Build GRCh38
rs758036081 Chr.16: 2597243-2597243 on Build GRCh38
rs764588339 Chr.16: 2597226-2597226 on Build GRCh38
rs878885480 Chr.16: 2597339-2597339 on Build GRCh38
rs746864089 Chr.16: 2597302-2597302 on Build GRCh38
rs553487099 Chr.16: 2597399-2597399 on Build GRCh38
rs567746314 Chr.16: 2597118-2597118 on Build GRCh38
rs760019244 Chr.16: 2597179-2597179 on Build GRCh38
rs774842989 Chr.16: 2597176-2597176 on Build GRCh38
rs887753551 Chr.16: 2597046-2597046 on Build GRCh38
rs549158095 Chr.16: 2597274-2597274 on Build GRCh38
rs113929832 Chr.16: 2597427-2597427 on Build GRCh38
rs1013268347 Chr.16: 2597474-2597474 on Build GRCh38
rs763017868 Chr.16: 2597158-2597158 on Build GRCh38
rs966963577 Chr.16: 2597196-2597196 on Build GRCh38
rs758054132 Chr.16: 2597097-2597097 on Build GRCh38
rs527719535 Chr.16: 2597483-2597483 on Build GRCh38
rs932076458 Chr.16: 2597394-2597394 on Build GRCh38
rs756079975 Chr.16: 2597321-2597321 on Build GRCh38
rs746868398 Chr.16: 2597106-2597106 on Build GRCh38
rs148940066 Chr.16: 2597476-2597476 on Build GRCh38
rs982704749 Chr.16: 2597336-2597336 on Build GRCh38
rs768116696 Chr.16: 2597192-2597192 on Build GRCh38
rs377482205 Chr.16: 2597411-2597411 on Build GRCh38
rs749966481 Chr.16: 2597230-2597230 on Build GRCh38
rs777654625 Chr.16: 2597327-2597327 on Build GRCh38
rs775814028 Chr.16: 2597193-2597193 on Build GRCh38
rs965782136 Chr.16: 2597289-2597289 on Build GRCh38
rs974348443 Chr.16: 2597264-2597264 on Build GRCh38
rs960374377 Chr.16: 2597117-2597117 on Build GRCh38
rs771291308 Chr.16: 2597173-2597173 on Build GRCh38
rs747992986 Chr.16: 2597315-2597315 on Build GRCh38
rs557965294 Chr.16: 2597354-2597354 on Build GRCh38
rs777394904 Chr.16: 2597113-2597113 on Build GRCh38
rs565556209 Chr.16: 2597477-2597477 on Build GRCh38
rs577192003 Chr.16: 2597373-2597373 on Build GRCh38
rs565242192 Chr.16: 2597047-2597047 on Build GRCh38
rs773372279 Chr.16: 2597152-2597152 on Build GRCh38
rs555913218 Chr.16: 2597284-2597284 on Build GRCh38
rs1032697746 Chr.16: 2597218-2597218 on Build GRCh38
rs533525003 Chr.16: 2597376-2597376 on Build GRCh38
rs1042558842 Chr.16: 2597442-2597442 on Build GRCh38
rs73492136 Chr.16: 2597076-2597076 on Build GRCh38
rs1040313092 Chr.16: 2597370-2597370 on Build GRCh38
rs538103841 Chr.16: 2597338-2597338 on Build GRCh38
rs910253610 Chr.16: 2597288-2597288 on Build GRCh38
rs750004515 Chr.16: 2597096-2597096 on Build GRCh38
rs944726197 Chr.16: 2597351-2597351 on Build GRCh38
rs376983918 Chr.16: 2597100-2597100 on Build GRCh38
rs751320393 Chr.16: 2597275-2597275 on Build GRCh38

View Full Product Details