The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016610.3 | NP_057694.2 | 2 - 3 | 224 | 89 |
XM_011545529.1 | XP_011543831.1 | 2 - 3 | 375 | 89 | |
XM_011545530.2 | XP_011543832.1 | 2 - 3 | 203 | 89 | |
GenBank mRNA | AF246971.1 | - | 2 - 3 | 205 | 89 |