Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 22206123-22206628 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EEF2K
Entrez Gene ID
29904
Gene Name
eukaryotic elongation factor 2 kinase
Gene Aliases
HSU93850, eEF-2K
UniGene
Hs.498892
Gene Chromosome Location
Chr.16: 22206264-22288745 on Build GRCh38
Transcripts
NM_013302.3 XM_017023194.1

SNP Detail

SNP ID Chromosome Location
rs1022050952 Chr.16: 22206316-22206316 on Build GRCh38
rs186505381 Chr.16: 22206578-22206578 on Build GRCh38
rs575235001 Chr.16: 22206225-22206225 on Build GRCh38
rs930343487 Chr.16: 22206402-22206402 on Build GRCh38
rs1029372503 Chr.16: 22206524-22206524 on Build GRCh38
rs1037403882 Chr.16: 22206154-22206154 on Build GRCh38
rs1055831941 Chr.16: 22206406-22206406 on Build GRCh38
rs898827317 Chr.16: 22206155-22206155 on Build GRCh38
rs987550359 Chr.16: 22206547-22206547 on Build GRCh38
rs922057487 Chr.16: 22206258-22206258 on Build GRCh38
rs965947727 Chr.16: 22206601-22206601 on Build GRCh38
rs906778289 Chr.16: 22206488-22206488 on Build GRCh38
rs568032810 Chr.16: 22206582-22206582 on Build GRCh38
rs989276628 Chr.16: 22206315-22206315 on Build GRCh38
rs1048849600 Chr.16: 22206589-22206589 on Build GRCh38
rs1015616506 Chr.16: 22206290-22206290 on Build GRCh38
rs564617777 Chr.16: 22206321-22206321 on Build GRCh38
rs975119153 Chr.16: 22206251-22206251 on Build GRCh38
rs940889310 Chr.16: 22206145-22206145 on Build GRCh38
rs978437284 Chr.16: 22206325-22206325 on Build GRCh38
rs895160031 Chr.16: 22206608-22206608 on Build GRCh38
rs774146507 Chr.16: 22206146-22206146 on Build GRCh38
rs978647281 Chr.16: 22206235-22206235 on Build GRCh38
rs529603384 Chr.16: 22206520-22206520 on Build GRCh38
rs903006387 Chr.16: 22206483-22206483 on Build GRCh38
rs866853108 Chr.16: 22206361-22206361 on Build GRCh38
rs944368769 Chr.16: 22206211-22206211 on Build GRCh38
rs940959327 Chr.16: 22206331-22206331 on Build GRCh38
rs967301641 Chr.16: 22206234-22206234 on Build GRCh38
rs895533515 Chr.16: 22206217-22206217 on Build GRCh38
rs759464924 Chr.16: 22206149-22206150 on Build GRCh38
rs1041336945 Chr.16: 22206291-22206302 on Build GRCh38
rs954807035 Chr.16: 22206534-22206534 on Build GRCh38
rs1034118339 Chr.16: 22206207-22206207 on Build GRCh38
rs528085838 Chr.16: 22206342-22206342 on Build GRCh38
rs920860491 Chr.16: 22206581-22206581 on Build GRCh38
rs1003534328 Chr.16: 22206291-22206291 on Build GRCh38
rs1037937810 Chr.16: 22206338-22206338 on Build GRCh38
rs1032782113 Chr.16: 22206239-22206239 on Build GRCh38
rs1019317917 Chr.16: 22206594-22206594 on Build GRCh38
rs966616748 Chr.16: 22206324-22206324 on Build GRCh38
rs1048604931 Chr.16: 22206421-22206421 on Build GRCh38
rs181260490 Chr.16: 22206477-22206477 on Build GRCh38
rs908064439 Chr.16: 22206557-22206557 on Build GRCh38
rs895576785 Chr.16: 22206439-22206439 on Build GRCh38
rs374884794 Chr.16: 22206423-22206423 on Build GRCh38
rs921680749 Chr.16: 22206389-22206389 on Build GRCh38
rs1022317689 Chr.16: 22206558-22206558 on Build GRCh38
rs777789402 Chr.16: 22206499-22206499 on Build GRCh38
rs962190235 Chr.16: 22206569-22206569 on Build GRCh38
rs1004441874 Chr.16: 22206268-22206268 on Build GRCh38
rs370724289 Chr.16: 22206559-22206559 on Build GRCh38
rs945256549 Chr.16: 22206165-22206165 on Build GRCh38
rs987645219 Chr.16: 22206271-22206271 on Build GRCh38
rs148149454 Chr.16: 22206513-22206513 on Build GRCh38
rs1014047017 Chr.16: 22206219-22206219 on Build GRCh38
rs544348875 Chr.16: 22206310-22206310 on Build GRCh38
rs944403588 Chr.16: 22206336-22206336 on Build GRCh38
rs958052927 Chr.16: 22206244-22206244 on Build GRCh38
rs1001713388 Chr.16: 22206193-22206193 on Build GRCh38
rs1013953651 Chr.16: 22206475-22206475 on Build GRCh38
rs933352358 Chr.16: 22206270-22206270 on Build GRCh38
rs533300246 Chr.16: 22206398-22206398 on Build GRCh38
rs908031536 Chr.16: 22206314-22206314 on Build GRCh38
rs888705699 Chr.16: 22206262-22206262 on Build GRCh38
rs937557133 Chr.16: 22206604-22206604 on Build GRCh38
rs977798728 Chr.16: 22206189-22206189 on Build GRCh38
rs1039222532 Chr.16: 22206213-22206213 on Build GRCh38
rs1032943076 Chr.16: 22206505-22206505 on Build GRCh38
rs940186755 Chr.16: 22206476-22206476 on Build GRCh38
rs767328649 Chr.16: 22206242-22206242 on Build GRCh38
rs1048552213 Chr.16: 22206253-22206253 on Build GRCh38
rs977942434 Chr.16: 22206568-22206568 on Build GRCh38

View Full Product Details