Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 22250910-22251404 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EEF2K
Entrez Gene ID
29904
Gene Name
eukaryotic elongation factor 2 kinase
Gene Aliases
HSU93850, eEF-2K
UniGene
Hs.498892
Gene Chromosome Location
Chr.16: 22206264-22288745 on Build GRCh38
Transcripts
NM_013302.3 XM_017023193.1 XM_017023194.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6555511 c.571G>T p.A191S Chr.16: 22251275-22251275 on Build GRCh38
6651801 c.506C>T p.A169V Chr.16: 22251210-22251210 on Build GRCh38
7158582 c.547G>C p.E183Q Chr.16: 22251251-22251251 on Build GRCh38
1376685 c.581delG p.E196fs*22 Chr.16: 22251285-22251285 on Build GRCh38
1376683 c.532C>T p.R178W Chr.16: 22251236-22251236 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765929243 Chr.16: 22251196-22251196 on Build GRCh38
rs140219296 Chr.16: 22251019-22251019 on Build GRCh38
rs752992920 Chr.16: 22251356-22251356 on Build GRCh38
rs947457502 Chr.16: 22251362-22251362 on Build GRCh38
rs947571370 Chr.16: 22251226-22251226 on Build GRCh38
rs146134752 Chr.16: 22251174-22251174 on Build GRCh38
rs767249667 Chr.16: 22251210-22251210 on Build GRCh38
rs144705546 Chr.16: 22251290-22251290 on Build GRCh38
rs375705424 Chr.16: 22251086-22251086 on Build GRCh38
rs760516944 Chr.16: 22251121-22251122 on Build GRCh38
rs767258504 Chr.16: 22251260-22251260 on Build GRCh38
rs756329303 Chr.16: 22251374-22251374 on Build GRCh38
rs997599754 Chr.16: 22251165-22251165 on Build GRCh38
rs139820091 Chr.16: 22251285-22251285 on Build GRCh38
rs777550326 Chr.16: 22251149-22251149 on Build GRCh38
rs370520965 Chr.16: 22251304-22251304 on Build GRCh38
rs758490176 Chr.16: 22251221-22251221 on Build GRCh38
rs766201074 Chr.16: 22251122-22251122 on Build GRCh38
rs371491303 Chr.16: 22251131-22251131 on Build GRCh38
rs749637240 Chr.16: 22251303-22251303 on Build GRCh38
rs2303186 Chr.16: 22251193-22251193 on Build GRCh38
rs1049968414 Chr.16: 22250941-22250941 on Build GRCh38
rs751136394 Chr.16: 22251147-22251147 on Build GRCh38
rs755041930 Chr.16: 22251292-22251292 on Build GRCh38
rs148576611 Chr.16: 22251224-22251224 on Build GRCh38
rs558121142 Chr.16: 22251347-22251347 on Build GRCh38
rs763061605 Chr.16: 22250946-22250946 on Build GRCh38
rs755849618 Chr.16: 22251215-22251215 on Build GRCh38
rs765221591 Chr.16: 22251338-22251338 on Build GRCh38
rs763742980 Chr.16: 22251281-22251281 on Build GRCh38
rs200101051 Chr.16: 22251325-22251325 on Build GRCh38
rs200577173 Chr.16: 22251276-22251276 on Build GRCh38
rs896811690 Chr.16: 22251269-22251269 on Build GRCh38
rs756522929 Chr.16: 22251295-22251295 on Build GRCh38
rs369413078 Chr.16: 22251148-22251148 on Build GRCh38
rs55959085 Chr.16: 22251205-22251205 on Build GRCh38
rs76898301 Chr.16: 22251330-22251330 on Build GRCh38
rs988859957 Chr.16: 22251223-22251223 on Build GRCh38
rs146399588 Chr.16: 22251211-22251211 on Build GRCh38
rs759223201 Chr.16: 22251206-22251206 on Build GRCh38
rs748609190 Chr.16: 22251236-22251236 on Build GRCh38
rs376970159 Chr.16: 22251286-22251286 on Build GRCh38
rs747180465 Chr.16: 22251175-22251175 on Build GRCh38
rs998548918 Chr.16: 22251066-22251066 on Build GRCh38
rs759095840 Chr.16: 22251257-22251257 on Build GRCh38
rs529777108 Chr.16: 22251240-22251240 on Build GRCh38
rs777834802 Chr.16: 22251216-22251216 on Build GRCh38
rs746018935 Chr.16: 22251153-22251153 on Build GRCh38
rs761579842 Chr.16: 22251331-22251331 on Build GRCh38
rs562798620 Chr.16: 22251098-22251098 on Build GRCh38
rs748259123 Chr.16: 22251294-22251294 on Build GRCh38
rs769034403 Chr.16: 22251176-22251176 on Build GRCh38
rs752609933 Chr.16: 22251132-22251132 on Build GRCh38
rs145216064 Chr.16: 22251192-22251192 on Build GRCh38
rs774988075 Chr.16: 22251318-22251318 on Build GRCh38
rs191986675 Chr.16: 22251237-22251237 on Build GRCh38
rs750290664 Chr.16: 22251343-22251343 on Build GRCh38
rs370671646 Chr.16: 22251326-22251326 on Build GRCh38
rs141676543 Chr.16: 22251229-22251229 on Build GRCh38
rs374224871 Chr.16: 22251327-22251327 on Build GRCh38
rs781477493 Chr.16: 22251293-22251293 on Build GRCh38
rs777957758 Chr.16: 22251302-22251302 on Build GRCh38
rs1012103022 Chr.16: 22251323-22251323 on Build GRCh38
rs745438208 Chr.16: 22251322-22251322 on Build GRCh38
rs773781398 Chr.16: 22251238-22251238 on Build GRCh38
rs751189128 Chr.16: 22251128-22251128 on Build GRCh38
rs961177726 Chr.16: 22251130-22251130 on Build GRCh38
rs765438321 Chr.16: 22251134-22251134 on Build GRCh38
rs768912029 Chr.16: 22251230-22251230 on Build GRCh38
rs956050461 Chr.16: 22251339-22251339 on Build GRCh38
rs543207264 Chr.16: 22251089-22251089 on Build GRCh38
rs1043142258 Chr.16: 22251253-22251253 on Build GRCh38
rs369010030 Chr.16: 22251116-22251116 on Build GRCh38
rs904292176 Chr.16: 22251026-22251026 on Build GRCh38
rs746824336 Chr.16: 22251371-22251371 on Build GRCh38
rs377653714 Chr.16: 22251178-22251178 on Build GRCh38
rs764118978 Chr.16: 22250954-22250954 on Build GRCh38
rs142805417 Chr.16: 22251256-22251256 on Build GRCh38
rs151023929 Chr.16: 22251261-22251261 on Build GRCh38
rs1042725413 Chr.16: 22251022-22251022 on Build GRCh38
rs377747666 Chr.16: 22251220-22251220 on Build GRCh38
rs373222313 Chr.16: 22251173-22251173 on Build GRCh38
rs770594193 Chr.16: 22251251-22251251 on Build GRCh38
rs529460713 Chr.16: 22250993-22250993 on Build GRCh38
rs763983728 Chr.16: 22251329-22251329 on Build GRCh38
rs751804990 Chr.16: 22250977-22250977 on Build GRCh38
rs993465042 Chr.16: 22251340-22251340 on Build GRCh38
rs985675433 Chr.16: 22251382-22251382 on Build GRCh38
rs762529883 Chr.16: 22251194-22251194 on Build GRCh38
rs1026854446 Chr.16: 22251137-22251137 on Build GRCh38
rs772138145 Chr.16: 22251158-22251158 on Build GRCh38
rs1019684606 Chr.16: 22250982-22250982 on Build GRCh38

View Full Product Details