The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164269.1 | NP_001157741.1 | 2 - 3 | 230 | 58 |
NM_001164270.1 | NP_001157742.1 | 2 - 3 | 385 | 58 | |
NM_002875.4 | NP_002866.2 | 2 - 3 | 385 | 58 | |
NM_133487.3 | NP_597994.3 | 2 - 3 | 385 | 58 | |
XM_006720626.3 | XP_006720689.1 | 2 - 3 | 436 | 58 | |
XM_011521857.2 | XP_011520159.1 | 2 - 3 | 135 | 58 | |
XM_011521858.2 | XP_011520160.1 | 2 - 3 | 182 | 58 | |
XM_011521859.2 | XP_011520161.1 | 2 - 3 | 319 | 58 | |
XM_011521860.2 | XP_011520162.1 | 2 - 3 | 230 | 58 | |
XM_011521861.2 | XP_011520163.1 | 2 - 3 | 135 | 58 | |
XM_011521862.2 | XP_011520164.1 | 2 - 3 | 353 | 58 | |
GenBank mRNA | AK131299.1 | - | 2 - 3 | 230 | 58 |
AK291969.1 | - | 2 - 3 | 340 | 58 | |
AK303089.1 | - | 2 - 3 | 341 | 58 | |
AK313503.1 | - | 2 - 3 | 340 | 58 | |
BC001459.2 | - | 2 - 3 | 342 | 58 | |
BT019705.1 | - | 1 - 2 | 86 | 58 | |
CR536559.1 | - | 1 - 2 | 86 | 58 | |
D13804.1 | - | 2 - 3 | 297 | 58 | |
D14134.1 | - | 2 - 3 | 318 | 58 | |
EU362635.1 | - | 2 - 3 | 385 | 58 |