The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142315.1 | NP_001135787.1 | 2 - 3 | 383 | 62 |
NM_001142316.1 | NP_001135788.1 | 1 - 2 | 483 | 62 | |
NM_005574.3 | NP_005565.2 | 4 - 5 | 1081 | 62 | |
XM_005252921.2 | XP_005252978.1 | 3 - 4 | 966 | 62 | |
XM_017017727.1 | XP_016873216.1 | 3 - 4 | 734 | 62 | |
XM_017017728.1 | XP_016873217.1 | 5 - 6 | 835 | 62 | |
XM_017017729.1 | XP_016873218.1 | 4 - 5 | 718 | 62 | |
XM_017017730.1 | XP_016873219.1 | 3 - 4 | 900 | 62 | |
XM_017017731.1 | XP_016873220.1 | 4 - 5 | 681 | 62 | |
XM_017017732.1 | XP_016873221.1 | 3 - 4 | 612 | 62 | |
XM_017017733.1 | XP_016873222.1 | 3 - 4 | 519 | 62 | |
GenBank mRNA | BC034041.1 | - | 1 - 2 | 485 | 62 |
BC035607.1 | - | 1 - 2 | 371 | 62 | |
BC042426.1 | - | 1 - 2 | 474 | 62 | |
BM690599.1 | - | 2 - 3 | 284 | 62 | |
X61118.1 | - | 4 - 5 | 1080 | 62 |