The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204126.1 | NP_001191055.1 | 13 - 14 | 1758 | 89 |
NM_001204127.1 | NP_001191056.1 | 13 - 14 | 1688 | 89 | |
NM_006152.3 | NP_006143.2 | 14 - 15 | 1839 | 89 | |
XM_005253370.3 | XP_005253427.1 | 15 - 16 | 1898 | 89 | |
XM_005253372.2 | XP_005253429.1 | 14 - 15 | 1899 | 89 | |
XM_005253374.3 | XP_005253431.1 | 12 - 13 | 1756 | 89 | |
XM_006719076.2 | XP_006719139.1 | 11 - 12 | 822 | 89 | |
XM_011520667.1 | XP_011518969.1 | 15 - 16 | 2047 | 89 | |
XM_011520668.2 | XP_011518970.1 | 11 - 12 | 1085 | 89 | |
XM_011520669.2 | XP_011518971.1 | 10 - 11 | 967 | 89 | |
XM_011520670.1 | XP_011518972.1 | 10 - 11 | 738 | 89 | |
XM_017019300.1 | XP_016874789.1 | 14 - 15 | 1982 | 89 | |
XM_017019301.1 | XP_016874790.1 | 13 - 14 | 1691 | 89 | |
XM_017019302.1 | XP_016874791.1 | 14 - 15 | 1896 | 89 | |
GenBank mRNA | AK303103.1 | - | 10 - 11 | 1091 | 89 |
AK303256.1 | - | 13 - 14 | 1297 | 89 | |
AW408383.1 | - | 4 - 5 | 359 | 89 | |
BC126417.1 | - | 14 - 15 | 1346 | 89 | |
BC144164.1 | - | 13 - 14 | 1265 | 89 | |
BC144165.1 | - | 14 - 15 | 1346 | 89 | |
U10485.1 | - | 14 - 15 | 1487 | 89 |