Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 24939279-24939777 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP17
Entrez Gene ID
55114
Gene Name
Rho GTPase activating protein 17
Gene Aliases
MST066, MST110, MSTP038, MSTP066, MSTP110, NADRIN, PP367, PP4534, RICH-1, RICH1, WBP15
UniGene
Hs.373793
Gene Chromosome Location
Chr.16: 24919384-25015601 on Build GRCh38
Transcripts
NM_001006634.2 NM_018054.5 XM_011545873.2 XM_011545874.2 XM_011545875.2 XM_011545876.2 XM_011545877.2 XM_011545878.2 XM_017023390.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7356121 c.1529G>A p.R510Q Chr.16: 24939559-24939559 on Build GRCh38
4059535 c.1620A>G p.P540P Chr.16: 24939468-24939468 on Build GRCh38
7307902 c.1564G>C p.A522P Chr.16: 24939524-24939524 on Build GRCh38
5562629 c.1573C>A p.P525T Chr.16: 24939515-24939515 on Build GRCh38
4059536 c.1572G>A p.Q524Q Chr.16: 24939516-24939516 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs78457529 Chr.16: 24939559-24939559 on Build GRCh38
rs199828507 Chr.16: 24939482-24939482 on Build GRCh38
rs746145116 Chr.16: 24939477-24939477 on Build GRCh38
rs142115593 Chr.16: 24939485-24939485 on Build GRCh38
rs774531530 Chr.16: 24939369-24939369 on Build GRCh38
rs778549625 Chr.16: 24939320-24939320 on Build GRCh38
rs905163942 Chr.16: 24939329-24939329 on Build GRCh38
rs775962191 Chr.16: 24939428-24939428 on Build GRCh38
rs149887796 Chr.16: 24939381-24939381 on Build GRCh38
rs919254835 Chr.16: 24939486-24939486 on Build GRCh38
rs753579121 Chr.16: 24939628-24939628 on Build GRCh38
rs140685431 Chr.16: 24939514-24939514 on Build GRCh38
rs571309783 Chr.16: 24939468-24939468 on Build GRCh38
rs200153867 Chr.16: 24939644-24939644 on Build GRCh38
rs775587518 Chr.16: 24939325-24939325 on Build GRCh38
rs769155474 Chr.16: 24939489-24939489 on Build GRCh38
rs772182265 Chr.16: 24939323-24939323 on Build GRCh38
rs867238495 Chr.16: 24939318-24939318 on Build GRCh38
rs768561975 Chr.16: 24939404-24939404 on Build GRCh38
rs546945788 Chr.16: 24939563-24939563 on Build GRCh38
rs956589752 Chr.16: 24939616-24939616 on Build GRCh38
rs766081670 Chr.16: 24939604-24939604 on Build GRCh38
rs942036857 Chr.16: 24939330-24939330 on Build GRCh38
rs754276172 Chr.16: 24939430-24939430 on Build GRCh38
rs369123400 Chr.16: 24939501-24939501 on Build GRCh38
rs956942077 Chr.16: 24939448-24939448 on Build GRCh38
rs112450100 Chr.16: 24939577-24939577 on Build GRCh38
rs970896717 Chr.16: 24939471-24939471 on Build GRCh38
rs748665526 Chr.16: 24939560-24939560 on Build GRCh38
rs762104093 Chr.16: 24939433-24939433 on Build GRCh38
rs745320640 Chr.16: 24939400-24939400 on Build GRCh38
rs750101106 Chr.16: 24939447-24939447 on Build GRCh38
rs944440460 Chr.16: 24939714-24939714 on Build GRCh38
rs1052589562 Chr.16: 24939305-24939305 on Build GRCh38
rs781419840 Chr.16: 24939355-24939355 on Build GRCh38
rs746756878 Chr.16: 24939326-24939326 on Build GRCh38
rs777611347 Chr.16: 24939525-24939525 on Build GRCh38
rs781146686 Chr.16: 24939520-24939520 on Build GRCh38
rs752864111 Chr.16: 24939383-24939384 on Build GRCh38
rs749361451 Chr.16: 24939724-24939724 on Build GRCh38
rs376769667 Chr.16: 24939391-24939391 on Build GRCh38
rs201096033 Chr.16: 24939626-24939626 on Build GRCh38
rs567384743 Chr.16: 24939671-24939671 on Build GRCh38
rs773253049 Chr.16: 24939355-24939355 on Build GRCh38
rs755068203 Chr.16: 24939636-24939636 on Build GRCh38
rs750090760 Chr.16: 24939505-24939505 on Build GRCh38
rs1033000716 Chr.16: 24939444-24939444 on Build GRCh38
rs185072394 Chr.16: 24939332-24939332 on Build GRCh38
rs762764341 Chr.16: 24939509-24939509 on Build GRCh38
rs768580330 Chr.16: 24939344-24939344 on Build GRCh38
rs571224945 Chr.16: 24939543-24939543 on Build GRCh38
rs566929405 Chr.16: 24939580-24939580 on Build GRCh38
rs374911769 Chr.16: 24939324-24939325 on Build GRCh38
rs756167651 Chr.16: 24939537-24939540 on Build GRCh38
rs774459430 Chr.16: 24939419-24939419 on Build GRCh38
rs764899218 Chr.16: 24939443-24939443 on Build GRCh38
rs554784020 Chr.16: 24939710-24939710 on Build GRCh38
rs754986220 Chr.16: 24939462-24939462 on Build GRCh38
rs780829391 Chr.16: 24939464-24939464 on Build GRCh38
rs760015909 Chr.16: 24939371-24939371 on Build GRCh38
rs145791095 Chr.16: 24939524-24939524 on Build GRCh38
rs926685406 Chr.16: 24939660-24939660 on Build GRCh38
rs768618730 Chr.16: 24939555-24939555 on Build GRCh38
rs541547174 Chr.16: 24939701-24939701 on Build GRCh38
rs542927086 Chr.16: 24939748-24939748 on Build GRCh38
rs149025741 Chr.16: 24939476-24939476 on Build GRCh38
rs375510367 Chr.16: 24939619-24939619 on Build GRCh38
rs1018008895 Chr.16: 24939414-24939414 on Build GRCh38
rs146682873 Chr.16: 24939504-24939504 on Build GRCh38
rs368132068 Chr.16: 24939451-24939451 on Build GRCh38
rs572415448 Chr.16: 24939689-24939689 on Build GRCh38
rs747068803 Chr.16: 24939554-24939554 on Build GRCh38
rs777033645 Chr.16: 24939496-24939496 on Build GRCh38
rs549344360 Chr.16: 24939479-24939479 on Build GRCh38
rs773997126 Chr.16: 24939692-24939692 on Build GRCh38
rs377481266 Chr.16: 24939537-24939537 on Build GRCh38
rs767964861 Chr.16: 24939372-24939372 on Build GRCh38
rs749631154 Chr.16: 24939664-24939664 on Build GRCh38
rs757072932 Chr.16: 24939630-24939630 on Build GRCh38
rs371175593 Chr.16: 24939633-24939633 on Build GRCh38
rs949456531 Chr.16: 24939317-24939317 on Build GRCh38
rs771428347 Chr.16: 24939684-24939684 on Build GRCh38
rs764333043 Chr.16: 24939429-24939429 on Build GRCh38
rs562576208 Chr.16: 24939743-24939743 on Build GRCh38
rs760868293 Chr.16: 24939461-24939461 on Build GRCh38
rs138492230 Chr.16: 24939532-24939532 on Build GRCh38
rs144901655 Chr.16: 24939405-24939405 on Build GRCh38
rs371811661 Chr.16: 24939388-24939388 on Build GRCh38
rs753427507 Chr.16: 24939382-24939382 on Build GRCh38
rs897507602 Chr.16: 24939754-24939754 on Build GRCh38
rs770475545 Chr.16: 24939650-24939650 on Build GRCh38
rs773755529 Chr.16: 24939575-24939575 on Build GRCh38
rs369553189 Chr.16: 24939368-24939368 on Build GRCh38
rs774180404 Chr.16: 24939607-24939607 on Build GRCh38
rs748983302 Chr.16: 24939315-24939315 on Build GRCh38
rs745759542 Chr.16: 24939322-24939322 on Build GRCh38
rs946856101 Chr.16: 24939650-24939651 on Build GRCh38
rs373850550 Chr.16: 24939409-24939409 on Build GRCh38
rs748662425 Chr.16: 24939647-24939647 on Build GRCh38
rs559129143 Chr.16: 24939661-24939661 on Build GRCh38
rs527665624 Chr.16: 24939402-24939402 on Build GRCh38
rs765958113 Chr.16: 24939513-24939513 on Build GRCh38
rs188795105 Chr.16: 24939632-24939632 on Build GRCh38
rs899243714 Chr.16: 24939530-24939530 on Build GRCh38
rs371724391 Chr.16: 24939359-24939359 on Build GRCh38
rs747929231 Chr.16: 24939465-24939465 on Build GRCh38
rs779773563 Chr.16: 24939458-24939458 on Build GRCh38
rs765002357 Chr.16: 24939410-24939410 on Build GRCh38
rs766950266 Chr.16: 24939613-24939613 on Build GRCh38
rs757927930 Chr.16: 24939455-24939455 on Build GRCh38
rs536547947 Chr.16: 24939617-24939617 on Build GRCh38
rs140540414 Chr.16: 24939725-24939725 on Build GRCh38
rs140774879 Chr.16: 24939403-24939403 on Build GRCh38
rs1040028131 Chr.16: 24939723-24939723 on Build GRCh38
rs752448050 Chr.16: 24939425-24939425 on Build GRCh38
rs370997584 Chr.16: 24939608-24939608 on Build GRCh38
rs1008793041 Chr.16: 24939363-24939363 on Build GRCh38

View Full Product Details