The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001229.4 | NP_001220.2 | 1 - 2 | 378 | 71 |
NM_001278054.1 | NP_001264983.1 | 1 - 2 | 378 | 71 | |
NR_102732.1 | 1 - 2 | 378 | 71 | ||
NR_102733.1 | 1 - 2 | 378 | 71 | ||
XM_011542273.2 | XP_011540575.1 | 1 - 2 | 172 | 71 | |
GenBank mRNA | AB015653.1 | - | 1 - 2 | 154 | 71 |
AB020979.1 | - | 1 - 2 | 133 | 71 | |
AB209147.1 | - | 1 - 2 | 378 | 71 | |
AF093130.1 | - | 1 - 2 | 133 | 71 | |
AF110376.1 | - | 1 - 2 | 133 | 71 | |
AJ781267.1 | - | 1 - 2 | 133 | 71 | |
AK292080.1 | - | 1 - 2 | 150 | 71 | |
AK292111.1 | - | 1 - 2 | 154 | 71 | |
AY732490.2 | - | 1 - 2 | 133 | 71 | |
BC002452.2 | - | 1 - 2 | 139 | 71 | |
BC006463.1 | - | 1 - 2 | 228 | 71 | |
BT006911.1 | - | 1 - 2 | 133 | 71 | |
HY027003.1 | - | 1 - 2 | 172 | 71 | |
U56390.1 | - | 1 - 2 | 136 | 71 | |
U60521.1 | - | 1 - 2 | 178 | 71 |