Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 30069407-30069902 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALDOA
Entrez Gene ID
226
Gene Name
aldolase, fructose-bisphosphate A
Gene Aliases
ALDA, GSD12, HEL-S-87p
UniGene
Hs.513490
Gene Chromosome Location
Chr.16: 30053090-30070420 on Build GRCh38
Transcripts
NM_000034.3 NM_001127617.2 NM_001243177.1 NM_184041.2 NM_184043.2 XM_011545768.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3111675 c.652A>T p.S218C Chr.16: 30069526-30069526 on Build GRCh38
7110025 c.825G>A p.Q275Q Chr.16: 30069855-30069855 on Build GRCh38
6500226 c.736C>T p.H246Y Chr.16: 30069610-30069610 on Build GRCh38
3402264 c.789C>T p.P263P Chr.16: 30069663-30069663 on Build GRCh38
435097 c.825G>C p.Q275H Chr.16: 30069855-30069855 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs113222518 Chr.16: 30069675-30069675 on Build GRCh38
rs746388986 Chr.16: 30069590-30069590 on Build GRCh38
rs140201659 Chr.16: 30069511-30069511 on Build GRCh38
rs199653623 Chr.16: 30069537-30069537 on Build GRCh38
rs763156670 Chr.16: 30069712-30069712 on Build GRCh38
rs200733968 Chr.16: 30069685-30069685 on Build GRCh38
rs780658545 Chr.16: 30069657-30069657 on Build GRCh38
rs879068017 Chr.16: 30069824-30069824 on Build GRCh38
rs367758674 Chr.16: 30069482-30069482 on Build GRCh38
rs748293364 Chr.16: 30069759-30069759 on Build GRCh38
rs575447672 Chr.16: 30069474-30069474 on Build GRCh38
rs563447533 Chr.16: 30069879-30069879 on Build GRCh38
rs189723657 Chr.16: 30069810-30069810 on Build GRCh38
rs992903483 Chr.16: 30069517-30069517 on Build GRCh38
rs750880048 Chr.16: 30069650-30069650 on Build GRCh38
rs878860332 Chr.16: 30069676-30069676 on Build GRCh38
rs763665203 Chr.16: 30069804-30069804 on Build GRCh38
rs774444763 Chr.16: 30069753-30069753 on Build GRCh38
rs201610579 Chr.16: 30069830-30069830 on Build GRCh38
rs768596159 Chr.16: 30069611-30069611 on Build GRCh38
rs141764904 Chr.16: 30069767-30069767 on Build GRCh38
rs75559164 Chr.16: 30069479-30069479 on Build GRCh38
rs759520049 Chr.16: 30069784-30069784 on Build GRCh38
rs752184548 Chr.16: 30069699-30069699 on Build GRCh38
rs189419268 Chr.16: 30069448-30069448 on Build GRCh38
rs372878864 Chr.16: 30069816-30069816 on Build GRCh38
rs752431778 Chr.16: 30069646-30069646 on Build GRCh38
rs752890295 Chr.16: 30069467-30069467 on Build GRCh38
rs766907546 Chr.16: 30069643-30069643 on Build GRCh38
rs780958986 Chr.16: 30069584-30069584 on Build GRCh38
rs550915077 Chr.16: 30069573-30069573 on Build GRCh38
rs377623839 Chr.16: 30069452-30069452 on Build GRCh38
rs3134626 Chr.16: 30069564-30069564 on Build GRCh38
rs560915717 Chr.16: 30069490-30069490 on Build GRCh38
rs763894241 Chr.16: 30069701-30069701 on Build GRCh38
rs770613633 Chr.16: 30069686-30069686 on Build GRCh38
rs368207656 Chr.16: 30069638-30069638 on Build GRCh38
rs182063751 Chr.16: 30069535-30069535 on Build GRCh38
rs775578709 Chr.16: 30069531-30069531 on Build GRCh38
rs761982199 Chr.16: 30069630-30069630 on Build GRCh38
rs1028052840 Chr.16: 30069661-30069661 on Build GRCh38
rs764984222 Chr.16: 30069540-30069540 on Build GRCh38
rs3098416 Chr.16: 30069868-30069868 on Build GRCh38
rs775439610 Chr.16: 30069827-30069828 on Build GRCh38
rs944420954 Chr.16: 30069596-30069596 on Build GRCh38
rs772200646 Chr.16: 30069786-30069786 on Build GRCh38
rs767685109 Chr.16: 30069465-30069465 on Build GRCh38
rs3134627 Chr.16: 30069609-30069609 on Build GRCh38
rs534465340 Chr.16: 30069691-30069691 on Build GRCh38
rs978113250 Chr.16: 30069720-30069720 on Build GRCh38
rs770793827 Chr.16: 30069775-30069775 on Build GRCh38
rs1020363624 Chr.16: 30069654-30069654 on Build GRCh38
rs781754795 Chr.16: 30069477-30069483 on Build GRCh38
rs540258225 Chr.16: 30069429-30069429 on Build GRCh38
rs754442562 Chr.16: 30069574-30069574 on Build GRCh38
rs879043021 Chr.16: 30069825-30069826 on Build GRCh38
rs766939993 Chr.16: 30069801-30069802 on Build GRCh38
rs781198918 Chr.16: 30069664-30069664 on Build GRCh38
rs768071615 Chr.16: 30069823-30069823 on Build GRCh38
rs202079871 Chr.16: 30069856-30069856 on Build GRCh38
rs982947672 Chr.16: 30069761-30069761 on Build GRCh38
rs770093075 Chr.16: 30069587-30069587 on Build GRCh38
rs747160542 Chr.16: 30069457-30069458 on Build GRCh38
rs772297021 Chr.16: 30069525-30069525 on Build GRCh38
rs773418197 Chr.16: 30069679-30069679 on Build GRCh38
rs901107752 Chr.16: 30069871-30069871 on Build GRCh38
rs879200660 Chr.16: 30069496-30069496 on Build GRCh38
rs773837231 Chr.16: 30069705-30069706 on Build GRCh38
rs748384981 Chr.16: 30069669-30069669 on Build GRCh38
rs879203182 Chr.16: 30069820-30069820 on Build GRCh38
rs886051893 Chr.16: 30069622-30069622 on Build GRCh38
rs760900489 Chr.16: 30069468-30069468 on Build GRCh38
rs151069138 Chr.16: 30069647-30069647 on Build GRCh38
rs370909055 Chr.16: 30069478-30069478 on Build GRCh38
rs878939622 Chr.16: 30069495-30069495 on Build GRCh38
rs1040451681 Chr.16: 30069603-30069603 on Build GRCh38
rs1048708464 Chr.16: 30069622-30069622 on Build GRCh38
rs770097934 Chr.16: 30069613-30069615 on Build GRCh38
rs769027218 Chr.16: 30069459-30069460 on Build GRCh38
rs146109120 Chr.16: 30069510-30069510 on Build GRCh38
rs767214133 Chr.16: 30069862-30069862 on Build GRCh38
rs147193284 Chr.16: 30069831-30069831 on Build GRCh38
rs373229343 Chr.16: 30069430-30069430 on Build GRCh38
rs888329113 Chr.16: 30069629-30069629 on Build GRCh38
rs762881835 Chr.16: 30069637-30069637 on Build GRCh38
rs180962387 Chr.16: 30069819-30069819 on Build GRCh38
rs749443540 Chr.16: 30069772-30069772 on Build GRCh38
rs757739728 Chr.16: 30069588-30069588 on Build GRCh38
rs930102805 Chr.16: 30069487-30069487 on Build GRCh38
rs769597382 Chr.16: 30069763-30069763 on Build GRCh38
rs758285007 Chr.16: 30069571-30069571 on Build GRCh38
rs1020311270 Chr.16: 30069433-30069433 on Build GRCh38
rs565013143 Chr.16: 30069864-30069864 on Build GRCh38
rs776950855 Chr.16: 30069617-30069617 on Build GRCh38
rs566755208 Chr.16: 30069690-30069690 on Build GRCh38
rs771967749 Chr.16: 30069863-30069863 on Build GRCh38
rs920112127 Chr.16: 30069484-30069484 on Build GRCh38
rs760307214 Chr.16: 30069803-30069803 on Build GRCh38
rs372148189 Chr.16: 30069493-30069493 on Build GRCh38
rs376977302 Chr.16: 30069723-30069723 on Build GRCh38
rs1002926886 Chr.16: 30069658-30069658 on Build GRCh38
rs573836554 Chr.16: 30069715-30069715 on Build GRCh38
rs760807537 Chr.16: 30069533-30069533 on Build GRCh38
rs760253906 Chr.16: 30069700-30069700 on Build GRCh38
rs879154964 Chr.16: 30069678-30069678 on Build GRCh38
rs148503847 Chr.16: 30069633-30069633 on Build GRCh38
rs779421883 Chr.16: 30069649-30069649 on Build GRCh38
rs148066566 Chr.16: 30069728-30069728 on Build GRCh38
rs779383902 Chr.16: 30069589-30069589 on Build GRCh38
rs11860935 Chr.16: 30069459-30069459 on Build GRCh38
rs113124815 Chr.16: 30069828-30069828 on Build GRCh38
rs780248284 Chr.16: 30069451-30069451 on Build GRCh38
rs779577709 Chr.16: 30069483-30069483 on Build GRCh38
rs11553109 Chr.16: 30069559-30069559 on Build GRCh38
rs749551179 Chr.16: 30069834-30069834 on Build GRCh38
rs752128129 Chr.16: 30069722-30069722 on Build GRCh38
rs763806503 Chr.16: 30069469-30069469 on Build GRCh38
rs769938234 Chr.16: 30069677-30069677 on Build GRCh38
rs773971050 Chr.16: 30069781-30069781 on Build GRCh38
rs748910173 Chr.16: 30069704-30069705 on Build GRCh38
rs546577561 Chr.16: 30069516-30069516 on Build GRCh38
rs758837447 Chr.16: 30069714-30069714 on Build GRCh38
rs142759891 Chr.16: 30069634-30069634 on Build GRCh38
rs111252736 Chr.16: 30069841-30069841 on Build GRCh38
rs532473202 Chr.16: 30069870-30069870 on Build GRCh38
rs775550918 Chr.16: 30069799-30069799 on Build GRCh38
rs374231136 Chr.16: 30069843-30069843 on Build GRCh38
rs375294816 Chr.16: 30069431-30069431 on Build GRCh38
rs375434995 Chr.16: 30069598-30069598 on Build GRCh38
rs773601711 Chr.16: 30069657-30069658 on Build GRCh38
rs986721970 Chr.16: 30069454-30069454 on Build GRCh38
rs900867408 Chr.16: 30069648-30069648 on Build GRCh38
rs577338176 Chr.16: 30069739-30069739 on Build GRCh38
rs754601084 Chr.16: 30069509-30069509 on Build GRCh38
rs776454021 Chr.16: 30069809-30069809 on Build GRCh38
rs751804439 Chr.16: 30069817-30069817 on Build GRCh38
rs775049666 Chr.16: 30069520-30069520 on Build GRCh38
rs201658390 Chr.16: 30069641-30069641 on Build GRCh38
rs746350079 Chr.16: 30069523-30069523 on Build GRCh38
rs767080400 Chr.16: 30069697-30069697 on Build GRCh38
rs368025105 Chr.16: 30069460-30069460 on Build GRCh38
rs747443135 Chr.16: 30069663-30069663 on Build GRCh38
rs748620627 Chr.16: 30069496-30069497 on Build GRCh38
rs142242422 Chr.16: 30069567-30069567 on Build GRCh38
rs772559518 Chr.16: 30069591-30069591 on Build GRCh38
rs879255334 Chr.16: 30069878-30069878 on Build GRCh38
rs759139411 Chr.16: 30069642-30069642 on Build GRCh38
rs563388888 Chr.16: 30069471-30069471 on Build GRCh38
rs770621179 Chr.16: 30069705-30069705 on Build GRCh38
rs144693278 Chr.16: 30069610-30069610 on Build GRCh38
rs764440580 Chr.16: 30069534-30069534 on Build GRCh38
rs186320122 Chr.16: 30069861-30069861 on Build GRCh38
rs753518435 Chr.16: 30069705-30069705 on Build GRCh38

View Full Product Details