Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 30429735-30430229 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCTPP1
Entrez Gene ID
79077
Gene Name
dCTP pyrophosphatase 1
Gene Aliases
CDA03, RS21C6, XTP3TPA
UniGene
Hs.632191
Gene Chromosome Location
Chr.16: 30423698-30430052 on Build GRCh38
Transcripts
NM_024096.1 NR_134470.1 NR_134471.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3112180 c.101T>C p.I34T Chr.16: 30429880-30429880 on Build GRCh38
4793214 c.23T>A p.I8N Chr.16: 30429958-30429958 on Build GRCh38
4802657 c.64C>A p.R22R Chr.16: 30429917-30429917 on Build GRCh38
184995 c.41G>A p.G14E Chr.16: 30429940-30429940 on Build GRCh38
6296654 c.25C>A p.R9S Chr.16: 30429956-30429956 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs748087811 Chr.16: 30429994-30429994 on Build GRCh38
rs576161653 Chr.16: 30429869-30429869 on Build GRCh38
rs906277523 Chr.16: 30430024-30430024 on Build GRCh38
rs180777562 Chr.16: 30429987-30429987 on Build GRCh38
rs1057490586 Chr.16: 30430050-30430050 on Build GRCh38
rs375167447 Chr.16: 30429999-30429999 on Build GRCh38
rs1035824586 Chr.16: 30429818-30429818 on Build GRCh38
rs563861068 Chr.16: 30430025-30430025 on Build GRCh38
rs892015282 Chr.16: 30429791-30429791 on Build GRCh38
rs200319978 Chr.16: 30429964-30429964 on Build GRCh38
rs750072598 Chr.16: 30429900-30429900 on Build GRCh38
rs543674443 Chr.16: 30429943-30429943 on Build GRCh38
rs754320814 Chr.16: 30429976-30429976 on Build GRCh38
rs894624821 Chr.16: 30430056-30430056 on Build GRCh38
rs769498113 Chr.16: 30430006-30430006 on Build GRCh38
rs767784123 Chr.16: 30429966-30429966 on Build GRCh38
rs768666125 Chr.16: 30429945-30429945 on Build GRCh38
rs764625693 Chr.16: 30429970-30429970 on Build GRCh38
rs905850693 Chr.16: 30429843-30429843 on Build GRCh38
rs977028957 Chr.16: 30429941-30429941 on Build GRCh38
rs763517545 Chr.16: 30429885-30429885 on Build GRCh38
rs953900661 Chr.16: 30430188-30430188 on Build GRCh38
rs868011705 Chr.16: 30430164-30430164 on Build GRCh38
rs771089155 Chr.16: 30429833-30429833 on Build GRCh38
rs936129333 Chr.16: 30429783-30429784 on Build GRCh38
rs913399467 Chr.16: 30430034-30430034 on Build GRCh38
rs781406115 Chr.16: 30429893-30429894 on Build GRCh38
rs994635122 Chr.16: 30430193-30430193 on Build GRCh38
rs1013066074 Chr.16: 30430068-30430068 on Build GRCh38
rs746779815 Chr.16: 30429989-30429989 on Build GRCh38
rs756606971 Chr.16: 30429969-30429969 on Build GRCh38
rs904714545 Chr.16: 30429776-30429776 on Build GRCh38
rs369889419 Chr.16: 30430115-30430115 on Build GRCh38
rs749322219 Chr.16: 30429929-30429929 on Build GRCh38
rs549211836 Chr.16: 30430082-30430082 on Build GRCh38
rs369621327 Chr.16: 30429916-30429916 on Build GRCh38
rs139575359 Chr.16: 30429888-30429888 on Build GRCh38
rs768543754 Chr.16: 30429990-30429990 on Build GRCh38
rs185805848 Chr.16: 30430112-30430112 on Build GRCh38
rs749515017 Chr.16: 30429794-30429794 on Build GRCh38
rs778957306 Chr.16: 30429985-30429985 on Build GRCh38
rs965878792 Chr.16: 30430151-30430151 on Build GRCh38
rs775345918 Chr.16: 30429864-30429864 on Build GRCh38
rs774163311 Chr.16: 30429839-30429839 on Build GRCh38
rs769942134 Chr.16: 30429773-30429773 on Build GRCh38
rs915166346 Chr.16: 30430195-30430195 on Build GRCh38
rs1013160637 Chr.16: 30429832-30429832 on Build GRCh38
rs769829600 Chr.16: 30429831-30429831 on Build GRCh38
rs376615500 Chr.16: 30429782-30429782 on Build GRCh38
rs532394112 Chr.16: 30429849-30429860 on Build GRCh38
rs372109503 Chr.16: 30429991-30429991 on Build GRCh38
rs965450062 Chr.16: 30429937-30429937 on Build GRCh38
rs771877218 Chr.16: 30429855-30429855 on Build GRCh38
rs773060140 Chr.16: 30429959-30429959 on Build GRCh38
rs921122118 Chr.16: 30430179-30430179 on Build GRCh38
rs762532977 Chr.16: 30430010-30430010 on Build GRCh38
rs945256726 Chr.16: 30430200-30430200 on Build GRCh38
rs777848541 Chr.16: 30429926-30429926 on Build GRCh38
rs570773277 Chr.16: 30429829-30429829 on Build GRCh38
rs1003292693 Chr.16: 30430030-30430030 on Build GRCh38
rs753342612 Chr.16: 30429887-30429887 on Build GRCh38
rs925661315 Chr.16: 30429989-30429989 on Build GRCh38
rs567522455 Chr.16: 30430083-30430083 on Build GRCh38
rs768763226 Chr.16: 30430065-30430065 on Build GRCh38
rs779154042 Chr.16: 30429936-30429936 on Build GRCh38
rs35384422 Chr.16: 30429823-30429824 on Build GRCh38
rs781610165 Chr.16: 30429917-30429917 on Build GRCh38
rs1038430386 Chr.16: 30430096-30430096 on Build GRCh38
rs531126058 Chr.16: 30430029-30430029 on Build GRCh38
rs748330412 Chr.16: 30429900-30429901 on Build GRCh38
rs761452897 Chr.16: 30429949-30429949 on Build GRCh38
rs202172957 Chr.16: 30429967-30429967 on Build GRCh38
rs201197422 Chr.16: 30429944-30429944 on Build GRCh38
rs201625150 Chr.16: 30429988-30429988 on Build GRCh38
rs759289671 Chr.16: 30429842-30429842 on Build GRCh38
rs1039187724 Chr.16: 30429760-30429760 on Build GRCh38
rs1046390602 Chr.16: 30430055-30430055 on Build GRCh38
rs764820586 Chr.16: 30429957-30429957 on Build GRCh38
rs374301748 Chr.16: 30429984-30429984 on Build GRCh38
rs1015590564 Chr.16: 30430145-30430145 on Build GRCh38
rs982749810 Chr.16: 30429903-30429903 on Build GRCh38
rs753147370 Chr.16: 30429925-30429925 on Build GRCh38
rs1004216245 Chr.16: 30430127-30430127 on Build GRCh38
rs989247125 Chr.16: 30430190-30430190 on Build GRCh38
rs544683617 Chr.16: 30429788-30429788 on Build GRCh38
rs952707983 Chr.16: 30429841-30429841 on Build GRCh38
rs761580953 Chr.16: 30429895-30429895 on Build GRCh38
rs766012185 Chr.16: 30430014-30430014 on Build GRCh38
rs776316015 Chr.16: 30429947-30429947 on Build GRCh38
rs757295928 Chr.16: 30429932-30429932 on Build GRCh38

View Full Product Details